Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 86565827-86566312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCA4
Entrez Gene ID
22802
Gene Name
chloride channel accessory 4
Gene Aliases
CaCC, CaCC2
UniGene
Hs.567422
Gene Chromosome Location
Chr.1: 86547076-86580758 on Build GRCh38
Transcripts
NM_012128.3 NR_024602.1 XM_011541015.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3493072 c.83G>A p.G28E Chr.1: 86566010-86566010 on Build GRCh38
6127960 c.23C>T p.P8L Chr.1: 86565950-86565950 on Build GRCh38
6064616 c.784C>A p.L262I Chr.1: 86565850-86565850 on Build GRCh38
6305840 c.6G>T p.V2V Chr.1: 86565933-86565933 on Build GRCh38
6333879 c.62G>T p.C21F Chr.1: 86565989-86565989 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770126242 Chr.1: 86566039-86566063 on Build GRCh38
rs1029477211 Chr.1: 86566163-86566163 on Build GRCh38
rs753088960 Chr.1: 86566015-86566015 on Build GRCh38
rs776946663 Chr.1: 86565989-86565989 on Build GRCh38
rs2231592 Chr.1: 86565973-86565973 on Build GRCh38
rs754154848 Chr.1: 86565866-86565866 on Build GRCh38
rs755758828 Chr.1: 86566050-86566050 on Build GRCh38
rs377545104 Chr.1: 86565904-86565904 on Build GRCh38
rs1010123598 Chr.1: 86566088-86566088 on Build GRCh38
rs763272861 Chr.1: 86566016-86566016 on Build GRCh38
rs984751333 Chr.1: 86566181-86566181 on Build GRCh38
rs774204047 Chr.1: 86565964-86565964 on Build GRCh38
rs766688618 Chr.1: 86565864-86565864 on Build GRCh38
rs181895259 Chr.1: 86566065-86566065 on Build GRCh38
rs552871030 Chr.1: 86566175-86566175 on Build GRCh38
rs879136520 Chr.1: 86565995-86565995 on Build GRCh38
rs1018884980 Chr.1: 86566121-86566121 on Build GRCh38
rs992347829 Chr.1: 86566223-86566223 on Build GRCh38
rs186153841 Chr.1: 86565886-86565886 on Build GRCh38
rs775266586 Chr.1: 86565928-86565928 on Build GRCh38
rs765638706 Chr.1: 86566231-86566231 on Build GRCh38
rs2231594 Chr.1: 86566087-86566087 on Build GRCh38
rs922057500 Chr.1: 86565897-86565897 on Build GRCh38
rs905642443 Chr.1: 86566036-86566036 on Build GRCh38
rs1031823837 Chr.1: 86566254-86566254 on Build GRCh38
rs965913722 Chr.1: 86565861-86565861 on Build GRCh38
rs756797338 Chr.1: 86565923-86565923 on Build GRCh38
rs759853572 Chr.1: 86565997-86565997 on Build GRCh38
rs943344629 Chr.1: 86566048-86566048 on Build GRCh38
rs746501996 Chr.1: 86565920-86565920 on Build GRCh38
rs753092233 Chr.1: 86565889-86565889 on Build GRCh38
rs146099598 Chr.1: 86566165-86566165 on Build GRCh38
rs758951749 Chr.1: 86565896-86565896 on Build GRCh38
rs934774674 Chr.1: 86566001-86566001 on Build GRCh38
rs370135331 Chr.1: 86566067-86566067 on Build GRCh38
rs899707518 Chr.1: 86566078-86566078 on Build GRCh38
rs376171151 Chr.1: 86566033-86566033 on Build GRCh38
rs77380561 Chr.1: 86565895-86565895 on Build GRCh38
rs952121137 Chr.1: 86565925-86565925 on Build GRCh38
rs569474373 Chr.1: 86566219-86566219 on Build GRCh38
rs112029751 Chr.1: 86566261-86566261 on Build GRCh38
rs755357644 Chr.1: 86565867-86565867 on Build GRCh38
rs750010270 Chr.1: 86566040-86566040 on Build GRCh38
rs760725513 Chr.1: 86565860-86565860 on Build GRCh38
rs762469356 Chr.1: 86566023-86566023 on Build GRCh38
rs765635225 Chr.1: 86566014-86566014 on Build GRCh38
rs992171986 Chr.1: 86565913-86565913 on Build GRCh38
rs756705925 Chr.1: 86566027-86566027 on Build GRCh38
rs79822589 Chr.1: 86565961-86565961 on Build GRCh38
rs2231593 Chr.1: 86566069-86566069 on Build GRCh38
rs200941009 Chr.1: 86565949-86565949 on Build GRCh38
rs962354275 Chr.1: 86565910-86565910 on Build GRCh38
rs996045640 Chr.1: 86566159-86566159 on Build GRCh38
rs766598785 Chr.1: 86565988-86565988 on Build GRCh38
rs914592440 Chr.1: 86566039-86566039 on Build GRCh38
rs551057337 Chr.1: 86566199-86566199 on Build GRCh38
rs190677985 Chr.1: 86566021-86566021 on Build GRCh38
rs772602 Chr.1: 86566247-86566247 on Build GRCh38
rs749103949 Chr.1: 86565952-86565952 on Build GRCh38
rs907849525 Chr.1: 86566197-86566197 on Build GRCh38
rs962034466 Chr.1: 86566214-86566214 on Build GRCh38
rs765721412 Chr.1: 86565882-86565882 on Build GRCh38
rs898179221 Chr.1: 86566196-86566196 on Build GRCh38
rs916137005 Chr.1: 86565916-86565916 on Build GRCh38
rs534475122 Chr.1: 86566291-86566291 on Build GRCh38
rs1016671909 Chr.1: 86566178-86566178 on Build GRCh38
rs971981455 Chr.1: 86565929-86565929 on Build GRCh38
rs908124082 Chr.1: 86565941-86565941 on Build GRCh38
rs1045270683 Chr.1: 86566274-86566274 on Build GRCh38
rs966246627 Chr.1: 86566147-86566147 on Build GRCh38
rs780779596 Chr.1: 86565926-86565926 on Build GRCh38
rs748394443 Chr.1: 86565971-86565974 on Build GRCh38

View Full Product Details