Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161222124-161222701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOA2
Entrez Gene ID
336
Gene Name
apolipoprotein A2
Gene Aliases
Apo-AII, ApoA-II, apoAII
UniGene
Hs.237658
Gene Chromosome Location
Chr.1: 161222293-161223628 on Build GRCh38
Transcripts
NM_001643.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3997018 c.186-4C>T p.? Chr.1: 161222526-161222526 on Build GRCh38
7079301 c.208C>T p.Q70* Chr.1: 161222483-161222483 on Build GRCh38
6508071 c.203C>T p.S68L Chr.1: 161222505-161222505 on Build GRCh38
1491816 c.222C>A p.P74P Chr.1: 161222486-161222486 on Build GRCh38
898538 c.207G>T p.K69N Chr.1: 161222501-161222501 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770351089 Chr.1: 161222252-161222252 on Build GRCh38
rs5088 Chr.1: 161222461-161222461 on Build GRCh38
rs1008701521 Chr.1: 161222626-161222626 on Build GRCh38
rs770336880 Chr.1: 161222398-161222398 on Build GRCh38
rs958816921 Chr.1: 161222657-161222657 on Build GRCh38
rs886108086 Chr.1: 161222183-161222183 on Build GRCh38
rs963076554 Chr.1: 161222214-161222214 on Build GRCh38
rs138954773 Chr.1: 161222437-161222437 on Build GRCh38
rs370320309 Chr.1: 161222212-161222212 on Build GRCh38
rs763419450 Chr.1: 161222318-161222318 on Build GRCh38
rs937394706 Chr.1: 161222396-161222396 on Build GRCh38
rs766546997 Chr.1: 161222284-161222284 on Build GRCh38
rs138452270 Chr.1: 161222465-161222465 on Build GRCh38
rs1014912024 Chr.1: 161222169-161222169 on Build GRCh38
rs773443521 Chr.1: 161222403-161222403 on Build GRCh38
rs775553389 Chr.1: 161222531-161222531 on Build GRCh38
rs200538010 Chr.1: 161222466-161222466 on Build GRCh38
rs987794829 Chr.1: 161222275-161222275 on Build GRCh38
rs371082660 Chr.1: 161222226-161222226 on Build GRCh38
rs757032616 Chr.1: 161222456-161222456 on Build GRCh38
rs759575953 Chr.1: 161222422-161222422 on Build GRCh38
rs774893741 Chr.1: 161222347-161222347 on Build GRCh38
rs768425737 Chr.1: 161222492-161222492 on Build GRCh38
rs76162486 Chr.1: 161222523-161222523 on Build GRCh38
rs781414435 Chr.1: 161222313-161222313 on Build GRCh38
rs6413453 Chr.1: 161222526-161222526 on Build GRCh38
rs754055541 Chr.1: 161222548-161222548 on Build GRCh38
rs750382331 Chr.1: 161222566-161222566 on Build GRCh38
rs773788407 Chr.1: 161222569-161222578 on Build GRCh38
rs755953730 Chr.1: 161222323-161222323 on Build GRCh38
rs867083217 Chr.1: 161222326-161222326 on Build GRCh38
rs748369938 Chr.1: 161222181-161222181 on Build GRCh38
rs1803091 Chr.1: 161222490-161222490 on Build GRCh38
rs80268898 Chr.1: 161222290-161222290 on Build GRCh38
rs36027516 Chr.1: 161222406-161222406 on Build GRCh38
rs778751126 Chr.1: 161222460-161222460 on Build GRCh38
rs896097854 Chr.1: 161222149-161222149 on Build GRCh38
rs371681039 Chr.1: 161222522-161222522 on Build GRCh38
rs369936134 Chr.1: 161222554-161222554 on Build GRCh38
rs756325696 Chr.1: 161222447-161222447 on Build GRCh38
rs775317995 Chr.1: 161222393-161222393 on Build GRCh38
rs760110117 Chr.1: 161222358-161222358 on Build GRCh38
rs76816717 Chr.1: 161222289-161222289 on Build GRCh38
rs940369794 Chr.1: 161222530-161222530 on Build GRCh38
rs775237655 Chr.1: 161222257-161222257 on Build GRCh38
rs745396210 Chr.1: 161222387-161222387 on Build GRCh38
rs763308512 Chr.1: 161222416-161222416 on Build GRCh38
rs773733645 Chr.1: 161222248-161222248 on Build GRCh38
rs116243194 Chr.1: 161222206-161222206 on Build GRCh38
rs777908160 Chr.1: 161222451-161222451 on Build GRCh38
rs1046104966 Chr.1: 161222177-161222177 on Build GRCh38
rs749159200 Chr.1: 161222244-161222244 on Build GRCh38
rs200191201 Chr.1: 161222541-161222541 on Build GRCh38
rs865834629 Chr.1: 161222414-161222415 on Build GRCh38
rs760378915 Chr.1: 161222394-161222394 on Build GRCh38
rs6413454 Chr.1: 161222420-161222420 on Build GRCh38
rs978786211 Chr.1: 161222645-161222645 on Build GRCh38
rs765054986 Chr.1: 161222561-161222561 on Build GRCh38
rs774770928 Chr.1: 161222521-161222521 on Build GRCh38
rs577866247 Chr.1: 161222155-161222155 on Build GRCh38
rs751011813 Chr.1: 161222277-161222277 on Build GRCh38
rs752976925 Chr.1: 161222438-161222438 on Build GRCh38
rs1006991518 Chr.1: 161222399-161222399 on Build GRCh38
rs1029859927 Chr.1: 161222379-161222379 on Build GRCh38
rs767563719 Chr.1: 161222429-161222429 on Build GRCh38
rs779695910 Chr.1: 161222478-161222478 on Build GRCh38
rs538876594 Chr.1: 161222579-161222579 on Build GRCh38
rs765663934 Chr.1: 161222262-161222262 on Build GRCh38
rs531133229 Chr.1: 161222612-161222612 on Build GRCh38
rs750873551 Chr.1: 161222632-161222632 on Build GRCh38
rs12824 Chr.1: 161222467-161222467 on Build GRCh38
rs776287664 Chr.1: 161222498-161222498 on Build GRCh38
rs770532529 Chr.1: 161222246-161222246 on Build GRCh38
rs755025275 Chr.1: 161222292-161222292 on Build GRCh38
rs773597165 Chr.1: 161222273-161222273 on Build GRCh38
rs762296656 Chr.1: 161222261-161222261 on Build GRCh38
rs777531516 Chr.1: 161222345-161222345 on Build GRCh38
rs967793822 Chr.1: 161222340-161222340 on Build GRCh38
rs995992227 Chr.1: 161222231-161222231 on Build GRCh38
rs947332365 Chr.1: 161222638-161222638 on Build GRCh38
rs757039675 Chr.1: 161222560-161222560 on Build GRCh38
rs749636227 Chr.1: 161222513-161222513 on Build GRCh38
rs764978622 Chr.1: 161222462-161222462 on Build GRCh38
rs1033003111 Chr.1: 161222656-161222656 on Build GRCh38
rs760691677 Chr.1: 161222538-161222538 on Build GRCh38
rs75163218 Chr.1: 161222291-161222291 on Build GRCh38
rs5087 Chr.1: 161222674-161222674 on Build GRCh38
rs1028824039 Chr.1: 161222576-161222576 on Build GRCh38
rs767294487 Chr.1: 161222255-161222255 on Build GRCh38
rs935199577 Chr.1: 161222646-161222646 on Build GRCh38
rs191480819 Chr.1: 161222630-161222630 on Build GRCh38
rs771421275 Chr.1: 161222343-161222343 on Build GRCh38
rs749315211 Chr.1: 161222349-161222349 on Build GRCh38
rs763424301 Chr.1: 161222574-161222574 on Build GRCh38
rs751627101 Chr.1: 161222194-161222194 on Build GRCh38
rs753545559 Chr.1: 161222428-161222428 on Build GRCh38
rs6413452 Chr.1: 161222644-161222644 on Build GRCh38
rs890291989 Chr.1: 161222586-161222586 on Build GRCh38
rs895093114 Chr.1: 161222271-161222271 on Build GRCh38
rs771665149 Chr.1: 161222392-161222392 on Build GRCh38
rs763351951 Chr.1: 161222283-161222283 on Build GRCh38
rs766810017 Chr.1: 161222419-161222419 on Build GRCh38
rs368862336 Chr.1: 161222356-161222356 on Build GRCh38
rs890897229 Chr.1: 161222664-161222664 on Build GRCh38
rs771286488 Chr.1: 161222514-161222514 on Build GRCh38

View Full Product Details