Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 35680640-35681164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARPP21
Entrez Gene ID
10777
Gene Name
cAMP regulated phosphoprotein 21
Gene Aliases
ARPP-21, R3HDM3, RCS, TARPP
UniGene
Hs.475902
Gene Chromosome Location
Chr.3: 35638981-35798933 on Build GRCh38
Transcripts
NM_001025068.1 NM_001025069.1 NM_001267616.1 NM_001267617.1 NM_001267618.1 NM_001267619.1 NM_016300.4 NM_198399.1 XM_005264811.2 XM_006712943.2 XM_006712944.2 XM_011533298.2 XM_011533299.2 XM_011533300.2 XM_011533301.2 XM_011533302.2 XM_011533303.2 XM_017005574.1 XM_017005575.1 XM_017005576.1 XM_017005577.1 XM_017005578.1 XM_017005579.1 XM_017005580.1 XM_017005581.1 XM_017005582.1 XM_017005583.1 XM_017005584.1 XM_017005585.1 XM_017005586.1 XM_017005587.1 XM_017005588.1 XM_017005589.1 XM_017005590.1 XM_017005591.1 XM_017005592.1 XM_017005593.1 XM_017005594.1 XM_017005595.1 XM_017005596.1 XM_017005597.1 XM_017005598.1 XM_017005599.1 XM_017005600.1 XM_017005601.1 XM_017005602.1 XM_017005603.1 XM_017005604.1 XM_017005605.1 XM_017005606.1 XM_017005607.1 XM_017005608.1 XM_017005609.1 XM_017005610.1 XM_017005611.1 XM_017005612.1

SNP Detail

SNP ID Chromosome Location
rs1052133841 Chr.3: 35680777-35680777 on Build GRCh38
rs938894525 Chr.3: 35681038-35681038 on Build GRCh38
rs867649449 Chr.3: 35681095-35681095 on Build GRCh38
rs1056307301 Chr.3: 35680893-35680893 on Build GRCh38
rs907814089 Chr.3: 35680804-35680804 on Build GRCh38
rs962066152 Chr.3: 35680952-35680952 on Build GRCh38
rs903474538 Chr.3: 35680988-35680988 on Build GRCh38
rs562688045 Chr.3: 35680811-35680811 on Build GRCh38
rs1023436356 Chr.3: 35680734-35680734 on Build GRCh38
rs966175522 Chr.3: 35681080-35681081 on Build GRCh38
rs1050478285 Chr.3: 35680907-35680907 on Build GRCh38
rs947567512 Chr.3: 35680980-35680980 on Build GRCh38
rs76233610 Chr.3: 35681084-35681084 on Build GRCh38
rs545417415 Chr.3: 35681104-35681104 on Build GRCh38
rs984569887 Chr.3: 35680802-35680802 on Build GRCh38
rs113710044 Chr.3: 35680723-35680725 on Build GRCh38
rs976147085 Chr.3: 35680821-35680821 on Build GRCh38
rs892168373 Chr.3: 35680787-35680787 on Build GRCh38
rs1011918764 Chr.3: 35680708-35680708 on Build GRCh38
rs1014386616 Chr.3: 35680947-35680947 on Build GRCh38
rs914644176 Chr.3: 35680959-35680959 on Build GRCh38
rs766727770 Chr.3: 35681017-35681017 on Build GRCh38
rs951856928 Chr.3: 35680741-35680741 on Build GRCh38
rs1056142588 Chr.3: 35681083-35681083 on Build GRCh38
rs887514919 Chr.3: 35681143-35681143 on Build GRCh38
rs572702385 Chr.3: 35681065-35681065 on Build GRCh38
rs754259551 Chr.3: 35681048-35681048 on Build GRCh38
rs397711772 Chr.3: 35680806-35680806 on Build GRCh38
rs1015008610 Chr.3: 35680950-35680950 on Build GRCh38
rs554473465 Chr.3: 35681027-35681027 on Build GRCh38
rs779202324 Chr.3: 35681096-35681096 on Build GRCh38
rs1042273849 Chr.3: 35680982-35680982 on Build GRCh38
rs960490769 Chr.3: 35680666-35680666 on Build GRCh38
rs184967286 Chr.3: 35681016-35681016 on Build GRCh38
rs781396680 Chr.3: 35680853-35680853 on Build GRCh38
rs912102741 Chr.3: 35681074-35681074 on Build GRCh38
rs1011968070 Chr.3: 35681087-35681087 on Build GRCh38
rs568344125 Chr.3: 35681015-35681015 on Build GRCh38
rs868497599 Chr.3: 35680977-35680977 on Build GRCh38
rs3836256 Chr.3: 35680798-35680798 on Build GRCh38
rs150775807 Chr.3: 35680858-35680858 on Build GRCh38
rs896281116 Chr.3: 35680896-35680896 on Build GRCh38
rs946373656 Chr.3: 35680797-35680798 on Build GRCh38
rs756656784 Chr.3: 35680891-35680891 on Build GRCh38
rs994827185 Chr.3: 35680978-35680978 on Build GRCh38
rs571879255 Chr.3: 35680804-35680807 on Build GRCh38
rs537859163 Chr.3: 35680737-35680737 on Build GRCh38
rs573906381 Chr.3: 35680761-35680761 on Build GRCh38

View Full Product Details