Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50606261-50606832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CISH
Entrez Gene ID
1154
Gene Name
cytokine inducible SH2 containing protein
Gene Aliases
BACTS2, CIS, CIS-1, G18, SOCS
UniGene
Hs.655334
Gene Chromosome Location
Chr.3: 50606454-50611831 on Build GRCh38
Transcripts
NM_013324.5 NM_145071.2 XM_011533329.2

SNP Detail

SNP ID Chromosome Location
rs566745192 Chr.3: 50606390-50606390 on Build GRCh38
rs1020363139 Chr.3: 50606611-50606611 on Build GRCh38
rs142646565 Chr.3: 50606502-50606502 on Build GRCh38
rs903298795 Chr.3: 50606633-50606633 on Build GRCh38
rs571443532 Chr.3: 50606700-50606700 on Build GRCh38
rs770231672 Chr.3: 50606296-50606296 on Build GRCh38
rs759177410 Chr.3: 50606568-50606568 on Build GRCh38
rs972174817 Chr.3: 50606709-50606709 on Build GRCh38
rs568795355 Chr.3: 50606787-50606787 on Build GRCh38
rs893109615 Chr.3: 50606596-50606596 on Build GRCh38
rs151010021 Chr.3: 50606607-50606607 on Build GRCh38
rs553666497 Chr.3: 50606326-50606326 on Build GRCh38
rs541856727 Chr.3: 50606469-50606469 on Build GRCh38
rs968576640 Chr.3: 50606491-50606491 on Build GRCh38
rs140905116 Chr.3: 50606649-50606649 on Build GRCh38
rs1000516669 Chr.3: 50606810-50606810 on Build GRCh38
rs1037579935 Chr.3: 50606357-50606357 on Build GRCh38
rs1030122405 Chr.3: 50606418-50606418 on Build GRCh38
rs1007276970 Chr.3: 50606761-50606761 on Build GRCh38
rs372260191 Chr.3: 50606527-50606527 on Build GRCh38
rs971804680 Chr.3: 50606395-50606395 on Build GRCh38
rs74957911 Chr.3: 50606790-50606790 on Build GRCh38
rs564262631 Chr.3: 50606627-50606627 on Build GRCh38
rs919001220 Chr.3: 50606720-50606720 on Build GRCh38
rs1037289921 Chr.3: 50606748-50606748 on Build GRCh38
rs1038639948 Chr.3: 50606793-50606793 on Build GRCh38
rs554862704 Chr.3: 50606334-50606334 on Build GRCh38
rs1053719253 Chr.3: 50606589-50606589 on Build GRCh38
rs949240932 Chr.3: 50606450-50606450 on Build GRCh38
rs890179534 Chr.3: 50606758-50606758 on Build GRCh38
rs948684941 Chr.3: 50606636-50606636 on Build GRCh38
rs773476245 Chr.3: 50606359-50606359 on Build GRCh38
rs908858313 Chr.3: 50606371-50606371 on Build GRCh38
rs1030940030 Chr.3: 50606663-50606663 on Build GRCh38
rs976694445 Chr.3: 50606310-50606310 on Build GRCh38
rs200195759 Chr.3: 50606323-50606324 on Build GRCh38
rs937836391 Chr.3: 50606794-50606794 on Build GRCh38
rs904199229 Chr.3: 50606797-50606797 on Build GRCh38
rs144186398 Chr.3: 50606704-50606704 on Build GRCh38
rs953656874 Chr.3: 50606333-50606333 on Build GRCh38
rs1046688396 Chr.3: 50606492-50606492 on Build GRCh38
rs771349626 Chr.3: 50606412-50606412 on Build GRCh38
rs28525150 Chr.3: 50606752-50606752 on Build GRCh38
rs980078049 Chr.3: 50606639-50606639 on Build GRCh38
rs753587796 Chr.3: 50606751-50606751 on Build GRCh38
rs955936160 Chr.3: 50606534-50606534 on Build GRCh38
rs1017241669 Chr.3: 50606470-50606470 on Build GRCh38
rs745714780 Chr.3: 50606708-50606708 on Build GRCh38
rs534955927 Chr.3: 50606520-50606524 on Build GRCh38
rs1051938014 Chr.3: 50606756-50606756 on Build GRCh38
rs175634 Chr.3: 50606703-50606703 on Build GRCh38

View Full Product Details