Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 62223689-62224224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GNT2
Entrez Gene ID
10678
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 2
Gene Aliases
3-Gn-T1, 3-Gn-T2, B3GN-T2, B3GNT, B3GNT-2, B3GNT1, BETA3GNT, BGNT2, BGnT-2, beta-1, beta3Gn-T1, beta3Gn-T2
UniGene
Hs.173203
Gene Chromosome Location
Chr.2: 62196127-62224731 on Build GRCh38
Transcripts
NM_001319075.1 NM_006577.5

SNP Detail

SNP ID Chromosome Location
rs1006114525 Chr.2: 62223801-62223801 on Build GRCh38
rs531270418 Chr.2: 62223831-62223831 on Build GRCh38
rs961394526 Chr.2: 62224034-62224034 on Build GRCh38
rs1009984245 Chr.2: 62223739-62223739 on Build GRCh38
rs954593409 Chr.2: 62223919-62223919 on Build GRCh38
rs1046829749 Chr.2: 62224027-62224027 on Build GRCh38
rs923000915 Chr.2: 62223872-62223872 on Build GRCh38
rs910689114 Chr.2: 62223967-62223967 on Build GRCh38
rs763280851 Chr.2: 62224127-62224127 on Build GRCh38
rs906604539 Chr.2: 62223747-62223747 on Build GRCh38
rs1009100087 Chr.2: 62224003-62224003 on Build GRCh38
rs552811124 Chr.2: 62224049-62224049 on Build GRCh38
rs961698313 Chr.2: 62223847-62223847 on Build GRCh38
rs192908054 Chr.2: 62224031-62224031 on Build GRCh38
rs1016049866 Chr.2: 62224032-62224032 on Build GRCh38
rs946621520 Chr.2: 62224174-62224174 on Build GRCh38
rs944373533 Chr.2: 62224192-62224192 on Build GRCh38
rs952736694 Chr.2: 62223788-62223788 on Build GRCh38
rs571295337 Chr.2: 62224095-62224095 on Build GRCh38
rs559763293 Chr.2: 62223716-62223716 on Build GRCh38
rs927822918 Chr.2: 62223903-62223903 on Build GRCh38
rs1053922380 Chr.2: 62223970-62223970 on Build GRCh38
rs761826180 Chr.2: 62224021-62224021 on Build GRCh38
rs977997691 Chr.2: 62224186-62224186 on Build GRCh38
rs11897899 Chr.2: 62223748-62223748 on Build GRCh38
rs936857862 Chr.2: 62223969-62223969 on Build GRCh38
rs77626683 Chr.2: 62224189-62224189 on Build GRCh38
rs1016208468 Chr.2: 62223834-62223834 on Build GRCh38
rs756766802 Chr.2: 62223999-62223999 on Build GRCh38
rs1055228650 Chr.2: 62224061-62224061 on Build GRCh38
rs937848021 Chr.2: 62223917-62223917 on Build GRCh38
rs1016155740 Chr.2: 62224184-62224184 on Build GRCh38
rs764724612 Chr.2: 62224008-62224008 on Build GRCh38
rs185113508 Chr.2: 62223759-62223759 on Build GRCh38
rs1055072776 Chr.2: 62223921-62223921 on Build GRCh38
rs985871580 Chr.2: 62223945-62223945 on Build GRCh38
rs972130612 Chr.2: 62223850-62223850 on Build GRCh38
rs1034168146 Chr.2: 62223757-62223757 on Build GRCh38
rs958557355 Chr.2: 62223797-62223797 on Build GRCh38
rs371596760 Chr.2: 62223843-62223843 on Build GRCh38
rs908522245 Chr.2: 62223804-62223804 on Build GRCh38
rs1002688541 Chr.2: 62224058-62224058 on Build GRCh38
rs543368346 Chr.2: 62224185-62224185 on Build GRCh38
rs190154708 Chr.2: 62224028-62224028 on Build GRCh38
rs765396282 Chr.2: 62224022-62224022 on Build GRCh38
rs150823555 Chr.2: 62223744-62223744 on Build GRCh38
rs923633791 Chr.2: 62224190-62224190 on Build GRCh38
rs199542169 Chr.2: 62223982-62223983 on Build GRCh38
rs150012028 Chr.2: 62223980-62223980 on Build GRCh38
rs894290371 Chr.2: 62224075-62224075 on Build GRCh38
rs564695399 Chr.2: 62224157-62224157 on Build GRCh38
rs771215608 Chr.2: 62223731-62223731 on Build GRCh38
rs915046566 Chr.2: 62224139-62224139 on Build GRCh38
rs915181479 Chr.2: 62223943-62223943 on Build GRCh38
rs982635033 Chr.2: 62224045-62224045 on Build GRCh38
rs892441291 Chr.2: 62223735-62223735 on Build GRCh38
rs928053996 Chr.2: 62224077-62224077 on Build GRCh38

View Full Product Details