Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 30931941-30932438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT14
Entrez Gene ID
79623
Gene Name
polypeptide N-acetylgalactosaminyltransferase 14
Gene Aliases
GALNT15, GalNac-T10, GalNac-T14
UniGene
Hs.468058
Gene Chromosome Location
Chr.2: 30888518-31138726 on Build GRCh38
Transcripts
NM_001253826.1 NM_001253827.1 NM_024572.3 XM_011533105.1 XM_017004906.1 XM_017004907.1 XM_017004908.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
98368 c.976G>A p.V326I Chr.2: 30932150-30932150 on Build GRCh38
2941692 c.962G>A p.G321D Chr.2: 30932164-30932164 on Build GRCh38
6669023 c.943C>T p.R315* Chr.2: 30932183-30932183 on Build GRCh38
4093795 c.1007G>A p.R336Q Chr.2: 30932119-30932119 on Build GRCh38
2941691 c.1008G>A p.R336R Chr.2: 30932118-30932118 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148988791 Chr.2: 30932151-30932151 on Build GRCh38
rs774065276 Chr.2: 30932034-30932034 on Build GRCh38
rs181159970 Chr.2: 30932290-30932290 on Build GRCh38
rs756796892 Chr.2: 30932059-30932059 on Build GRCh38
rs372202464 Chr.2: 30932339-30932339 on Build GRCh38
rs773513395 Chr.2: 30932021-30932021 on Build GRCh38
rs143757721 Chr.2: 30932154-30932154 on Build GRCh38
rs760473532 Chr.2: 30932108-30932108 on Build GRCh38
rs553549873 Chr.2: 30932103-30932103 on Build GRCh38
rs559500240 Chr.2: 30932306-30932306 on Build GRCh38
rs771870260 Chr.2: 30932183-30932183 on Build GRCh38
rs967866720 Chr.2: 30932357-30932357 on Build GRCh38
rs896808975 Chr.2: 30932289-30932289 on Build GRCh38
rs759533365 Chr.2: 30932038-30932038 on Build GRCh38
rs368328840 Chr.2: 30932119-30932119 on Build GRCh38
rs753018629 Chr.2: 30932319-30932319 on Build GRCh38
rs550987390 Chr.2: 30932401-30932401 on Build GRCh38
rs530891022 Chr.2: 30932400-30932400 on Build GRCh38
rs768469194 Chr.2: 30932213-30932213 on Build GRCh38
rs368072213 Chr.2: 30932046-30932046 on Build GRCh38
rs959357920 Chr.2: 30932304-30932304 on Build GRCh38
rs552383139 Chr.2: 30931973-30931973 on Build GRCh38
rs576710368 Chr.2: 30931987-30931987 on Build GRCh38
rs533619083 Chr.2: 30932052-30932052 on Build GRCh38
rs185872317 Chr.2: 30931983-30931983 on Build GRCh38
rs1047468542 Chr.2: 30932011-30932011 on Build GRCh38
rs201271515 Chr.2: 30932077-30932077 on Build GRCh38
rs1032920235 Chr.2: 30932278-30932278 on Build GRCh38
rs568124717 Chr.2: 30932041-30932041 on Build GRCh38
rs751627966 Chr.2: 30932010-30932010 on Build GRCh38
rs376736831 Chr.2: 30932168-30932168 on Build GRCh38
rs532452109 Chr.2: 30932282-30932282 on Build GRCh38
rs369811254 Chr.2: 30932169-30932169 on Build GRCh38
rs976811388 Chr.2: 30932406-30932406 on Build GRCh38
rs769585692 Chr.2: 30932354-30932354 on Build GRCh38
rs867639335 Chr.2: 30932109-30932109 on Build GRCh38
rs753258718 Chr.2: 30932143-30932143 on Build GRCh38
rs746508405 Chr.2: 30932053-30932053 on Build GRCh38
rs755426989 Chr.2: 30932130-30932130 on Build GRCh38
rs776254943 Chr.2: 30932216-30932216 on Build GRCh38
rs867414539 Chr.2: 30932321-30932321 on Build GRCh38
rs922907629 Chr.2: 30932182-30932182 on Build GRCh38
rs537089531 Chr.2: 30932045-30932045 on Build GRCh38
rs573448407 Chr.2: 30932120-30932120 on Build GRCh38
rs1035340752 Chr.2: 30932291-30932291 on Build GRCh38
rs1012581340 Chr.2: 30932330-30932330 on Build GRCh38
rs944453307 Chr.2: 30932253-30932253 on Build GRCh38
rs752251149 Chr.2: 30932044-30932044 on Build GRCh38
rs751959763 Chr.2: 30932074-30932074 on Build GRCh38
rs548229476 Chr.2: 30932001-30932001 on Build GRCh38
rs878931558 Chr.2: 30932283-30932283 on Build GRCh38
rs528229507 Chr.2: 30932317-30932317 on Build GRCh38
rs34910566 Chr.2: 30932124-30932124 on Build GRCh38
rs374361605 Chr.2: 30932102-30932102 on Build GRCh38
rs372818470 Chr.2: 30932076-30932076 on Build GRCh38
rs372673493 Chr.2: 30932396-30932396 on Build GRCh38
rs1036803017 Chr.2: 30932270-30932270 on Build GRCh38
rs771414437 Chr.2: 30932130-30932130 on Build GRCh38
rs748565330 Chr.2: 30932078-30932078 on Build GRCh38
rs539731537 Chr.2: 30932088-30932088 on Build GRCh38
rs968735687 Chr.2: 30932138-30932138 on Build GRCh38
rs796373355 Chr.2: 30932367-30932367 on Build GRCh38
rs767943091 Chr.2: 30932133-30932133 on Build GRCh38
rs959456579 Chr.2: 30932082-30932082 on Build GRCh38
rs766920792 Chr.2: 30932067-30932067 on Build GRCh38
rs771053156 Chr.2: 30932091-30932091 on Build GRCh38
rs756238746 Chr.2: 30932079-30932079 on Build GRCh38
rs900170591 Chr.2: 30932271-30932271 on Build GRCh38
rs376518376 Chr.2: 30932221-30932221 on Build GRCh38
rs139793019 Chr.2: 30932150-30932150 on Build GRCh38
rs202109411 Chr.2: 30932209-30932209 on Build GRCh38
rs372423583 Chr.2: 30932127-30932127 on Build GRCh38
rs1003440487 Chr.2: 30932057-30932057 on Build GRCh38
rs930148073 Chr.2: 30932210-30932210 on Build GRCh38
rs777654707 Chr.2: 30932080-30932080 on Build GRCh38
rs774186098 Chr.2: 30932092-30932092 on Build GRCh38
rs570725104 Chr.2: 30932405-30932405 on Build GRCh38
rs775083709 Chr.2: 30932201-30932201 on Build GRCh38
rs770773384 Chr.2: 30932025-30932025 on Build GRCh38
rs1034437272 Chr.2: 30932316-30932316 on Build GRCh38
rs185747835 Chr.2: 30932391-30932391 on Build GRCh38
rs764961383 Chr.2: 30932126-30932126 on Build GRCh38
rs1038469399 Chr.2: 30931985-30931985 on Build GRCh38
rs1005270771 Chr.2: 30932252-30932252 on Build GRCh38
rs547973457 Chr.2: 30932352-30932352 on Build GRCh38
rs199843526 Chr.2: 30932058-30932058 on Build GRCh38
rs763034516 Chr.2: 30932023-30932023 on Build GRCh38
rs201293423 Chr.2: 30932236-30932236 on Build GRCh38

View Full Product Details