Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 165680075-165680656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSRNP3
Entrez Gene ID
80034
Gene Name
cysteine and serine rich nuclear protein 3
Gene Aliases
FAM130A2, PPP1R73, TAIP-2, TAIP2
UniGene
Hs.470479
Gene Chromosome Location
Chr.2: 165469647-165689407 on Build GRCh38
Transcripts
NM_001172173.1 NM_024969.3 XM_005246865.4

SNP Detail

SNP ID Chromosome Location
rs186607292 Chr.2: 165680264-165680264 on Build GRCh38
rs975935462 Chr.2: 165680489-165680489 on Build GRCh38
rs139145976 Chr.2: 165680502-165680502 on Build GRCh38
rs895452566 Chr.2: 165680447-165680447 on Build GRCh38
rs376913235 Chr.2: 165680618-165680618 on Build GRCh38
rs904391139 Chr.2: 165680408-165680408 on Build GRCh38
rs944057852 Chr.2: 165680242-165680242 on Build GRCh38
rs987329009 Chr.2: 165680402-165680402 on Build GRCh38
rs563517686 Chr.2: 165680109-165680109 on Build GRCh38
rs913316398 Chr.2: 165680315-165680315 on Build GRCh38
rs994577992 Chr.2: 165680609-165680609 on Build GRCh38
rs945862359 Chr.2: 165680378-165680378 on Build GRCh38
rs559701487 Chr.2: 165680267-165680267 on Build GRCh38
rs374517228 Chr.2: 165680611-165680611 on Build GRCh38
rs547881750 Chr.2: 165680550-165680550 on Build GRCh38
rs557585496 Chr.2: 165680120-165680120 on Build GRCh38
rs1042908907 Chr.2: 165680387-165680387 on Build GRCh38
rs560429894 Chr.2: 165680296-165680296 on Build GRCh38
rs1013121518 Chr.2: 165680207-165680207 on Build GRCh38
rs1036236268 Chr.2: 165680169-165680169 on Build GRCh38
rs962097175 Chr.2: 165680504-165680504 on Build GRCh38
rs182261680 Chr.2: 165680188-165680188 on Build GRCh38
rs956121828 Chr.2: 165680379-165680379 on Build GRCh38
rs369436754 Chr.2: 165680373-165680373 on Build GRCh38
rs751765796 Chr.2: 165680442-165680442 on Build GRCh38
rs943212643 Chr.2: 165680463-165680463 on Build GRCh38
rs1055738357 Chr.2: 165680445-165680446 on Build GRCh38
rs1022714725 Chr.2: 165680241-165680241 on Build GRCh38
rs1031612042 Chr.2: 165680362-165680362 on Build GRCh38
rs911674104 Chr.2: 165680460-165680460 on Build GRCh38

View Full Product Details