Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 168714629-168715226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERS6
Entrez Gene ID
253782
Gene Name
ceramide synthase 6
Gene Aliases
CERS5, LASS6
UniGene
Hs.506829
Gene Chromosome Location
Chr.2: 168456249-168775134 on Build GRCh38
Transcripts
NM_001256126.1 NM_203463.2 XM_005246440.4 XM_017003749.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7238058 c.686G>T p.R229L Chr.2: 168715077-168715077 on Build GRCh38
3569581 c.691G>A p.G231R Chr.2: 168715082-168715082 on Build GRCh38
4595784 c.730C>A p.L244I Chr.2: 168715121-168715121 on Build GRCh38
3569582 c.730C>T p.L244F Chr.2: 168715121-168715121 on Build GRCh38
7316832 c.654G>A p.L218L Chr.2: 168715045-168715045 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138089929 Chr.2: 168714956-168714956 on Build GRCh38
rs774014075 Chr.2: 168715079-168715079 on Build GRCh38
rs553664265 Chr.2: 168714720-168714720 on Build GRCh38
rs957091373 Chr.2: 168714756-168714756 on Build GRCh38
rs961502643 Chr.2: 168714662-168714662 on Build GRCh38
rs774724981 Chr.2: 168714745-168714745 on Build GRCh38
rs1000846114 Chr.2: 168714819-168714819 on Build GRCh38
rs904811643 Chr.2: 168714811-168714811 on Build GRCh38
rs909532356 Chr.2: 168714894-168714894 on Build GRCh38
rs989821665 Chr.2: 168714987-168714987 on Build GRCh38
rs781249433 Chr.2: 168715071-168715071 on Build GRCh38
rs779197751 Chr.2: 168714995-168714995 on Build GRCh38
rs759923701 Chr.2: 168714766-168714766 on Build GRCh38
rs561887347 Chr.2: 168714947-168714947 on Build GRCh38
rs747182302 Chr.2: 168715010-168715010 on Build GRCh38
rs368211276 Chr.2: 168715038-168715038 on Build GRCh38
rs909806073 Chr.2: 168714654-168714654 on Build GRCh38
rs116506340 Chr.2: 168715009-168715009 on Build GRCh38
rs953760190 Chr.2: 168714878-168714878 on Build GRCh38
rs985133659 Chr.2: 168714879-168714879 on Build GRCh38
rs372323865 Chr.2: 168715076-168715076 on Build GRCh38
rs755225302 Chr.2: 168715135-168715135 on Build GRCh38
rs577523815 Chr.2: 168714853-168714853 on Build GRCh38
rs923136136 Chr.2: 168714817-168714817 on Build GRCh38
rs536031451 Chr.2: 168714741-168714741 on Build GRCh38
rs941318692 Chr.2: 168714899-168714899 on Build GRCh38
rs61096247 Chr.2: 168714851-168714851 on Build GRCh38
rs540534695 Chr.2: 168714855-168714855 on Build GRCh38
rs529171384 Chr.2: 168714699-168714699 on Build GRCh38
rs764137443 Chr.2: 168715098-168715098 on Build GRCh38
rs913163776 Chr.2: 168714773-168714773 on Build GRCh38
rs749744852 Chr.2: 168715168-168715168 on Build GRCh38
rs116510340 Chr.2: 168715100-168715100 on Build GRCh38
rs760583543 Chr.2: 168715096-168715096 on Build GRCh38
rs748560125 Chr.2: 168715149-168715149 on Build GRCh38
rs1024426286 Chr.2: 168714919-168714919 on Build GRCh38
rs115265576 Chr.2: 168714806-168714806 on Build GRCh38
rs773816652 Chr.2: 168715176-168715176 on Build GRCh38
rs764377085 Chr.2: 168714885-168714885 on Build GRCh38
rs113951892 Chr.2: 168714946-168714946 on Build GRCh38
rs747608813 Chr.2: 168715181-168715181 on Build GRCh38
rs62174433 Chr.2: 168714868-168714868 on Build GRCh38
rs761186044 Chr.2: 168714666-168714666 on Build GRCh38
rs1032356843 Chr.2: 168715108-168715108 on Build GRCh38
rs781470081 Chr.2: 168715145-168715145 on Build GRCh38
rs756044038 Chr.2: 168714981-168714981 on Build GRCh38
rs780954291 Chr.2: 168715077-168715077 on Build GRCh38
rs1050247899 Chr.2: 168715095-168715095 on Build GRCh38
rs375567826 Chr.2: 168715166-168715166 on Build GRCh38
rs146657043 Chr.2: 168715087-168715087 on Build GRCh38
rs780260977 Chr.2: 168715121-168715121 on Build GRCh38
rs768953712 Chr.2: 168715024-168715024 on Build GRCh38
rs770077691 Chr.2: 168715161-168715161 on Build GRCh38
rs542396028 Chr.2: 168714937-168714937 on Build GRCh38
rs758488825 Chr.2: 168715000-168715000 on Build GRCh38
rs939180983 Chr.2: 168714949-168714949 on Build GRCh38
rs781591758 Chr.2: 168715064-168715064 on Build GRCh38
rs1049353123 Chr.2: 168714652-168714652 on Build GRCh38
rs759203411 Chr.2: 168715085-168715085 on Build GRCh38
rs150365971 Chr.2: 168714825-168714825 on Build GRCh38
rs766235835 Chr.2: 168714997-168714997 on Build GRCh38
rs941274437 Chr.2: 168714675-168714675 on Build GRCh38
rs892017039 Chr.2: 168715136-168715136 on Build GRCh38
rs751771710 Chr.2: 168715122-168715122 on Build GRCh38
rs775415372 Chr.2: 168714839-168714839 on Build GRCh38
rs758577445 Chr.2: 168715112-168715112 on Build GRCh38
rs754814456 Chr.2: 168715065-168715065 on Build GRCh38
rs752622547 Chr.2: 168714977-168714977 on Build GRCh38
rs140341827 Chr.2: 168714690-168714690 on Build GRCh38
rs13014488 Chr.2: 168714904-168714904 on Build GRCh38
rs773651193 Chr.2: 168715073-168715073 on Build GRCh38
rs895875243 Chr.2: 168714783-168714783 on Build GRCh38
rs1049865598 Chr.2: 168714840-168714840 on Build GRCh38
rs770321860 Chr.2: 168715066-168715066 on Build GRCh38
rs369910905 Chr.2: 168714965-168714965 on Build GRCh38
rs575286600 Chr.2: 168714809-168714809 on Build GRCh38
rs75359850 Chr.2: 168714734-168714734 on Build GRCh38
rs767291291 Chr.2: 168714971-168714971 on Build GRCh38
rs1038561391 Chr.2: 168714867-168714867 on Build GRCh38
rs985603559 Chr.2: 168715190-168715190 on Build GRCh38

View Full Product Details