Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 112679206-112679772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP11AUN
Entrez Gene ID
400165
Gene Name
ATP11A upstream neighbor
Gene Aliases
C13orf35, SMABLO1
UniGene
Hs.591213
Gene Chromosome Location
Chr.13: 112647044-112684497 on Build GRCh38
Transcripts
NM_207440.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6521247 c.222G>A p.R74R Chr.13: 112679601-112679601 on Build GRCh38
6993081 c.18G>A p.A6A Chr.13: 112679397-112679397 on Build GRCh38
6217777 c.159G>A p.P53P Chr.13: 112679538-112679538 on Build GRCh38
6463071 c.134G>A p.R45H Chr.13: 112679513-112679513 on Build GRCh38
7185517 c.233G>T p.G78V Chr.13: 112679612-112679612 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757840603 Chr.13: 112679658-112679658 on Build GRCh38
rs1040082031 Chr.13: 112679245-112679245 on Build GRCh38
rs7338610 Chr.13: 112679370-112679370 on Build GRCh38
rs1047451651 Chr.13: 112679234-112679234 on Build GRCh38
rs549923457 Chr.13: 112679306-112679306 on Build GRCh38
rs73574563 Chr.13: 112679415-112679415 on Build GRCh38
rs200954527 Chr.13: 112679564-112679564 on Build GRCh38
rs267603760 Chr.13: 112679458-112679458 on Build GRCh38
rs778304928 Chr.13: 112679510-112679521 on Build GRCh38
rs750843436 Chr.13: 112679390-112679390 on Build GRCh38
rs759534437 Chr.13: 112679350-112679350 on Build GRCh38
rs934042463 Chr.13: 112679502-112679502 on Build GRCh38
rs759287684 Chr.13: 112679425-112679425 on Build GRCh38
rs532070406 Chr.13: 112679278-112679278 on Build GRCh38
rs756425294 Chr.13: 112679508-112679508 on Build GRCh38
rs941735164 Chr.13: 112679446-112679446 on Build GRCh38
rs939950123 Chr.13: 112679530-112679530 on Build GRCh38
rs780924753 Chr.13: 112679671-112679671 on Build GRCh38
rs891796733 Chr.13: 112679292-112679292 on Build GRCh38
rs1030370619 Chr.13: 112679290-112679290 on Build GRCh38
rs751191674 Chr.13: 112679660-112679660 on Build GRCh38
rs764666222 Chr.13: 112679645-112679645 on Build GRCh38
rs745471588 Chr.13: 112679509-112679509 on Build GRCh38
rs765512223 Chr.13: 112679460-112679460 on Build GRCh38
rs777382327 Chr.13: 112679406-112679406 on Build GRCh38
rs752645498 Chr.13: 112679359-112679359 on Build GRCh38
rs974893329 Chr.13: 112679729-112679729 on Build GRCh38
rs117237810 Chr.13: 112679479-112679479 on Build GRCh38
rs201453616 Chr.13: 112679382-112679382 on Build GRCh38
rs929097235 Chr.13: 112679232-112679232 on Build GRCh38
rs747680883 Chr.13: 112679330-112679330 on Build GRCh38
rs779204611 Chr.13: 112679286-112679286 on Build GRCh38
rs891830358 Chr.13: 112679500-112679500 on Build GRCh38
rs750628088 Chr.13: 112679463-112679463 on Build GRCh38
rs200943748 Chr.13: 112679512-112679512 on Build GRCh38
rs1038142484 Chr.13: 112679533-112679533 on Build GRCh38
rs773048164 Chr.13: 112679339-112679339 on Build GRCh38
rs369042046 Chr.13: 112679396-112679396 on Build GRCh38
rs761923080 Chr.13: 112679536-112679536 on Build GRCh38
rs769716880 Chr.13: 112679689-112679689 on Build GRCh38
rs767941308 Chr.13: 112679621-112679621 on Build GRCh38
rs114091399 Chr.13: 112679495-112679495 on Build GRCh38
rs201512112 Chr.13: 112679489-112679489 on Build GRCh38
rs959182914 Chr.13: 112679586-112679586 on Build GRCh38
rs773329231 Chr.13: 112679542-112679542 on Build GRCh38
rs116657983 Chr.13: 112679443-112679443 on Build GRCh38
rs745508072 Chr.13: 112679559-112679559 on Build GRCh38
rs144955786 Chr.13: 112679535-112679535 on Build GRCh38
rs771024276 Chr.13: 112679327-112679327 on Build GRCh38
rs761865865 Chr.13: 112679459-112679459 on Build GRCh38
rs754791551 Chr.13: 112679668-112679668 on Build GRCh38
rs201364555 Chr.13: 112679556-112679556 on Build GRCh38
rs192073132 Chr.13: 112679363-112679363 on Build GRCh38
rs1024863286 Chr.13: 112679624-112679624 on Build GRCh38
rs780467695 Chr.13: 112679397-112679397 on Build GRCh38
rs979292972 Chr.13: 112679362-112679362 on Build GRCh38
rs147355248 Chr.13: 112679275-112679275 on Build GRCh38
rs941821217 Chr.13: 112679727-112679727 on Build GRCh38
rs765280632 Chr.13: 112679537-112679537 on Build GRCh38
rs566038787 Chr.13: 112679742-112679742 on Build GRCh38
rs769211198 Chr.13: 112679402-112679402 on Build GRCh38
rs527274699 Chr.13: 112679259-112679259 on Build GRCh38
rs774224491 Chr.13: 112679343-112679343 on Build GRCh38
rs763196221 Chr.13: 112679543-112679543 on Build GRCh38
rs917577527 Chr.13: 112679229-112679229 on Build GRCh38
rs760024974 Chr.13: 112679610-112679610 on Build GRCh38
rs1022006151 Chr.13: 112679534-112679534 on Build GRCh38
rs761194667 Chr.13: 112679640-112679640 on Build GRCh38
rs141230308 Chr.13: 112679506-112679506 on Build GRCh38
rs917606966 Chr.13: 112679407-112679407 on Build GRCh38
rs773826640 Chr.13: 112679726-112679726 on Build GRCh38
rs564116015 Chr.13: 112679527-112679527 on Build GRCh38
rs539964531 Chr.13: 112679753-112679753 on Build GRCh38
rs369426969 Chr.13: 112679477-112679477 on Build GRCh38
rs753865659 Chr.13: 112679244-112679244 on Build GRCh38
rs746932458 Chr.13: 112679524-112679524 on Build GRCh38
rs769317234 Chr.13: 112679337-112679337 on Build GRCh38
rs200914561 Chr.13: 112679597-112679597 on Build GRCh38
rs868395798 Chr.13: 112679559-112679564 on Build GRCh38
rs560155493 Chr.13: 112679250-112679250 on Build GRCh38
rs896136324 Chr.13: 112679594-112679594 on Build GRCh38
rs556365416 Chr.13: 112679456-112679456 on Build GRCh38
rs373171726 Chr.13: 112679483-112679483 on Build GRCh38
rs74446403 Chr.13: 112679322-112679322 on Build GRCh38
rs1033557756 Chr.13: 112679576-112679576 on Build GRCh38
rs777562625 Chr.13: 112679690-112679690 on Build GRCh38
rs746886400 Chr.13: 112679563-112679563 on Build GRCh38
rs539429434 Chr.13: 112679383-112679383 on Build GRCh38
rs752041784 Chr.13: 112679470-112679470 on Build GRCh38
rs200305243 Chr.13: 112679412-112679412 on Build GRCh38
rs192007935 Chr.13: 112679560-112679560 on Build GRCh38
rs1010346335 Chr.13: 112679294-112679294 on Build GRCh38
rs866895795 Chr.13: 112679580-112679580 on Build GRCh38
rs762649705 Chr.13: 112679340-112679340 on Build GRCh38
rs367674877 Chr.13: 112679659-112679659 on Build GRCh38
rs150373081 Chr.13: 112679469-112679469 on Build GRCh38
rs925986882 Chr.13: 112679236-112679236 on Build GRCh38
rs138886345 Chr.13: 112679596-112679596 on Build GRCh38
rs767418384 Chr.13: 112679358-112679358 on Build GRCh38
rs754314917 Chr.13: 112679647-112679647 on Build GRCh38
rs753284014 Chr.13: 112679629-112679629 on Build GRCh38
rs993478947 Chr.13: 112679400-112679400 on Build GRCh38
rs977688628 Chr.13: 112679655-112679655 on Build GRCh38
rs769893730 Chr.13: 112679593-112679593 on Build GRCh38
rs373831748 Chr.13: 112679648-112679648 on Build GRCh38
rs778442948 Chr.13: 112679558-112679558 on Build GRCh38
rs748029542 Chr.13: 112679589-112679589 on Build GRCh38
rs774434934 Chr.13: 112679607-112679607 on Build GRCh38
rs770653326 Chr.13: 112679422-112679422 on Build GRCh38
rs983139460 Chr.13: 112679709-112679709 on Build GRCh38
rs997732818 Chr.13: 112679281-112679281 on Build GRCh38
rs908835542 Chr.13: 112679723-112679723 on Build GRCh38
rs201611037 Chr.13: 112679687-112679687 on Build GRCh38
rs754089948 Chr.13: 112679378-112679378 on Build GRCh38
rs372159514 Chr.13: 112679423-112679423 on Build GRCh38
rs1033743228 Chr.13: 112679354-112679354 on Build GRCh38
rs747530091 Chr.13: 112679401-112679401 on Build GRCh38
rs139762375 Chr.13: 112679740-112679740 on Build GRCh38
rs775288269 Chr.13: 112679515-112679515 on Build GRCh38
rs751897797 Chr.13: 112679544-112679544 on Build GRCh38
rs151189351 Chr.13: 112679417-112679417 on Build GRCh38
rs199805592 Chr.13: 112679467-112679467 on Build GRCh38
rs753176662 Chr.13: 112679370-112679370 on Build GRCh38
rs755384113 Chr.13: 112679553-112679553 on Build GRCh38
rs771875325 Chr.13: 112679429-112679429 on Build GRCh38
rs571051739 Chr.13: 112679276-112679276 on Build GRCh38
rs995647656 Chr.13: 112679720-112679720 on Build GRCh38
rs758739806 Chr.13: 112679468-112679468 on Build GRCh38
rs768208765 Chr.13: 112679555-112679555 on Build GRCh38
rs375489837 Chr.13: 112679434-112679434 on Build GRCh38

View Full Product Details