Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103559119-103559668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG5
Entrez Gene ID
9529
Gene Name
BCL2 associated athanogene 5
Gene Aliases
BAG-5
UniGene
Hs.5443
Gene Chromosome Location
Chr.14: 103556544-103562814 on Build GRCh38
Transcripts
NM_001015048.2 NM_001015049.2 NM_004873.3

SNP Detail

SNP ID Chromosome Location
rs76189229 Chr.14: 103559349-103559349 on Build GRCh38
rs914648111 Chr.14: 103559628-103559628 on Build GRCh38
rs759734713 Chr.14: 103559429-103559429 on Build GRCh38
rs753356882 Chr.14: 103559643-103559643 on Build GRCh38
rs1047678272 Chr.14: 103559631-103559631 on Build GRCh38
rs996255516 Chr.14: 103559510-103559510 on Build GRCh38
rs544136238 Chr.14: 103559436-103559436 on Build GRCh38
rs1023256944 Chr.14: 103559497-103559497 on Build GRCh38
rs1005808489 Chr.14: 103559192-103559192 on Build GRCh38
rs572584714 Chr.14: 103559348-103559348 on Build GRCh38
rs774608905 Chr.14: 103559329-103559329 on Build GRCh38
rs938490537 Chr.14: 103559414-103559414 on Build GRCh38
rs766408838 Chr.14: 103559319-103559319 on Build GRCh38
rs529821986 Chr.14: 103559460-103559460 on Build GRCh38
rs187913611 Chr.14: 103559591-103559591 on Build GRCh38
rs934755166 Chr.14: 103559279-103559279 on Build GRCh38
rs530751868 Chr.14: 103559373-103559373 on Build GRCh38
rs1054442293 Chr.14: 103559315-103559315 on Build GRCh38
rs867084786 Chr.14: 103559554-103559554 on Build GRCh38
rs551841163 Chr.14: 103559593-103559593 on Build GRCh38
rs1047563097 Chr.14: 103559420-103559421 on Build GRCh38
rs886309759 Chr.14: 103559441-103559442 on Build GRCh38
rs991763772 Chr.14: 103559222-103559222 on Build GRCh38
rs770188687 Chr.14: 103559193-103559193 on Build GRCh38
rs567019925 Chr.14: 103559511-103559511 on Build GRCh38
rs139246381 Chr.14: 103559372-103559372 on Build GRCh38
rs924765478 Chr.14: 103559234-103559234 on Build GRCh38
rs542731960 Chr.14: 103559261-103559261 on Build GRCh38
rs1037438037 Chr.14: 103559455-103559455 on Build GRCh38
rs1029019552 Chr.14: 103559262-103559262 on Build GRCh38
rs552761945 Chr.14: 103559294-103559294 on Build GRCh38
rs1025997698 Chr.14: 103559182-103559182 on Build GRCh38
rs764716392 Chr.14: 103559606-103559606 on Build GRCh38
rs990213962 Chr.14: 103559584-103559584 on Build GRCh38
rs537439153 Chr.14: 103559272-103559272 on Build GRCh38
rs970061652 Chr.14: 103559603-103559605 on Build GRCh38
rs377404109 Chr.14: 103559583-103559583 on Build GRCh38
rs566205967 Chr.14: 103559573-103559573 on Build GRCh38
rs1040617486 Chr.14: 103559405-103559405 on Build GRCh38
rs561139553 Chr.14: 103559450-103559450 on Build GRCh38
rs1050396374 Chr.14: 103559350-103559350 on Build GRCh38
rs971624429 Chr.14: 103559228-103559228 on Build GRCh38
rs897631719 Chr.14: 103559407-103559407 on Build GRCh38
rs1012989947 Chr.14: 103559484-103559484 on Build GRCh38
rs900154659 Chr.14: 103559477-103559477 on Build GRCh38
rs956186075 Chr.14: 103559574-103559574 on Build GRCh38
rs972060145 Chr.14: 103559518-103559518 on Build GRCh38
rs1028549528 Chr.14: 103559553-103559554 on Build GRCh38
rs980514144 Chr.14: 103559613-103559613 on Build GRCh38
rs574106714 Chr.14: 103559227-103559227 on Build GRCh38
rs987390748 Chr.14: 103559311-103559311 on Build GRCh38
rs981684375 Chr.14: 103559544-103559544 on Build GRCh38
rs1044292328 Chr.14: 103559401-103559406 on Build GRCh38
rs956232438 Chr.14: 103559575-103559576 on Build GRCh38
rs35498576 Chr.14: 103559556-103559556 on Build GRCh38
rs955537394 Chr.14: 103559299-103559299 on Build GRCh38
rs948752860 Chr.14: 103559383-103559383 on Build GRCh38
rs955813970 Chr.14: 103559553-103559557 on Build GRCh38
rs982015128 Chr.14: 103559230-103559230 on Build GRCh38
rs887470113 Chr.14: 103559464-103559464 on Build GRCh38
rs886191800 Chr.14: 103559187-103559189 on Build GRCh38

View Full Product Details