Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 82990699-82991214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf40
Entrez Gene ID
123207
Gene Name
chromosome 15 open reading frame 40
Gene Aliases
-
UniGene
Hs.352541
Gene Chromosome Location
Chr.15: 82988297-83012298 on Build GRCh38
Transcripts
NM_001160115.1 NM_001160116.1 XM_011521213.2 NM_031452.3

Gene Symbol
FAM103A1
Entrez Gene ID
83640
Gene Name
family with sequence similarity 103 member A1
Gene Aliases
C15orf18, HsT19360, RAM
UniGene
Hs.80624
Gene Chromosome Location
Chr.15: 82986203-82991057 on Build GRCh38
Transcripts
NM_001160115.1 NM_001160116.1 XM_011521213.2 NM_031452.3

SNP Detail

SNP ID Chromosome Location
rs971323055 Chr.15: 82991043-82991043 on Build GRCh38
rs530283938 Chr.15: 82990785-82990785 on Build GRCh38
rs979780098 Chr.15: 82991065-82991065 on Build GRCh38
rs371975154 Chr.15: 82990788-82990788 on Build GRCh38
rs553759390 Chr.15: 82991086-82991086 on Build GRCh38
rs777090530 Chr.15: 82990879-82990879 on Build GRCh38
rs552197394 Chr.15: 82991061-82991061 on Build GRCh38
rs748250172 Chr.15: 82990958-82990958 on Build GRCh38
rs773489171 Chr.15: 82990997-82990999 on Build GRCh38
rs554777203 Chr.15: 82991126-82991126 on Build GRCh38
rs891385950 Chr.15: 82990838-82990838 on Build GRCh38
rs565820007 Chr.15: 82991085-82991085 on Build GRCh38
rs962424376 Chr.15: 82991113-82991113 on Build GRCh38
rs930140650 Chr.15: 82990786-82990786 on Build GRCh38
rs1040142071 Chr.15: 82990920-82990920 on Build GRCh38
rs997232675 Chr.15: 82991107-82991107 on Build GRCh38
rs1024174656 Chr.15: 82991026-82991026 on Build GRCh38
rs1047850021 Chr.15: 82991130-82991130 on Build GRCh38
rs901585177 Chr.15: 82991106-82991106 on Build GRCh38
rs891599253 Chr.15: 82990969-82990969 on Build GRCh38
rs1010029305 Chr.15: 82990984-82990984 on Build GRCh38
rs913194011 Chr.15: 82990935-82990938 on Build GRCh38
rs988781387 Chr.15: 82990915-82990915 on Build GRCh38
rs911409109 Chr.15: 82991008-82991008 on Build GRCh38
rs550220601 Chr.15: 82990866-82990866 on Build GRCh38
rs950699482 Chr.15: 82990772-82990772 on Build GRCh38
rs903922998 Chr.15: 82990940-82990940 on Build GRCh38
rs570286700 Chr.15: 82990899-82990899 on Build GRCh38
rs942827582 Chr.15: 82991064-82991064 on Build GRCh38
rs947647155 Chr.15: 82990935-82990935 on Build GRCh38
rs1031100587 Chr.15: 82990964-82990964 on Build GRCh38
rs1026263297 Chr.15: 82990725-82990725 on Build GRCh38
rs928723543 Chr.15: 82990889-82990889 on Build GRCh38
rs781299819 Chr.15: 82991175-82991175 on Build GRCh38
rs974249996 Chr.15: 82990774-82990774 on Build GRCh38
rs374287215 Chr.15: 82990942-82990942 on Build GRCh38
rs561311740 Chr.15: 82990743-82990743 on Build GRCh38
rs886079503 Chr.15: 82991158-82991158 on Build GRCh38

View Full Product Details