Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90617493-90618065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTC3
Entrez Gene ID
64784
Gene Name
CREB regulated transcription coactivator 3
Gene Aliases
TORC-3, TORC3
UniGene
Hs.567572
Gene Chromosome Location
Chr.15: 90529886-90645345 on Build GRCh38
Transcripts
NM_001042574.2 NM_022769.4 XM_005254968.2 XM_011521906.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1708548 c.631C>T p.P211S Chr.15: 90617900-90617900 on Build GRCh38
966511 c.662C>T p.P221L Chr.15: 90617931-90617931 on Build GRCh38
4822972 c.646A>G p.N216D Chr.15: 90617915-90617915 on Build GRCh38
7248319 c.663G>A p.P221P Chr.15: 90617932-90617932 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs575049896 Chr.15: 90617841-90617841 on Build GRCh38
rs141737123 Chr.15: 90617769-90617769 on Build GRCh38
rs368205917 Chr.15: 90617997-90617997 on Build GRCh38
rs769978211 Chr.15: 90617866-90617866 on Build GRCh38
rs952786287 Chr.15: 90617561-90617561 on Build GRCh38
rs1014704840 Chr.15: 90617679-90617679 on Build GRCh38
rs180843251 Chr.15: 90617663-90617663 on Build GRCh38
rs535194864 Chr.15: 90617768-90617768 on Build GRCh38
rs4932357 Chr.15: 90617652-90617652 on Build GRCh38
rs979058533 Chr.15: 90617740-90617740 on Build GRCh38
rs764874770 Chr.15: 90617903-90617903 on Build GRCh38
rs749351359 Chr.15: 90617972-90617972 on Build GRCh38
rs976904329 Chr.15: 90617597-90617597 on Build GRCh38
rs751055533 Chr.15: 90617866-90617866 on Build GRCh38
rs546302908 Chr.15: 90617596-90617596 on Build GRCh38
rs968828155 Chr.15: 90617535-90617535 on Build GRCh38
rs773174310 Chr.15: 90617995-90617995 on Build GRCh38
rs750211393 Chr.15: 90617895-90617895 on Build GRCh38
rs980919018 Chr.15: 90617813-90617813 on Build GRCh38
rs763443566 Chr.15: 90617777-90617777 on Build GRCh38
rs944294717 Chr.15: 90617594-90617594 on Build GRCh38
rs911451284 Chr.15: 90617593-90617593 on Build GRCh38
rs530567159 Chr.15: 90617786-90617786 on Build GRCh38
rs142491811 Chr.15: 90617902-90617902 on Build GRCh38
rs200082382 Chr.15: 90617884-90617884 on Build GRCh38
rs540129338 Chr.15: 90617865-90617866 on Build GRCh38
rs543085250 Chr.15: 90617950-90617950 on Build GRCh38
rs965611006 Chr.15: 90617751-90617751 on Build GRCh38
rs924140349 Chr.15: 90617607-90617607 on Build GRCh38
rs1027454558 Chr.15: 90617718-90617718 on Build GRCh38
rs746087617 Chr.15: 90617850-90617850 on Build GRCh38
rs776413991 Chr.15: 90618003-90618003 on Build GRCh38
rs771397244 Chr.15: 90617846-90617846 on Build GRCh38
rs766158496 Chr.15: 90617897-90617897 on Build GRCh38
rs994685582 Chr.15: 90617682-90617682 on Build GRCh38
rs890321699 Chr.15: 90617713-90617713 on Build GRCh38
rs1030338892 Chr.15: 90617703-90617703 on Build GRCh38
rs935092314 Chr.15: 90617614-90617614 on Build GRCh38
rs1044699819 Chr.15: 90617828-90617828 on Build GRCh38
rs952843683 Chr.15: 90617728-90617728 on Build GRCh38
rs760512655 Chr.15: 90617896-90617896 on Build GRCh38
rs148259578 Chr.15: 90617629-90617629 on Build GRCh38
rs757873752 Chr.15: 90617913-90617913 on Build GRCh38
rs144818109 Chr.15: 90617959-90617959 on Build GRCh38
rs550099752 Chr.15: 90617653-90617653 on Build GRCh38
rs1037719576 Chr.15: 90617671-90617671 on Build GRCh38
rs935464061 Chr.15: 90617778-90617778 on Build GRCh38
rs376557472 Chr.15: 90617681-90617681 on Build GRCh38
rs778300144 Chr.15: 90617838-90617838 on Build GRCh38
rs575646187 Chr.15: 90617826-90617826 on Build GRCh38
rs1018285557 Chr.15: 90618027-90618027 on Build GRCh38
rs985846172 Chr.15: 90617730-90617730 on Build GRCh38
rs752176911 Chr.15: 90617911-90617911 on Build GRCh38
rs111569950 Chr.15: 90617764-90617765 on Build GRCh38
rs116946190 Chr.15: 90617821-90617821 on Build GRCh38
rs1048908529 Chr.15: 90617971-90617971 on Build GRCh38
rs77748519 Chr.15: 90617932-90617932 on Build GRCh38
rs947683440 Chr.15: 90617648-90617648 on Build GRCh38
rs551061685 Chr.15: 90617789-90617789 on Build GRCh38
rs547339285 Chr.15: 90617719-90617719 on Build GRCh38
rs924385106 Chr.15: 90617566-90617566 on Build GRCh38
rs374979539 Chr.15: 90618002-90618002 on Build GRCh38
rs370846835 Chr.15: 90617912-90617912 on Build GRCh38
rs755295040 Chr.15: 90617980-90617980 on Build GRCh38
rs146224437 Chr.15: 90617939-90617939 on Build GRCh38
rs906245048 Chr.15: 90617877-90617877 on Build GRCh38
rs973265275 Chr.15: 90617532-90617532 on Build GRCh38
rs139851694 Chr.15: 90617945-90617945 on Build GRCh38
rs565525756 Chr.15: 90617731-90617731 on Build GRCh38
rs893564510 Chr.15: 90617634-90617634 on Build GRCh38
rs773086130 Chr.15: 90617894-90617894 on Build GRCh38
rs763025184 Chr.15: 90617874-90617874 on Build GRCh38
rs9888660 Chr.15: 90617536-90617536 on Build GRCh38
rs753545725 Chr.15: 90617899-90617899 on Build GRCh38
rs149408148 Chr.15: 90617954-90617954 on Build GRCh38
rs368247464 Chr.15: 90617848-90617848 on Build GRCh38
rs985490300 Chr.15: 90617592-90617592 on Build GRCh38
rs184470048 Chr.15: 90617811-90617811 on Build GRCh38
rs758226599 Chr.15: 90617931-90617931 on Build GRCh38
rs775448244 Chr.15: 90617871-90617871 on Build GRCh38

View Full Product Details