Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 72097537-72097998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX38
Entrez Gene ID
9785
Gene Name
DEAH-box helicase 38
Gene Aliases
DDX38, PRP16, PRPF16
UniGene
Hs.151218
Gene Chromosome Location
Chr.16: 72093665-72112912 on Build GRCh38
Transcripts
NM_014003.3 XM_005256269.1 XM_011523484.1 XM_011523485.1 XM_017023913.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1203560 c.607C>T p.R203* Chr.16: 72097772-72097772 on Build GRCh38
3421150 c.608G>A p.R203Q Chr.16: 72097773-72097773 on Build GRCh38
4438940 c.560G>A p.G187E Chr.16: 72097725-72097725 on Build GRCh38
1379685 c.591C>A p.P197P Chr.16: 72097756-72097756 on Build GRCh38
6347509 c.565G>T p.E189* Chr.16: 72097730-72097730 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558767894 Chr.16: 72097943-72097943 on Build GRCh38
rs977001844 Chr.16: 72097793-72097793 on Build GRCh38
rs11862496 Chr.16: 72097561-72097561 on Build GRCh38
rs535966988 Chr.16: 72097831-72097831 on Build GRCh38
rs898036746 Chr.16: 72097938-72097938 on Build GRCh38
rs148657156 Chr.16: 72097559-72097559 on Build GRCh38
rs935662577 Chr.16: 72097660-72097660 on Build GRCh38
rs3036401 Chr.16: 72097842-72097844 on Build GRCh38
rs775129865 Chr.16: 72097753-72097753 on Build GRCh38
rs1046321974 Chr.16: 72097936-72097936 on Build GRCh38
rs767489479 Chr.16: 72097768-72097768 on Build GRCh38
rs778329320 Chr.16: 72097811-72097811 on Build GRCh38
rs151219524 Chr.16: 72097603-72097603 on Build GRCh38
rs977179024 Chr.16: 72097834-72097834 on Build GRCh38
rs35420744 Chr.16: 72097904-72097905 on Build GRCh38
rs753918448 Chr.16: 72097772-72097772 on Build GRCh38
rs1028947237 Chr.16: 72097972-72097972 on Build GRCh38
rs925581281 Chr.16: 72097659-72097659 on Build GRCh38
rs774114256 Chr.16: 72097757-72097757 on Build GRCh38
rs776044870 Chr.16: 72097727-72097727 on Build GRCh38
rs10537374 Chr.16: 72097845-72097846 on Build GRCh38
rs750674805 Chr.16: 72097830-72097831 on Build GRCh38
rs935693640 Chr.16: 72097848-72097848 on Build GRCh38
rs374944590 Chr.16: 72097789-72097789 on Build GRCh38
rs963971164 Chr.16: 72097773-72097773 on Build GRCh38
rs975874260 Chr.16: 72097637-72097637 on Build GRCh38
rs184925614 Chr.16: 72097881-72097881 on Build GRCh38
rs757366316 Chr.16: 72097661-72097661 on Build GRCh38
rs576128612 Chr.16: 72097949-72097949 on Build GRCh38
rs950296841 Chr.16: 72097904-72097904 on Build GRCh38
rs762555103 Chr.16: 72097755-72097755 on Build GRCh38
rs1004580275 Chr.16: 72097592-72097592 on Build GRCh38
rs901725838 Chr.16: 72097933-72097942 on Build GRCh38
rs778241309 Chr.16: 72097715-72097715 on Build GRCh38
rs770968364 Chr.16: 72097763-72097763 on Build GRCh38
rs540151474 Chr.16: 72097896-72097896 on Build GRCh38
rs200666031 Chr.16: 72097799-72097799 on Build GRCh38
rs559408140 Chr.16: 72097899-72097899 on Build GRCh38
rs752996092 Chr.16: 72097797-72097797 on Build GRCh38
rs758835301 Chr.16: 72097710-72097710 on Build GRCh38
rs770376548 Chr.16: 72097638-72097638 on Build GRCh38
rs763901646 Chr.16: 72097756-72097756 on Build GRCh38
rs757762935 Chr.16: 72097820-72097820 on Build GRCh38
rs1049201713 Chr.16: 72097965-72097965 on Build GRCh38
rs945769148 Chr.16: 72097621-72097621 on Build GRCh38
rs747451837 Chr.16: 72097635-72097635 on Build GRCh38
rs140736447 Chr.16: 72097684-72097684 on Build GRCh38
rs34749771 Chr.16: 72097842-72097843 on Build GRCh38
rs922362170 Chr.16: 72097614-72097614 on Build GRCh38
rs781729705 Chr.16: 72097824-72097824 on Build GRCh38
rs554249100 Chr.16: 72097854-72097854 on Build GRCh38
rs775322616 Chr.16: 72097682-72097682 on Build GRCh38
rs568581243 Chr.16: 72097698-72097698 on Build GRCh38
rs769129571 Chr.16: 72097734-72097734 on Build GRCh38
rs1045843478 Chr.16: 72097847-72097847 on Build GRCh38
rs747539910 Chr.16: 72097716-72097716 on Build GRCh38
rs1041371351 Chr.16: 72097649-72097649 on Build GRCh38
rs144391201 Chr.16: 72097705-72097705 on Build GRCh38
rs964248341 Chr.16: 72097572-72097572 on Build GRCh38
rs748320622 Chr.16: 72097709-72097709 on Build GRCh38
rs764939717 Chr.16: 72097677-72097677 on Build GRCh38
rs887548021 Chr.16: 72097877-72097877 on Build GRCh38
rs751531063 Chr.16: 72097685-72097685 on Build GRCh38
rs757140615 Chr.16: 72097686-72097686 on Build GRCh38
rs773866098 Chr.16: 72097631-72097631 on Build GRCh38
rs937564954 Chr.16: 72097867-72097867 on Build GRCh38
rs755096342 Chr.16: 72097784-72097784 on Build GRCh38
rs776156852 Chr.16: 72097647-72097647 on Build GRCh38
rs745370411 Chr.16: 72097731-72097731 on Build GRCh38
rs1049272913 Chr.16: 72097871-72097871 on Build GRCh38
rs1014600916 Chr.16: 72097913-72097913 on Build GRCh38
rs771446107 Chr.16: 72097718-72097718 on Build GRCh38
rs755218183 Chr.16: 72097703-72097703 on Build GRCh38
rs894711614 Chr.16: 72097663-72097663 on Build GRCh38
rs1014820728 Chr.16: 72097593-72097593 on Build GRCh38
rs766421564 Chr.16: 72097689-72097689 on Build GRCh38
rs925693465 Chr.16: 72097846-72097846 on Build GRCh38
rs913464685 Chr.16: 72097838-72097838 on Build GRCh38
rs538803789 Chr.16: 72097566-72097566 on Build GRCh38
rs779523896 Chr.16: 72097817-72097817 on Build GRCh38

View Full Product Details