Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 83031853-83032397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH13
Entrez Gene ID
1012
Gene Name
cadherin 13
Gene Aliases
CDHH, P105
UniGene
Hs.654386
Gene Chromosome Location
Chr.16: 82626794-83796610 on Build GRCh38
Transcripts
NM_001220488.1 NM_001220489.1 NM_001220490.1 NM_001220491.1 NM_001220492.1 NM_001257.4 XM_017022848.1 XM_017022849.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7228654 c.22G>C p.G8R Chr.16: 83032189-83032189 on Build GRCh38
3712111 c.320C>A p.A107E Chr.16: 83032172-83032172 on Build GRCh38
6145311 c.303C>A p.P101P Chr.16: 83032155-83032155 on Build GRCh38
6498501 c.334G>A p.V112I Chr.16: 83032186-83032186 on Build GRCh38
379116 c.295C>G p.R99G Chr.16: 83032147-83032147 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764433326 Chr.16: 83032054-83032054 on Build GRCh38
rs919884884 Chr.16: 83031874-83031874 on Build GRCh38
rs760117321 Chr.16: 83032239-83032239 on Build GRCh38
rs748842714 Chr.16: 83032215-83032215 on Build GRCh38
rs371552389 Chr.16: 83032346-83032346 on Build GRCh38
rs751991343 Chr.16: 83032056-83032056 on Build GRCh38
rs188513868 Chr.16: 83031931-83031931 on Build GRCh38
rs530736609 Chr.16: 83031950-83031950 on Build GRCh38
rs893119452 Chr.16: 83032118-83032118 on Build GRCh38
rs766475325 Chr.16: 83032101-83032101 on Build GRCh38
rs371572523 Chr.16: 83032063-83032063 on Build GRCh38
rs931245498 Chr.16: 83032041-83032041 on Build GRCh38
rs1001574840 Chr.16: 83031903-83031903 on Build GRCh38
rs967913119 Chr.16: 83031955-83031955 on Build GRCh38
rs770121469 Chr.16: 83031968-83031968 on Build GRCh38
rs374734713 Chr.16: 83032058-83032058 on Build GRCh38
rs955014747 Chr.16: 83031915-83031915 on Build GRCh38
rs748410884 Chr.16: 83032121-83032121 on Build GRCh38
rs575477444 Chr.16: 83032190-83032190 on Build GRCh38
rs758199647 Chr.16: 83032123-83032123 on Build GRCh38
rs762111262 Chr.16: 83032267-83032267 on Build GRCh38
rs781266482 Chr.16: 83031963-83031963 on Build GRCh38
rs375003818 Chr.16: 83032073-83032073 on Build GRCh38
rs760131685 Chr.16: 83032193-83032193 on Build GRCh38
rs183211035 Chr.16: 83031997-83031997 on Build GRCh38
rs982459451 Chr.16: 83032174-83032174 on Build GRCh38
rs569386167 Chr.16: 83032279-83032279 on Build GRCh38
rs757196752 Chr.16: 83032167-83032167 on Build GRCh38
rs367975473 Chr.16: 83032305-83032305 on Build GRCh38
rs377383586 Chr.16: 83032153-83032153 on Build GRCh38
rs745840757 Chr.16: 83032179-83032179 on Build GRCh38
rs762601185 Chr.16: 83032145-83032145 on Build GRCh38
rs148997929 Chr.16: 83032235-83032235 on Build GRCh38
rs959562932 Chr.16: 83032332-83032332 on Build GRCh38
rs767121144 Chr.16: 83031979-83031979 on Build GRCh38
rs564171901 Chr.16: 83031969-83031969 on Build GRCh38
rs779566316 Chr.16: 83032173-83032173 on Build GRCh38
rs766100928 Chr.16: 83031996-83031996 on Build GRCh38
rs2228685 Chr.16: 83032360-83032360 on Build GRCh38
rs143774800 Chr.16: 83032256-83032256 on Build GRCh38
rs867078223 Chr.16: 83032031-83032031 on Build GRCh38
rs1041213234 Chr.16: 83032242-83032242 on Build GRCh38
rs564135228 Chr.16: 83032196-83032196 on Build GRCh38
rs541667190 Chr.16: 83031916-83031916 on Build GRCh38
rs1056635050 Chr.16: 83031984-83031984 on Build GRCh38
rs544219355 Chr.16: 83032192-83032192 on Build GRCh38
rs145871512 Chr.16: 83032084-83032084 on Build GRCh38
rs755224823 Chr.16: 83032113-83032113 on Build GRCh38
rs774362680 Chr.16: 83031974-83031974 on Build GRCh38
rs147586379 Chr.16: 83031905-83031905 on Build GRCh38
rs963764359 Chr.16: 83031992-83031992 on Build GRCh38
rs746922215 Chr.16: 83032075-83032075 on Build GRCh38
rs1033373638 Chr.16: 83032310-83032310 on Build GRCh38
rs746069538 Chr.16: 83031967-83031967 on Build GRCh38
rs986575826 Chr.16: 83031987-83031987 on Build GRCh38
rs7197530 Chr.16: 83032299-83032299 on Build GRCh38
rs780973504 Chr.16: 83032207-83032207 on Build GRCh38
rs752907921 Chr.16: 83032159-83032159 on Build GRCh38
rs111370214 Chr.16: 83032335-83032335 on Build GRCh38
rs187937615 Chr.16: 83032001-83032001 on Build GRCh38
rs371079121 Chr.16: 83032130-83032130 on Build GRCh38
rs199539898 Chr.16: 83032160-83032160 on Build GRCh38
rs749510568 Chr.16: 83032065-83032065 on Build GRCh38
rs762117667 Chr.16: 83032090-83032090 on Build GRCh38
rs770465360 Chr.16: 83032250-83032250 on Build GRCh38
rs191402207 Chr.16: 83032029-83032029 on Build GRCh38
rs765834816 Chr.16: 83032047-83032047 on Build GRCh38
rs549329150 Chr.16: 83032036-83032036 on Build GRCh38
rs538944333 Chr.16: 83032038-83032038 on Build GRCh38
rs549533705 Chr.16: 83032278-83032278 on Build GRCh38
rs778819651 Chr.16: 83032071-83032071 on Build GRCh38
rs753900035 Chr.16: 83032106-83032106 on Build GRCh38
rs534685566 Chr.16: 83031930-83031930 on Build GRCh38
rs770842301 Chr.16: 83032137-83032137 on Build GRCh38
rs749506528 Chr.16: 83032035-83032035 on Build GRCh38
rs376010391 Chr.16: 83032249-83032249 on Build GRCh38
rs533111528 Chr.16: 83032225-83032225 on Build GRCh38
rs764269261 Chr.16: 83032258-83032258 on Build GRCh38
rs959159104 Chr.16: 83032302-83032302 on Build GRCh38
rs760209670 Chr.16: 83031991-83031991 on Build GRCh38
rs772176185 Chr.16: 83032042-83032042 on Build GRCh38
rs371659407 Chr.16: 83032046-83032046 on Build GRCh38
rs201332621 Chr.16: 83032147-83032147 on Build GRCh38
rs775689913 Chr.16: 83032253-83032253 on Build GRCh38
rs755317016 Chr.16: 83032068-83032068 on Build GRCh38
rs370986033 Chr.16: 83032188-83032188 on Build GRCh38
rs541772361 Chr.16: 83032083-83032083 on Build GRCh38
rs975074520 Chr.16: 83031995-83031995 on Build GRCh38
rs982590192 Chr.16: 83031922-83031922 on Build GRCh38
rs762857450 Chr.16: 83031970-83031970 on Build GRCh38
rs766424984 Chr.16: 83032147-83032148 on Build GRCh38
rs571729482 Chr.16: 83032155-83032155 on Build GRCh38
rs369679692 Chr.16: 83032080-83032080 on Build GRCh38
rs759127708 Chr.16: 83032052-83032052 on Build GRCh38
rs996517326 Chr.16: 83031909-83031909 on Build GRCh38
rs112485535 Chr.16: 83031887-83031887 on Build GRCh38
rs6565105 Chr.16: 83032059-83032059 on Build GRCh38
rs192942006 Chr.16: 83031986-83031986 on Build GRCh38
rs368685803 Chr.16: 83032045-83032045 on Build GRCh38
rs1020922042 Chr.16: 83032132-83032132 on Build GRCh38
rs971065691 Chr.16: 83032288-83032288 on Build GRCh38
rs746000723 Chr.16: 83032023-83032023 on Build GRCh38
rs750878531 Chr.16: 83032092-83032092 on Build GRCh38
rs866019796 Chr.16: 83032124-83032124 on Build GRCh38
rs780275996 Chr.16: 83032034-83032034 on Build GRCh38
rs764273398 Chr.16: 83032195-83032195 on Build GRCh38
rs753461003 Chr.16: 83032050-83032050 on Build GRCh38
rs918157242 Chr.16: 83032166-83032166 on Build GRCh38
rs764913956 Chr.16: 83032089-83032089 on Build GRCh38
rs200199969 Chr.16: 83032186-83032186 on Build GRCh38
rs140344950 Chr.16: 83031913-83031913 on Build GRCh38
rs749708145 Chr.16: 83032265-83032265 on Build GRCh38
rs781249825 Chr.16: 83032017-83032017 on Build GRCh38
rs368675786 Chr.16: 83032194-83032194 on Build GRCh38
rs7197352 Chr.16: 83032214-83032214 on Build GRCh38
rs780946158 Chr.16: 83032176-83032176 on Build GRCh38
rs189485423 Chr.16: 83032293-83032293 on Build GRCh38
rs907276242 Chr.16: 83031893-83031893 on Build GRCh38
rs768596627 Chr.16: 83031972-83031972 on Build GRCh38
rs778118910 Chr.16: 83031998-83031998 on Build GRCh38
rs751351911 Chr.16: 83032161-83032161 on Build GRCh38
rs944308528 Chr.16: 83032344-83032344 on Build GRCh38
rs746954334 Chr.16: 83032136-83032136 on Build GRCh38
rs746609469 Chr.16: 83032152-83032152 on Build GRCh38
rs758138760 Chr.16: 83032143-83032143 on Build GRCh38
rs769320614 Chr.16: 83032142-83032142 on Build GRCh38
rs765410639 Chr.16: 83032156-83032156 on Build GRCh38
rs756966285 Chr.16: 83032198-83032198 on Build GRCh38
rs1015641693 Chr.16: 83032271-83032271 on Build GRCh38
rs773157567 Chr.16: 83032074-83032074 on Build GRCh38
rs779228407 Chr.16: 83032119-83032119 on Build GRCh38
rs774678387 Chr.16: 83032359-83032359 on Build GRCh38
rs7189859 Chr.16: 83031886-83031886 on Build GRCh38
rs761836771 Chr.16: 83032040-83032040 on Build GRCh38
rs183971768 Chr.16: 83032189-83032189 on Build GRCh38
rs756635742 Chr.16: 83032096-83032096 on Build GRCh38
rs1025242610 Chr.16: 83032339-83032339 on Build GRCh38
rs200439811 Chr.16: 83032138-83032138 on Build GRCh38
rs372694159 Chr.16: 83032228-83032228 on Build GRCh38
rs945409195 Chr.16: 83032184-83032184 on Build GRCh38
rs151097273 Chr.16: 83032292-83032292 on Build GRCh38
rs767804761 Chr.16: 83032091-83032091 on Build GRCh38
rs771358036 Chr.16: 83032191-83032191 on Build GRCh38
rs375168918 Chr.16: 83032180-83032180 on Build GRCh38
rs369916041 Chr.16: 83032148-83032148 on Build GRCh38
rs1019817868 Chr.16: 83031988-83031988 on Build GRCh38
rs761854858 Chr.16: 83031975-83031975 on Build GRCh38
rs556704424 Chr.16: 83031885-83031885 on Build GRCh38
rs550927856 Chr.16: 83031962-83031962 on Build GRCh38
rs778389493 Chr.16: 83032223-83032223 on Build GRCh38
rs752068202 Chr.16: 83032150-83032151 on Build GRCh38
rs1015245316 Chr.16: 83032280-83032280 on Build GRCh38
rs372200232 Chr.16: 83032212-83032212 on Build GRCh38

View Full Product Details