Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70258863-70259358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AARS
Entrez Gene ID
16
Gene Name
alanyl-tRNA synthetase
Gene Aliases
CMT2N, EIEE29
UniGene
Hs.315137
Gene Chromosome Location
Chr.16: 70252295-70289552 on Build GRCh38
Transcripts
NM_001605.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
242702 c.1840G>A p.A614T Chr.16: 70259132-70259132 on Build GRCh38
278486 c.1904G>T p.R635I Chr.16: 70259068-70259068 on Build GRCh38
6480116 c.1847G>A p.R616H Chr.16: 70259125-70259125 on Build GRCh38
5441260 c.1957G>A p.E653K Chr.16: 70259015-70259015 on Build GRCh38
3272813 c.1897C>T p.R633C Chr.16: 70259075-70259075 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756739953 Chr.16: 70258969-70258969 on Build GRCh38
rs1026305039 Chr.16: 70259244-70259244 on Build GRCh38
rs147291818 Chr.16: 70258942-70258942 on Build GRCh38
rs549523841 Chr.16: 70259277-70259277 on Build GRCh38
rs763252550 Chr.16: 70259036-70259036 on Build GRCh38
rs143855726 Chr.16: 70258909-70258909 on Build GRCh38
rs867893652 Chr.16: 70259245-70259245 on Build GRCh38
rs752673097 Chr.16: 70259094-70259094 on Build GRCh38
rs908986006 Chr.16: 70259280-70259280 on Build GRCh38
rs368023282 Chr.16: 70258965-70258965 on Build GRCh38
rs756118408 Chr.16: 70259184-70259184 on Build GRCh38
rs770013009 Chr.16: 70259129-70259129 on Build GRCh38
rs1054341533 Chr.16: 70259209-70259209 on Build GRCh38
rs933827393 Chr.16: 70258892-70258892 on Build GRCh38
rs200406732 Chr.16: 70258898-70258898 on Build GRCh38
rs749039663 Chr.16: 70259003-70259003 on Build GRCh38
rs373963866 Chr.16: 70258939-70258939 on Build GRCh38
rs945873486 Chr.16: 70258966-70258966 on Build GRCh38
rs1036069166 Chr.16: 70259326-70259326 on Build GRCh38
rs758423103 Chr.16: 70259075-70259075 on Build GRCh38
rs371595630 Chr.16: 70259161-70259161 on Build GRCh38
rs765959454 Chr.16: 70259174-70259174 on Build GRCh38
rs552900235 Chr.16: 70258901-70258901 on Build GRCh38
rs77814513 Chr.16: 70259191-70259191 on Build GRCh38
rs774290442 Chr.16: 70259200-70259201 on Build GRCh38
rs112620320 Chr.16: 70258978-70258978 on Build GRCh38
rs773357160 Chr.16: 70258948-70258948 on Build GRCh38
rs780198710 Chr.16: 70259132-70259132 on Build GRCh38
rs781270199 Chr.16: 70259125-70259125 on Build GRCh38
rs746047868 Chr.16: 70259029-70259029 on Build GRCh38
rs748142922 Chr.16: 70259009-70259009 on Build GRCh38
rs765106728 Chr.16: 70259179-70259179 on Build GRCh38
rs759349135 Chr.16: 70259211-70259212 on Build GRCh38
rs971554160 Chr.16: 70258955-70258955 on Build GRCh38
rs767054861 Chr.16: 70258973-70258973 on Build GRCh38
rs759201092 Chr.16: 70259230-70259230 on Build GRCh38
rs763453406 Chr.16: 70258956-70258956 on Build GRCh38
rs773583428 Chr.16: 70259260-70259260 on Build GRCh38
rs780249378 Chr.16: 70259186-70259186 on Build GRCh38
rs769917416 Chr.16: 70259030-70259030 on Build GRCh38
rs941040970 Chr.16: 70259074-70259074 on Build GRCh38
rs758375657 Chr.16: 70259181-70259181 on Build GRCh38
rs35986765 Chr.16: 70258907-70258907 on Build GRCh38
rs1038620594 Chr.16: 70259076-70259076 on Build GRCh38
rs758427335 Chr.16: 70259108-70259108 on Build GRCh38
rs564005034 Chr.16: 70259261-70259261 on Build GRCh38
rs1006948771 Chr.16: 70259163-70259163 on Build GRCh38
rs775379073 Chr.16: 70259238-70259238 on Build GRCh38
rs779839889 Chr.16: 70259001-70259001 on Build GRCh38
rs398058353 Chr.16: 70258898-70258899 on Build GRCh38
rs958804412 Chr.16: 70259268-70259268 on Build GRCh38
rs1003014467 Chr.16: 70259255-70259257 on Build GRCh38
rs34846578 Chr.16: 70258896-70258897 on Build GRCh38
rs928474071 Chr.16: 70259020-70259020 on Build GRCh38
rs573789791 Chr.16: 70259024-70259024 on Build GRCh38
rs199644417 Chr.16: 70259148-70259148 on Build GRCh38
rs769989483 Chr.16: 70259255-70259255 on Build GRCh38
rs545686456 Chr.16: 70258930-70258930 on Build GRCh38
rs769363058 Chr.16: 70258960-70258960 on Build GRCh38
rs770339465 Chr.16: 70259226-70259226 on Build GRCh38
rs148686377 Chr.16: 70258989-70258989 on Build GRCh38
rs768830699 Chr.16: 70259183-70259183 on Build GRCh38
rs777597230 Chr.16: 70259182-70259182 on Build GRCh38
rs985951789 Chr.16: 70258895-70258895 on Build GRCh38
rs776336061 Chr.16: 70259228-70259228 on Build GRCh38
rs187627813 Chr.16: 70259287-70259287 on Build GRCh38
rs552080527 Chr.16: 70259213-70259213 on Build GRCh38
rs373852619 Chr.16: 70258951-70258951 on Build GRCh38
rs755780106 Chr.16: 70258993-70258993 on Build GRCh38
rs1055964123 Chr.16: 70259032-70259032 on Build GRCh38
rs765016757 Chr.16: 70259234-70259234 on Build GRCh38
rs765977339 Chr.16: 70259078-70259078 on Build GRCh38
rs761281864 Chr.16: 70259224-70259224 on Build GRCh38
rs557679947 Chr.16: 70258934-70258934 on Build GRCh38
rs753617292 Chr.16: 70259085-70259085 on Build GRCh38
rs372221218 Chr.16: 70259126-70259126 on Build GRCh38
rs775734426 Chr.16: 70258954-70258954 on Build GRCh38
rs191745414 Chr.16: 70259305-70259305 on Build GRCh38
rs779779037 Chr.16: 70258940-70258940 on Build GRCh38
rs774810867 Chr.16: 70259048-70259048 on Build GRCh38
rs957615750 Chr.16: 70258910-70258910 on Build GRCh38
rs766999932 Chr.16: 70259065-70259065 on Build GRCh38
rs889720866 Chr.16: 70258904-70258904 on Build GRCh38
rs141374386 Chr.16: 70259133-70259133 on Build GRCh38
rs553681520 Chr.16: 70259285-70259286 on Build GRCh38
rs367579673 Chr.16: 70259154-70259154 on Build GRCh38
rs148608437 Chr.16: 70258941-70258941 on Build GRCh38
rs772774828 Chr.16: 70259033-70259033 on Build GRCh38
rs753398944 Chr.16: 70259175-70259175 on Build GRCh38
rs907296734 Chr.16: 70259239-70259239 on Build GRCh38
rs531124165 Chr.16: 70259262-70259262 on Build GRCh38
rs772735487 Chr.16: 70259170-70259170 on Build GRCh38
rs368548525 Chr.16: 70259042-70259042 on Build GRCh38
rs184444743 Chr.16: 70259212-70259212 on Build GRCh38
rs765625673 Chr.16: 70258953-70258953 on Build GRCh38
rs897445579 Chr.16: 70259115-70259115 on Build GRCh38
rs200896501 Chr.16: 70259090-70259090 on Build GRCh38
rs747848503 Chr.16: 70258949-70258950 on Build GRCh38
rs760123813 Chr.16: 70259171-70259171 on Build GRCh38
rs754924454 Chr.16: 70259004-70259004 on Build GRCh38
rs757181086 Chr.16: 70259243-70259243 on Build GRCh38
rs373438232 Chr.16: 70259218-70259218 on Build GRCh38
rs561750417 Chr.16: 70259068-70259068 on Build GRCh38
rs749210185 Chr.16: 70259259-70259259 on Build GRCh38
rs762373456 Chr.16: 70259056-70259056 on Build GRCh38
rs752548366 Chr.16: 70259180-70259180 on Build GRCh38
rs535074752 Chr.16: 70259299-70259299 on Build GRCh38
rs893129887 Chr.16: 70259209-70259210 on Build GRCh38
rs770901079 Chr.16: 70259150-70259151 on Build GRCh38
rs771099188 Chr.16: 70259019-70259019 on Build GRCh38
rs747114635 Chr.16: 70258950-70258950 on Build GRCh38
rs781224347 Chr.16: 70259189-70259189 on Build GRCh38
rs147822575 Chr.16: 70259034-70259034 on Build GRCh38
rs762414186 Chr.16: 70259148-70259149 on Build GRCh38
rs377247120 Chr.16: 70259153-70259153 on Build GRCh38
rs142233951 Chr.16: 70259023-70259023 on Build GRCh38
rs909529951 Chr.16: 70259041-70259041 on Build GRCh38
rs748542233 Chr.16: 70259210-70259210 on Build GRCh38
rs779013701 Chr.16: 70259007-70259007 on Build GRCh38
rs368681927 Chr.16: 70259257-70259259 on Build GRCh38
rs201177309 Chr.16: 70258945-70258945 on Build GRCh38
rs779182382 Chr.16: 70259208-70259208 on Build GRCh38
rs1018795546 Chr.16: 70259199-70259199 on Build GRCh38

View Full Product Details