Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 21183251-21183729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS7B
Entrez Gene ID
25979
Gene Name
dehydrogenase/reductase 7B
Gene Aliases
CGI-93, SDR32C1
UniGene
Hs.386989
Gene Chromosome Location
Chr.17: 21123369-21191972 on Build GRCh38
Transcripts
NM_015510.4 XM_005256595.4 XM_005256596.3 XM_011523786.2 XM_017024425.1 XM_017024426.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5481357 c.372A>T p.A124A Chr.17: 21183656-21183656 on Build GRCh38
6556232 c.352G>T p.D118Y Chr.17: 21183636-21183636 on Build GRCh38
6392854 c.333G>T p.L111F Chr.17: 21183617-21183617 on Build GRCh38
4677322 c.373G>A p.A125T Chr.17: 21183657-21183657 on Build GRCh38
6390057 c.358G>A p.G120R Chr.17: 21183642-21183642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777571423 Chr.17: 21183596-21183596 on Build GRCh38
rs1007208031 Chr.17: 21183521-21183521 on Build GRCh38
rs988895451 Chr.17: 21183492-21183492 on Build GRCh38
rs928653565 Chr.17: 21183315-21183315 on Build GRCh38
rs746754851 Chr.17: 21183708-21183708 on Build GRCh38
rs941872189 Chr.17: 21183344-21183344 on Build GRCh38
rs768069944 Chr.17: 21183358-21183358 on Build GRCh38
rs374662243 Chr.17: 21183699-21183699 on Build GRCh38
rs374983442 Chr.17: 21183528-21183528 on Build GRCh38
rs753469366 Chr.17: 21183557-21183557 on Build GRCh38
rs751420575 Chr.17: 21183698-21183698 on Build GRCh38
rs1051509146 Chr.17: 21183491-21183491 on Build GRCh38
rs771103238 Chr.17: 21183408-21183408 on Build GRCh38
rs770787003 Chr.17: 21183610-21183610 on Build GRCh38
rs371553142 Chr.17: 21183656-21183656 on Build GRCh38
rs115819229 Chr.17: 21183626-21183626 on Build GRCh38
rs762266141 Chr.17: 21183679-21183679 on Build GRCh38
rs925972248 Chr.17: 21183428-21183428 on Build GRCh38
rs890130189 Chr.17: 21183498-21183498 on Build GRCh38
rs1035751584 Chr.17: 21183657-21183657 on Build GRCh38
rs991410820 Chr.17: 21183661-21183661 on Build GRCh38
rs768200460 Chr.17: 21183685-21183685 on Build GRCh38
rs945203466 Chr.17: 21183526-21183526 on Build GRCh38
rs1021838703 Chr.17: 21183579-21183579 on Build GRCh38
rs960334334 Chr.17: 21183658-21183658 on Build GRCh38
rs957633729 Chr.17: 21183440-21183440 on Build GRCh38
rs929086966 Chr.17: 21183423-21183423 on Build GRCh38
rs752311534 Chr.17: 21183693-21183693 on Build GRCh38
rs545725296 Chr.17: 21183574-21183574 on Build GRCh38
rs938685375 Chr.17: 21183320-21183320 on Build GRCh38
rs563915865 Chr.17: 21183623-21183623 on Build GRCh38
rs754524846 Chr.17: 21183562-21183562 on Build GRCh38
rs373616709 Chr.17: 21183543-21183543 on Build GRCh38
rs977528751 Chr.17: 21183299-21183299 on Build GRCh38
rs367713441 Chr.17: 21183620-21183620 on Build GRCh38
rs777434900 Chr.17: 21183697-21183697 on Build GRCh38
rs1037464634 Chr.17: 21183354-21183354 on Build GRCh38
rs758155459 Chr.17: 21183577-21183577 on Build GRCh38
rs778477283 Chr.17: 21183567-21183567 on Build GRCh38
rs200070541 Chr.17: 21183692-21183692 on Build GRCh38
rs961549583 Chr.17: 21183296-21183296 on Build GRCh38
rs553054335 Chr.17: 21183581-21183581 on Build GRCh38
rs747130977 Chr.17: 21183322-21183322 on Build GRCh38
rs763186459 Chr.17: 21183648-21183648 on Build GRCh38
rs774715342 Chr.17: 21183664-21183664 on Build GRCh38
rs781149318 Chr.17: 21183709-21183709 on Build GRCh38
rs915838784 Chr.17: 21183695-21183695 on Build GRCh38
rs758010682 Chr.17: 21183694-21183694 on Build GRCh38
rs915806442 Chr.17: 21183332-21183332 on Build GRCh38
rs144330164 Chr.17: 21183688-21183688 on Build GRCh38
rs999376348 Chr.17: 21183652-21183652 on Build GRCh38

View Full Product Details