Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 15452699-15453166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDRT4
Entrez Gene ID
284040
Gene Name
CMT1A duplicated region transcript 4
Gene Aliases
-
UniGene
Hs.164595
Gene Chromosome Location
Chr.17: 15436015-15467611 on Build GRCh38
Transcripts
NM_001204477.1 NM_001204478.1 NR_037924.1

Gene Symbol
TVP23C-CDRT4
Entrez Gene ID
100533496
Gene Name
TVP23C-CDRT4 readthrough
Gene Aliases
FAM18B2-CDRT4
UniGene
Hs.164595
Gene Chromosome Location
Chr.17: 15436015-15563631 on Build GRCh38
Transcripts
NM_001204477.1 NM_001204478.1 NR_037924.1

SNP Detail

SNP ID Chromosome Location
rs764436651 Chr.17: 15452765-15452765 on Build GRCh38
rs889709112 Chr.17: 15452966-15452966 on Build GRCh38
rs1033353713 Chr.17: 15452823-15452823 on Build GRCh38
rs1029809882 Chr.17: 15452859-15452862 on Build GRCh38
rs1045619309 Chr.17: 15452780-15452780 on Build GRCh38
rs915650326 Chr.17: 15452982-15452982 on Build GRCh38
rs1019491227 Chr.17: 15453058-15453058 on Build GRCh38
rs903544853 Chr.17: 15452892-15452892 on Build GRCh38
rs550845528 Chr.17: 15452768-15452768 on Build GRCh38
rs961195987 Chr.17: 15453114-15453114 on Build GRCh38
rs576537304 Chr.17: 15453063-15453063 on Build GRCh38
rs77821761 Chr.17: 15453037-15453037 on Build GRCh38
rs972550121 Chr.17: 15452725-15452725 on Build GRCh38
rs983026630 Chr.17: 15452906-15452906 on Build GRCh38
rs945165062 Chr.17: 15452840-15452840 on Build GRCh38
rs952124083 Chr.17: 15452748-15452748 on Build GRCh38
rs964132687 Chr.17: 15452851-15452854 on Build GRCh38
rs1000951313 Chr.17: 15452934-15452934 on Build GRCh38
rs934495406 Chr.17: 15453108-15453108 on Build GRCh38
rs991596298 Chr.17: 15452979-15452979 on Build GRCh38
rs549835124 Chr.17: 15452856-15452857 on Build GRCh38
rs1042530926 Chr.17: 15452847-15452847 on Build GRCh38
rs910128819 Chr.17: 15452952-15452952 on Build GRCh38
rs949803307 Chr.17: 15452987-15452987 on Build GRCh38
rs951649543 Chr.17: 15452871-15452874 on Build GRCh38
rs765246387 Chr.17: 15452846-15452846 on Build GRCh38
rs191154803 Chr.17: 15452975-15452975 on Build GRCh38
rs2954778 Chr.17: 15452816-15452816 on Build GRCh38
rs1049380164 Chr.17: 15452958-15452958 on Build GRCh38
rs1056562344 Chr.17: 15452800-15452800 on Build GRCh38
rs7223838 Chr.17: 15452792-15452792 on Build GRCh38
rs775473770 Chr.17: 15452895-15452895 on Build GRCh38
rs976877611 Chr.17: 15452857-15452862 on Build GRCh38
rs894269019 Chr.17: 15452762-15452762 on Build GRCh38
rs985089426 Chr.17: 15452751-15452751 on Build GRCh38
rs919303687 Chr.17: 15452738-15452738 on Build GRCh38
rs902987464 Chr.17: 15453044-15453045 on Build GRCh38
rs372541102 Chr.17: 15453142-15453142 on Build GRCh38
rs573250551 Chr.17: 15452778-15452778 on Build GRCh38
rs183658666 Chr.17: 15452998-15452998 on Build GRCh38

View Full Product Details