Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50375030-50375614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EME1
Entrez Gene ID
146956
Gene Name
essential meiotic structure-specific endonuclease 1
Gene Aliases
MMS4L, SLX2A
UniGene
Hs.514330
Gene Chromosome Location
Chr.17: 50373220-50381479 on Build GRCh38
Transcripts
NM_001166131.1 NM_152463.2 XM_005257081.3 XM_011524395.2 XM_017024236.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
981003 c.278T>G p.F93C Chr.17: 50375486-50375486 on Build GRCh38
4945429 c.404G>T p.W135L Chr.17: 50375612-50375612 on Build GRCh38
4649581 c.124G>A p.E42K Chr.17: 50375332-50375332 on Build GRCh38
981001 c.277T>A p.F93I Chr.17: 50375485-50375485 on Build GRCh38
980999 c.268_270delGAA p.E92delE Chr.17: 50375476-50375478 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772389421 Chr.17: 50375434-50375434 on Build GRCh38
rs761429359 Chr.17: 50375203-50375203 on Build GRCh38
rs199769548 Chr.17: 50375300-50375300 on Build GRCh38
rs753452084 Chr.17: 50375476-50375478 on Build GRCh38
rs752663381 Chr.17: 50375391-50375391 on Build GRCh38
rs375893410 Chr.17: 50375581-50375581 on Build GRCh38
rs370330243 Chr.17: 50375539-50375539 on Build GRCh38
rs150029835 Chr.17: 50375405-50375405 on Build GRCh38
rs146276559 Chr.17: 50375246-50375246 on Build GRCh38
rs35248609 Chr.17: 50375223-50375223 on Build GRCh38
rs754099573 Chr.17: 50375525-50375525 on Build GRCh38
rs754940303 Chr.17: 50375351-50375351 on Build GRCh38
rs767615386 Chr.17: 50375275-50375275 on Build GRCh38
rs779198639 Chr.17: 50375538-50375538 on Build GRCh38
rs996422546 Chr.17: 50375055-50375055 on Build GRCh38
rs146890422 Chr.17: 50375465-50375465 on Build GRCh38
rs141978919 Chr.17: 50375548-50375548 on Build GRCh38
rs748101073 Chr.17: 50375579-50375581 on Build GRCh38
rs200504397 Chr.17: 50375273-50375273 on Build GRCh38
rs368526105 Chr.17: 50375474-50375474 on Build GRCh38
rs919853584 Chr.17: 50375195-50375195 on Build GRCh38
rs377462951 Chr.17: 50375586-50375586 on Build GRCh38
rs775021174 Chr.17: 50375189-50375189 on Build GRCh38
rs777335812 Chr.17: 50375567-50375567 on Build GRCh38
rs372340954 Chr.17: 50375196-50375196 on Build GRCh38
rs1029235885 Chr.17: 50375356-50375356 on Build GRCh38
rs746411096 Chr.17: 50375283-50375283 on Build GRCh38
rs747603787 Chr.17: 50375552-50375552 on Build GRCh38
rs770711136 Chr.17: 50375476-50375476 on Build GRCh38
rs756225076 Chr.17: 50375253-50375253 on Build GRCh38
rs149195681 Chr.17: 50375418-50375418 on Build GRCh38
rs746936742 Chr.17: 50375532-50375532 on Build GRCh38
rs755863877 Chr.17: 50375423-50375423 on Build GRCh38
rs553645610 Chr.17: 50375059-50375059 on Build GRCh38
rs987989093 Chr.17: 50375088-50375088 on Build GRCh38
rs780621573 Chr.17: 50375451-50375451 on Build GRCh38
rs138848514 Chr.17: 50375156-50375156 on Build GRCh38
rs764807730 Chr.17: 50375302-50375302 on Build GRCh38
rs935504870 Chr.17: 50375298-50375298 on Build GRCh38
rs1029037769 Chr.17: 50375067-50375067 on Build GRCh38
rs756711970 Chr.17: 50375541-50375541 on Build GRCh38
rs776069771 Chr.17: 50375296-50375296 on Build GRCh38
rs752526564 Chr.17: 50375305-50375322 on Build GRCh38
rs777423518 Chr.17: 50375470-50375478 on Build GRCh38
rs369680589 Chr.17: 50375274-50375274 on Build GRCh38
rs769283796 Chr.17: 50375561-50375561 on Build GRCh38
rs1053990714 Chr.17: 50375304-50375304 on Build GRCh38
rs776243726 Chr.17: 50375580-50375580 on Build GRCh38
rs372148977 Chr.17: 50375588-50375588 on Build GRCh38
rs768197209 Chr.17: 50375295-50375295 on Build GRCh38
rs954675145 Chr.17: 50375075-50375075 on Build GRCh38
rs778157348 Chr.17: 50375255-50375255 on Build GRCh38
rs140517067 Chr.17: 50375404-50375404 on Build GRCh38
rs749620888 Chr.17: 50375256-50375256 on Build GRCh38
rs751504552 Chr.17: 50375339-50375339 on Build GRCh38
rs748401009 Chr.17: 50375251-50375251 on Build GRCh38
rs962883465 Chr.17: 50375108-50375108 on Build GRCh38
rs931265378 Chr.17: 50375200-50375200 on Build GRCh38
rs776322631 Chr.17: 50375201-50375201 on Build GRCh38
rs373305910 Chr.17: 50375327-50375327 on Build GRCh38
rs543903545 Chr.17: 50375215-50375215 on Build GRCh38
rs570990672 Chr.17: 50375515-50375515 on Build GRCh38
rs757617549 Chr.17: 50375533-50375533 on Build GRCh38
rs750149608 Chr.17: 50375214-50375214 on Build GRCh38
rs551030336 Chr.17: 50375466-50375466 on Build GRCh38
rs926760704 Chr.17: 50375125-50375125 on Build GRCh38
rs766214551 Chr.17: 50375332-50375332 on Build GRCh38
rs751542825 Chr.17: 50375227-50375227 on Build GRCh38
rs200348764 Chr.17: 50375184-50375184 on Build GRCh38
rs1026796787 Chr.17: 50375493-50375493 on Build GRCh38
rs755067908 Chr.17: 50375230-50375230 on Build GRCh38
rs752712544 Chr.17: 50375511-50375511 on Build GRCh38
rs766253005 Chr.17: 50375224-50375224 on Build GRCh38
rs9896405 Chr.17: 50375353-50375353 on Build GRCh38
rs368491563 Chr.17: 50375192-50375192 on Build GRCh38
rs773954367 Chr.17: 50375483-50375483 on Build GRCh38
rs143639554 Chr.17: 50375361-50375361 on Build GRCh38
rs201551910 Chr.17: 50375381-50375381 on Build GRCh38
rs767310180 Chr.17: 50375491-50375491 on Build GRCh38
rs201338741 Chr.17: 50375318-50375318 on Build GRCh38
rs751505961 Chr.17: 50375159-50375160 on Build GRCh38
rs3760413 Chr.17: 50375415-50375415 on Build GRCh38
rs17714829 Chr.17: 50375120-50375120 on Build GRCh38
rs541428836 Chr.17: 50375130-50375130 on Build GRCh38
rs762656214 Chr.17: 50375468-50375468 on Build GRCh38
rs759519532 Chr.17: 50375258-50375259 on Build GRCh38
rs765009105 Chr.17: 50375209-50375209 on Build GRCh38
rs772911278 Chr.17: 50375433-50375433 on Build GRCh38
rs17714854 Chr.17: 50375395-50375395 on Build GRCh38

View Full Product Details