Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 64519075-64519622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP95
Entrez Gene ID
90799
Gene Name
centrosomal protein 95
Gene Aliases
CCDC45
UniGene
Hs.569713
Gene Chromosome Location
Chr.17: 64506736-64537951 on Build GRCh38
Transcripts
NM_001316990.1 NM_138363.2 NR_133644.1 XM_005257779.3 XM_005257780.2 XM_006722166.3 XM_011525458.2 XM_017025287.1 XM_017025288.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1316092 c.523G>A p.A175T Chr.17: 64519370-64519370 on Build GRCh38
4498936 c.530C>T p.S177F Chr.17: 64519377-64519377 on Build GRCh38
5149510 c.543C>A p.I181I Chr.17: 64519390-64519390 on Build GRCh38
5364433 c.501C>A p.G167G Chr.17: 64519348-64519348 on Build GRCh38
5387050 c.557_558AC>TT p.D186V Chr.17: 64519404-64519405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782496000 Chr.17: 64519451-64519451 on Build GRCh38
rs1031795680 Chr.17: 64519116-64519116 on Build GRCh38
rs782088529 Chr.17: 64519455-64519455 on Build GRCh38
rs781899752 Chr.17: 64519328-64519328 on Build GRCh38
rs1048762027 Chr.17: 64519332-64519332 on Build GRCh38
rs781961293 Chr.17: 64519410-64519410 on Build GRCh38
rs761868571 Chr.17: 64519142-64519143 on Build GRCh38
rs930394975 Chr.17: 64519317-64519317 on Build GRCh38
rs782002161 Chr.17: 64519443-64519443 on Build GRCh38
rs939190747 Chr.17: 64519248-64519248 on Build GRCh38
rs781825137 Chr.17: 64519347-64519347 on Build GRCh38
rs1053703957 Chr.17: 64519597-64519597 on Build GRCh38
rs565672100 Chr.17: 64519202-64519202 on Build GRCh38
rs782786473 Chr.17: 64519298-64519298 on Build GRCh38
rs370442455 Chr.17: 64519535-64519535 on Build GRCh38
rs943027217 Chr.17: 64519377-64519377 on Build GRCh38
rs970022755 Chr.17: 64519205-64519205 on Build GRCh38
rs541853074 Chr.17: 64519506-64519506 on Build GRCh38
rs188830334 Chr.17: 64519405-64519405 on Build GRCh38
rs782337349 Chr.17: 64519407-64519407 on Build GRCh38
rs782065405 Chr.17: 64519297-64519297 on Build GRCh38
rs9910506 Chr.17: 64519342-64519342 on Build GRCh38
rs782164211 Chr.17: 64519426-64519427 on Build GRCh38
rs145687161 Chr.17: 64519228-64519228 on Build GRCh38
rs972304145 Chr.17: 64519292-64519292 on Build GRCh38
rs781994195 Chr.17: 64519281-64519281 on Build GRCh38
rs782156591 Chr.17: 64519422-64519422 on Build GRCh38
rs782679583 Chr.17: 64519393-64519393 on Build GRCh38
rs781962359 Chr.17: 64519133-64519133 on Build GRCh38
rs782176032 Chr.17: 64519411-64519411 on Build GRCh38
rs782502570 Chr.17: 64519333-64519333 on Build GRCh38
rs369713673 Chr.17: 64519302-64519302 on Build GRCh38
rs957677797 Chr.17: 64519127-64519127 on Build GRCh38
rs999412522 Chr.17: 64519106-64519106 on Build GRCh38
rs782325750 Chr.17: 64519412-64519412 on Build GRCh38
rs782571395 Chr.17: 64519375-64519375 on Build GRCh38
rs781792861 Chr.17: 64519474-64519474 on Build GRCh38
rs781783570 Chr.17: 64519290-64519290 on Build GRCh38
rs999067936 Chr.17: 64519576-64519576 on Build GRCh38
rs919045083 Chr.17: 64519315-64519315 on Build GRCh38
rs782017181 Chr.17: 64519417-64519417 on Build GRCh38
rs782746306 Chr.17: 64519466-64519466 on Build GRCh38
rs782079594 Chr.17: 64519286-64519286 on Build GRCh38
rs184094387 Chr.17: 64519387-64519387 on Build GRCh38
rs75606420 Chr.17: 64519224-64519224 on Build GRCh38
rs782198833 Chr.17: 64519383-64519383 on Build GRCh38
rs782739237 Chr.17: 64519288-64519288 on Build GRCh38
rs562102852 Chr.17: 64519507-64519507 on Build GRCh38
rs782490099 Chr.17: 64519350-64519350 on Build GRCh38
rs781940912 Chr.17: 64519477-64519477 on Build GRCh38
rs554768438 Chr.17: 64519132-64519132 on Build GRCh38
rs181848970 Chr.17: 64519433-64519433 on Build GRCh38
rs16947841 Chr.17: 64519218-64519218 on Build GRCh38
rs548978319 Chr.17: 64519186-64519186 on Build GRCh38
rs782425479 Chr.17: 64519274-64519274 on Build GRCh38

View Full Product Details