Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 80437423-80437978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENDOV
Entrez Gene ID
284131
Gene Name
endonuclease V
Gene Aliases
-
UniGene
Hs.389678
Gene Chromosome Location
Chr.17: 80415117-80438086 on Build GRCh38
Transcripts
NM_001164637.2 NM_173627.4 XM_011524658.2 XM_011524669.2 XM_011524670.1 XM_011524671.2 XM_017024511.1 XM_017024512.1 XM_017024514.1 XM_017024515.1 XM_017024516.1 XM_017024517.1 XM_017024518.1 XM_017024519.1 XM_017024520.1

SNP Detail

SNP ID Chromosome Location
rs560797128 Chr.17: 80437827-80437827 on Build GRCh38
rs566990942 Chr.17: 80437442-80437442 on Build GRCh38
rs41303574 Chr.17: 80437620-80437620 on Build GRCh38
rs969818363 Chr.17: 80437754-80437754 on Build GRCh38
rs377218883 Chr.17: 80437531-80437531 on Build GRCh38
rs1049720787 Chr.17: 80437688-80437688 on Build GRCh38
rs920535411 Chr.17: 80437584-80437584 on Build GRCh38
rs1053404352 Chr.17: 80437923-80437923 on Build GRCh38
rs1000378664 Chr.17: 80437498-80437498 on Build GRCh38
rs574847891 Chr.17: 80437569-80437569 on Build GRCh38
rs746590415 Chr.17: 80437544-80437544 on Build GRCh38
rs747871712 Chr.17: 80437616-80437616 on Build GRCh38
rs1034561208 Chr.17: 80437513-80437514 on Build GRCh38
rs891767407 Chr.17: 80437697-80437697 on Build GRCh38
rs925909774 Chr.17: 80437915-80437915 on Build GRCh38
rs887916091 Chr.17: 80437949-80437949 on Build GRCh38
rs1006351526 Chr.17: 80437954-80437954 on Build GRCh38
rs922692459 Chr.17: 80437853-80437853 on Build GRCh38
rs907680841 Chr.17: 80437628-80437628 on Build GRCh38
rs549382908 Chr.17: 80437612-80437612 on Build GRCh38
rs776279919 Chr.17: 80437583-80437583 on Build GRCh38
rs41303576 Chr.17: 80437635-80437635 on Build GRCh38
rs41303578 Chr.17: 80437735-80437735 on Build GRCh38
rs932729271 Chr.17: 80437875-80437875 on Build GRCh38
rs530159351 Chr.17: 80437784-80437784 on Build GRCh38
rs983368807 Chr.17: 80437828-80437828 on Build GRCh38
rs952279518 Chr.17: 80437752-80437752 on Build GRCh38
rs1046180898 Chr.17: 80437937-80437937 on Build GRCh38
rs932674067 Chr.17: 80437680-80437680 on Build GRCh38
rs1024496523 Chr.17: 80437746-80437746 on Build GRCh38
rs147715261 Chr.17: 80437917-80437917 on Build GRCh38
rs892002047 Chr.17: 80437734-80437734 on Build GRCh38
rs41303580 Chr.17: 80437840-80437840 on Build GRCh38
rs570929707 Chr.17: 80437939-80437939 on Build GRCh38
rs965875203 Chr.17: 80437837-80437837 on Build GRCh38
rs41303572 Chr.17: 80437545-80437545 on Build GRCh38
rs534443838 Chr.17: 80437469-80437469 on Build GRCh38
rs946066910 Chr.17: 80437936-80437936 on Build GRCh38
rs1050193585 Chr.17: 80437879-80437879 on Build GRCh38
rs755378987 Chr.17: 80437740-80437740 on Build GRCh38
rs564914290 Chr.17: 80437698-80437698 on Build GRCh38
rs376777532 Chr.17: 80437946-80437946 on Build GRCh38
rs772972721 Chr.17: 80437851-80437851 on Build GRCh38
rs74720915 Chr.17: 80437727-80437727 on Build GRCh38
rs936503584 Chr.17: 80437905-80437905 on Build GRCh38
rs985944879 Chr.17: 80437859-80437859 on Build GRCh38
rs866597302 Chr.17: 80437582-80437582 on Build GRCh38
rs1015135552 Chr.17: 80437955-80437955 on Build GRCh38
rs1027285022 Chr.17: 80437777-80437777 on Build GRCh38
rs762924908 Chr.17: 80437874-80437874 on Build GRCh38
rs183714012 Chr.17: 80437713-80437713 on Build GRCh38
rs930530433 Chr.17: 80437608-80437608 on Build GRCh38
rs116748335 Chr.17: 80437530-80437530 on Build GRCh38
rs907796663 Chr.17: 80437836-80437836 on Build GRCh38
rs1045867591 Chr.17: 80437947-80437947 on Build GRCh38
rs765656807 Chr.17: 80437855-80437855 on Build GRCh38
rs548601622 Chr.17: 80437796-80437796 on Build GRCh38
rs540065634 Chr.17: 80437637-80437637 on Build GRCh38
rs558864955 Chr.17: 80437502-80437502 on Build GRCh38
rs907319613 Chr.17: 80437736-80437736 on Build GRCh38
rs769662414 Chr.17: 80437650-80437650 on Build GRCh38
rs904760860 Chr.17: 80437443-80437443 on Build GRCh38
rs147575863 Chr.17: 80437622-80437622 on Build GRCh38
rs892381031 Chr.17: 80437931-80437931 on Build GRCh38
rs931834026 Chr.17: 80437585-80437585 on Build GRCh38
rs1031820004 Chr.17: 80437511-80437511 on Build GRCh38
rs1001618611 Chr.17: 80437769-80437769 on Build GRCh38
rs1034507535 Chr.17: 80437751-80437751 on Build GRCh38
rs1031850736 Chr.17: 80437716-80437716 on Build GRCh38
rs562513736 Chr.17: 80437953-80437953 on Build GRCh38
rs952331583 Chr.17: 80437515-80437515 on Build GRCh38
rs538321155 Chr.17: 80437543-80437543 on Build GRCh38
rs976154242 Chr.17: 80437795-80437795 on Build GRCh38

View Full Product Details