Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 35774087-35774669 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCLK1
Entrez Gene ID
9201
Gene Name
doublecortin like kinase 1
Gene Aliases
CL1, CLICK1, DCAMKL1, DCDC3A, DCLK
UniGene
Hs.507755
Gene Chromosome Location
Chr.13: 35768652-36132020 on Build GRCh38
Transcripts
NM_001195415.1 NM_001195416.1 NM_004734.4 XM_005266592.2 XM_006719893.2 XM_017020847.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4775384 c.1239G>A p.S413S Chr.13: 35774598-35774598 on Build GRCh38
3417560 c.1279C>T p.R427C Chr.13: 35774558-35774558 on Build GRCh38
6553908 c.1192G>A p.V398M Chr.13: 35774645-35774645 on Build GRCh38
7233479 c.2181C>T p.T727T Chr.13: 35774651-35774651 on Build GRCh38
1587086 c.1228G>A p.E410K Chr.13: 35774609-35774609 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565397452 Chr.13: 35774476-35774476 on Build GRCh38
rs542604662 Chr.13: 35774277-35774277 on Build GRCh38
rs767914664 Chr.13: 35774586-35774586 on Build GRCh38
rs752171277 Chr.13: 35774139-35774139 on Build GRCh38
rs1033491991 Chr.13: 35774326-35774326 on Build GRCh38
rs112739939 Chr.13: 35774630-35774630 on Build GRCh38
rs910844578 Chr.13: 35774167-35774167 on Build GRCh38
rs763910744 Chr.13: 35774231-35774231 on Build GRCh38
rs375500006 Chr.13: 35774520-35774520 on Build GRCh38
rs1040631172 Chr.13: 35774557-35774557 on Build GRCh38
rs976211824 Chr.13: 35774172-35774172 on Build GRCh38
rs377204170 Chr.13: 35774625-35774625 on Build GRCh38
rs569155696 Chr.13: 35774610-35774610 on Build GRCh38
rs757007588 Chr.13: 35774635-35774635 on Build GRCh38
rs777529442 Chr.13: 35774492-35774492 on Build GRCh38
rs1013519904 Chr.13: 35774343-35774343 on Build GRCh38
rs775139421 Chr.13: 35774225-35774225 on Build GRCh38
rs182194897 Chr.13: 35774409-35774409 on Build GRCh38
rs761166313 Chr.13: 35774605-35774605 on Build GRCh38
rs755788470 Chr.13: 35774568-35774568 on Build GRCh38
rs369449567 Chr.13: 35774599-35774599 on Build GRCh38
rs745824879 Chr.13: 35774558-35774558 on Build GRCh38
rs762850996 Chr.13: 35774116-35774116 on Build GRCh38
rs959582735 Chr.13: 35774329-35774329 on Build GRCh38
rs902064643 Chr.13: 35774445-35774445 on Build GRCh38
rs545597760 Chr.13: 35774219-35774219 on Build GRCh38
rs901526765 Chr.13: 35774197-35774197 on Build GRCh38
rs950937477 Chr.13: 35774472-35774472 on Build GRCh38
rs769680190 Chr.13: 35774642-35774642 on Build GRCh38
rs909188447 Chr.13: 35774280-35774280 on Build GRCh38
rs370155230 Chr.13: 35774320-35774320 on Build GRCh38
rs746686672 Chr.13: 35774634-35774634 on Build GRCh38
rs376910686 Chr.13: 35774479-35774479 on Build GRCh38
rs145262053 Chr.13: 35774626-35774626 on Build GRCh38
rs943619976 Chr.13: 35774169-35774169 on Build GRCh38
rs138931047 Chr.13: 35774139-35774140 on Build GRCh38
rs1007794786 Chr.13: 35774123-35774123 on Build GRCh38
rs924420154 Chr.13: 35774580-35774580 on Build GRCh38
rs919377986 Chr.13: 35774474-35774474 on Build GRCh38
rs764103051 Chr.13: 35774501-35774501 on Build GRCh38
rs773678179 Chr.13: 35774591-35774591 on Build GRCh38
rs546690018 Chr.13: 35774463-35774463 on Build GRCh38
rs938043404 Chr.13: 35774282-35774283 on Build GRCh38
rs990741518 Chr.13: 35774493-35774493 on Build GRCh38
rs1007201349 Chr.13: 35774428-35774428 on Build GRCh38
rs915806058 Chr.13: 35774293-35774293 on Build GRCh38
rs372450457 Chr.13: 35774616-35774616 on Build GRCh38
rs780857999 Chr.13: 35774636-35774636 on Build GRCh38
rs578080649 Chr.13: 35774174-35774174 on Build GRCh38
rs186689649 Chr.13: 35774161-35774161 on Build GRCh38
rs533695794 Chr.13: 35774142-35774142 on Build GRCh38
rs749915788 Chr.13: 35774486-35774486 on Build GRCh38
rs372395267 Chr.13: 35774607-35774607 on Build GRCh38
rs934402210 Chr.13: 35774358-35774358 on Build GRCh38
rs139135390 Chr.13: 35774489-35774489 on Build GRCh38
rs115023965 Chr.13: 35774241-35774241 on Build GRCh38
rs943108835 Chr.13: 35774618-35774618 on Build GRCh38
rs762594588 Chr.13: 35774435-35774435 on Build GRCh38
rs575568336 Chr.13: 35774265-35774265 on Build GRCh38
rs1054706373 Chr.13: 35774609-35774609 on Build GRCh38
rs528471820 Chr.13: 35774386-35774386 on Build GRCh38
rs532182307 Chr.13: 35774480-35774480 on Build GRCh38
rs776582217 Chr.13: 35774527-35774527 on Build GRCh38
rs1054233216 Chr.13: 35774369-35774369 on Build GRCh38
rs189537845 Chr.13: 35774283-35774283 on Build GRCh38
rs971426667 Chr.13: 35774460-35774460 on Build GRCh38
rs745764381 Chr.13: 35774640-35774640 on Build GRCh38
rs9574561 Chr.13: 35774357-35774357 on Build GRCh38
rs949973263 Chr.13: 35774324-35774324 on Build GRCh38
rs762116664 Chr.13: 35774583-35774583 on Build GRCh38
rs774312251 Chr.13: 35774581-35774581 on Build GRCh38

View Full Product Details