Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14746773-14747051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRE2
Entrez Gene ID
30817
Gene Name
adhesion G protein-coupled receptor E2
Gene Aliases
CD312, EMR2, VBU
UniGene
Hs.531619
Gene Chromosome Location
Chr.19: 14732697-14778541 on Build GRCh38
Transcripts
NM_001271052.1 NM_013447.3 XM_011527948.2 XM_011527949.2 XM_011527951.2 XM_011527952.2 XM_011527953.2 XM_011527954.2 XM_011527955.2 XM_017026726.1 XM_017026727.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3670219 c.2031G>A p.W677* Chr.19: 14746956-14746956 on Build GRCh38
7422968 c.2029T>A p.W677R Chr.19: 14746958-14746958 on Build GRCh38
7366225 c.2030G>A p.W677* Chr.19: 14746957-14746957 on Build GRCh38
6653911 c.2068G>T p.G690* Chr.19: 14746919-14746919 on Build GRCh38
5899756 c.2065C>T p.L689F Chr.19: 14746922-14746922 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs200942496 Chr.19: 14746953-14746953 on Build GRCh38
rs34186967 Chr.19: 14746979-14746980 on Build GRCh38
rs774513835 Chr.19: 14746922-14746922 on Build GRCh38
rs772418492 Chr.19: 14746927-14746927 on Build GRCh38
rs1052171115 Chr.19: 14746837-14746837 on Build GRCh38
rs763674532 Chr.19: 14746971-14746972 on Build GRCh38
rs772149607 Chr.19: 14746865-14746865 on Build GRCh38
rs570053595 Chr.19: 14747028-14747028 on Build GRCh38
rs1048310091 Chr.19: 14746841-14746841 on Build GRCh38
rs751228441 Chr.19: 14746869-14746869 on Build GRCh38
rs61732001 Chr.19: 14746908-14746908 on Build GRCh38
rs377454798 Chr.19: 14746907-14746907 on Build GRCh38
rs201569047 Chr.19: 14746885-14746885 on Build GRCh38
rs777202442 Chr.19: 14746941-14746941 on Build GRCh38
rs185356117 Chr.19: 14746855-14746855 on Build GRCh38
rs370665322 Chr.19: 14746950-14746950 on Build GRCh38
rs1016953564 Chr.19: 14746979-14746979 on Build GRCh38
rs767526220 Chr.19: 14746980-14746980 on Build GRCh38
rs935000115 Chr.19: 14746806-14746806 on Build GRCh38
rs754506183 Chr.19: 14746852-14746852 on Build GRCh38
rs749136916 Chr.19: 14746911-14746911 on Build GRCh38
rs748439255 Chr.19: 14746926-14746926 on Build GRCh38
rs371200658 Chr.19: 14746915-14746915 on Build GRCh38
rs749260103 Chr.19: 14747013-14747013 on Build GRCh38
rs779645867 Chr.19: 14747011-14747011 on Build GRCh38
rs373118812 Chr.19: 14746882-14746882 on Build GRCh38
rs377637436 Chr.19: 14746939-14746939 on Build GRCh38
rs200940536 Chr.19: 14746931-14746931 on Build GRCh38
rs766888206 Chr.19: 14746854-14746854 on Build GRCh38
rs751390004 Chr.19: 14746977-14746978 on Build GRCh38
rs893584283 Chr.19: 14746850-14746850 on Build GRCh38
rs188483876 Chr.19: 14746862-14746862 on Build GRCh38
rs536793741 Chr.19: 14746967-14746967 on Build GRCh38
rs963157150 Chr.19: 14746993-14746993 on Build GRCh38
rs773762022 Chr.19: 14746930-14746930 on Build GRCh38
rs767130765 Chr.19: 14746960-14746960 on Build GRCh38
rs907856253 Chr.19: 14747027-14747027 on Build GRCh38
rs996638603 Chr.19: 14746893-14746894 on Build GRCh38
rs993394400 Chr.19: 14746990-14746990 on Build GRCh38
rs763205246 Chr.19: 14747019-14747019 on Build GRCh38
rs978981150 Chr.19: 14746948-14746948 on Build GRCh38
rs781035186 Chr.19: 14746893-14746893 on Build GRCh38
rs970505824 Chr.19: 14747026-14747026 on Build GRCh38
rs756940070 Chr.19: 14746955-14746955 on Build GRCh38
rs755966222 Chr.19: 14746980-14746980 on Build GRCh38
rs117913748 Chr.19: 14746861-14746861 on Build GRCh38
rs398040937 Chr.19: 14746980-14746981 on Build GRCh38
rs956341932 Chr.19: 14746989-14746989 on Build GRCh38

View Full Product Details