Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 49286152-49286660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDADC1
Entrez Gene ID
81602
Gene Name
cytidine and dCMP deaminase domain containing 1
Gene Aliases
NYD-SP15, bA103J18.1
UniGene
Hs.388220
Gene Chromosome Location
Chr.13: 49247875-49293486 on Build GRCh38
Transcripts
NM_001193478.1 NM_030911.3 NR_036439.1 XM_006719871.3 XM_011535250.2 XM_011535251.1 XM_011535253.1 XM_017020778.1 XM_017020779.1 XM_017020780.1 XM_017020781.1 XM_017020782.1 XM_017020783.1 XM_017020784.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3469185 c.1468G>A p.E490K Chr.13: 49286279-49286279 on Build GRCh38
4527066 c.1438G>A p.G480S Chr.13: 49286249-49286249 on Build GRCh38
6000949 c.1458T>C p.P486P Chr.13: 49286269-49286269 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1042776263 Chr.13: 49286547-49286547 on Build GRCh38
rs762047248 Chr.13: 49286324-49286324 on Build GRCh38
rs775143364 Chr.13: 49286301-49286301 on Build GRCh38
rs199798467 Chr.13: 49286259-49286259 on Build GRCh38
rs1036766845 Chr.13: 49286381-49286381 on Build GRCh38
rs941782494 Chr.13: 49286440-49286440 on Build GRCh38
rs201465059 Chr.13: 49286291-49286291 on Build GRCh38
rs896929019 Chr.13: 49286411-49286411 on Build GRCh38
rs760999977 Chr.13: 49286215-49286215 on Build GRCh38
rs117126694 Chr.13: 49286457-49286457 on Build GRCh38
rs185892130 Chr.13: 49286361-49286361 on Build GRCh38
rs753434032 Chr.13: 49286228-49286228 on Build GRCh38
rs1044084943 Chr.13: 49286466-49286466 on Build GRCh38
rs768178196 Chr.13: 49286202-49286202 on Build GRCh38
rs903819848 Chr.13: 49286548-49286548 on Build GRCh38
rs1057006800 Chr.13: 49286509-49286509 on Build GRCh38
rs757874124 Chr.13: 49286263-49286263 on Build GRCh38
rs769048385 Chr.13: 49286294-49286294 on Build GRCh38
rs537917316 Chr.13: 49286310-49286310 on Build GRCh38
rs777997188 Chr.13: 49286257-49286257 on Build GRCh38
rs752257540 Chr.13: 49286251-49286251 on Build GRCh38
rs546419459 Chr.13: 49286520-49286520 on Build GRCh38
rs778421457 Chr.13: 49286246-49286246 on Build GRCh38
rs938490218 Chr.13: 49286486-49286486 on Build GRCh38
rs9316437 Chr.13: 49286555-49286555 on Build GRCh38
rs190608705 Chr.13: 49286532-49286532 on Build GRCh38
rs1054796694 Chr.13: 49286233-49286235 on Build GRCh38
rs780999420 Chr.13: 49286283-49286283 on Build GRCh38
rs555383356 Chr.13: 49286195-49286195 on Build GRCh38
rs754621429 Chr.13: 49286231-49286231 on Build GRCh38
rs778532704 Chr.13: 49286281-49286284 on Build GRCh38
rs769130169 Chr.13: 49286216-49286217 on Build GRCh38
rs772262138 Chr.13: 49286327-49286327 on Build GRCh38
rs780567865 Chr.13: 49286522-49286522 on Build GRCh38
rs766704650 Chr.13: 49286219-49286219 on Build GRCh38
rs777756150 Chr.13: 49286277-49286277 on Build GRCh38
rs964902911 Chr.13: 49286601-49286601 on Build GRCh38
rs746954267 Chr.13: 49286279-49286279 on Build GRCh38
rs779896184 Chr.13: 49286436-49286436 on Build GRCh38
rs746583264 Chr.13: 49286333-49286334 on Build GRCh38
rs1048216586 Chr.13: 49286471-49286471 on Build GRCh38
rs527972888 Chr.13: 49286290-49286290 on Build GRCh38
rs995265156 Chr.13: 49286444-49286444 on Build GRCh38
rs1017671685 Chr.13: 49286597-49286597 on Build GRCh38
rs761961316 Chr.13: 49286200-49286200 on Build GRCh38
rs774522211 Chr.13: 49286185-49286185 on Build GRCh38
rs751016446 Chr.13: 49286472-49286472 on Build GRCh38
rs750933810 Chr.13: 49286209-49286209 on Build GRCh38
rs770646478 Chr.13: 49286282-49286282 on Build GRCh38
rs754450237 Chr.13: 49286485-49286485 on Build GRCh38
rs745841828 Chr.13: 49286288-49286291 on Build GRCh38
rs938266720 Chr.13: 49286211-49286211 on Build GRCh38

View Full Product Details