Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 49633645-49634221 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL11
Entrez Gene ID
115761
Gene Name
ADP ribosylation factor like GTPase 11
Gene Aliases
ARLTS1
UniGene
Hs.558599
Gene Chromosome Location
Chr.13: 49628299-49633872 on Build GRCh38
Transcripts
NM_138450.5

SNP Detail

SNP ID Chromosome Location
rs374853258 Chr.13: 49633859-49633859 on Build GRCh38
rs972844339 Chr.13: 49634074-49634074 on Build GRCh38
rs942986213 Chr.13: 49634122-49634122 on Build GRCh38
rs949524636 Chr.13: 49633719-49633719 on Build GRCh38
rs760990066 Chr.13: 49634195-49634195 on Build GRCh38
rs933643065 Chr.13: 49634200-49634200 on Build GRCh38
rs945749278 Chr.13: 49634182-49634182 on Build GRCh38
rs114815894 Chr.13: 49633963-49633963 on Build GRCh38
rs747360841 Chr.13: 49634073-49634078 on Build GRCh38
rs775942330 Chr.13: 49634124-49634124 on Build GRCh38
rs1039349453 Chr.13: 49633756-49633756 on Build GRCh38
rs904309794 Chr.13: 49634193-49634193 on Build GRCh38
rs189519981 Chr.13: 49633814-49633814 on Build GRCh38
rs181187405 Chr.13: 49633979-49633979 on Build GRCh38
rs569893144 Chr.13: 49633812-49633812 on Build GRCh38
rs765279738 Chr.13: 49633775-49633776 on Build GRCh38
rs958633819 Chr.13: 49633973-49633975 on Build GRCh38
rs946226480 Chr.13: 49634059-49634059 on Build GRCh38
rs901361753 Chr.13: 49634137-49634137 on Build GRCh38
rs900873958 Chr.13: 49634190-49634190 on Build GRCh38
rs747849621 Chr.13: 49633727-49633727 on Build GRCh38
rs913446773 Chr.13: 49634037-49634037 on Build GRCh38
rs540677934 Chr.13: 49634147-49634147 on Build GRCh38
rs913433821 Chr.13: 49633709-49633709 on Build GRCh38
rs539623388 Chr.13: 49633790-49633790 on Build GRCh38
rs369330204 Chr.13: 49633838-49633838 on Build GRCh38
rs998131693 Chr.13: 49633766-49633766 on Build GRCh38
rs556618877 Chr.13: 49634066-49634066 on Build GRCh38
rs781073355 Chr.13: 49633683-49633683 on Build GRCh38
rs918612482 Chr.13: 49633770-49633770 on Build GRCh38
rs911457311 Chr.13: 49634107-49634107 on Build GRCh38
rs963467849 Chr.13: 49633916-49633916 on Build GRCh38
rs999895449 Chr.13: 49633958-49633958 on Build GRCh38
rs968417920 Chr.13: 49633937-49633937 on Build GRCh38
rs370769835 Chr.13: 49634095-49634095 on Build GRCh38
rs772415905 Chr.13: 49634096-49634096 on Build GRCh38
rs61959910 Chr.13: 49633749-49633749 on Build GRCh38
rs868532095 Chr.13: 49634077-49634077 on Build GRCh38
rs541943665 Chr.13: 49634019-49634019 on Build GRCh38
rs769609521 Chr.13: 49633748-49633748 on Build GRCh38
rs925711162 Chr.13: 49634081-49634081 on Build GRCh38
rs758310167 Chr.13: 49634073-49634080 on Build GRCh38
rs551979043 Chr.13: 49633788-49633788 on Build GRCh38
rs796770107 Chr.13: 49633765-49633765 on Build GRCh38
rs534326513 Chr.13: 49633929-49633929 on Build GRCh38
rs372910479 Chr.13: 49633848-49633848 on Build GRCh38
rs757539921 Chr.13: 49634080-49634081 on Build GRCh38
rs1020348021 Chr.13: 49633935-49633935 on Build GRCh38
rs992645584 Chr.13: 49633980-49633980 on Build GRCh38
rs565742435 Chr.13: 49633898-49633898 on Build GRCh38
rs960019773 Chr.13: 49633987-49633987 on Build GRCh38
rs1016770130 Chr.13: 49633849-49633849 on Build GRCh38
rs566059420 Chr.13: 49633692-49633692 on Build GRCh38
rs1043201413 Chr.13: 49633746-49633746 on Build GRCh38

View Full Product Details