Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54889716-54890315 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCAR
Entrez Gene ID
2204
Gene Name
Fc fragment of IgA receptor
Gene Aliases
CD89, CTB-61M7.2, FcalphaRI
UniGene
Hs.659872
Gene Chromosome Location
Chr.19: 54874093-54891420 on Build GRCh38
Transcripts
NM_002000.3 NM_133269.3 NM_133271.3 NM_133272.3 NM_133273.3 NM_133274.3 NM_133277.3 NM_133278.3 XM_011526625.2 XM_017026473.1 XM_017026474.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1631088 c.731T>C p.I244T Chr.19: 54889730-54889730 on Build GRCh38
4581416 c.739G>T p.E247* Chr.19: 54889738-54889738 on Build GRCh38
6525390 c.797C>A p.P266Q Chr.19: 54889796-54889796 on Build GRCh38
6539293 c.835T>G p.F279V Chr.19: 54889834-54889834 on Build GRCh38
6566078 c.761C>G p.A254G Chr.19: 54889760-54889760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780137525 Chr.19: 54890222-54890222 on Build GRCh38
rs776384079 Chr.19: 54889779-54889779 on Build GRCh38
rs751353715 Chr.19: 54889749-54889749 on Build GRCh38
rs768079215 Chr.19: 54889908-54889908 on Build GRCh38
rs895490816 Chr.19: 54890095-54890095 on Build GRCh38
rs776595662 Chr.19: 54890120-54890121 on Build GRCh38
rs758323699 Chr.19: 54889745-54889745 on Build GRCh38
rs149025092 Chr.19: 54889825-54889825 on Build GRCh38
rs1056549712 Chr.19: 54889973-54889973 on Build GRCh38
rs900027767 Chr.19: 54889753-54889753 on Build GRCh38
rs759241783 Chr.19: 54889784-54889784 on Build GRCh38
rs75257909 Chr.19: 54889738-54889738 on Build GRCh38
rs79267258 Chr.19: 54889835-54889835 on Build GRCh38
rs896597394 Chr.19: 54889995-54889995 on Build GRCh38
rs748488778 Chr.19: 54889776-54889776 on Build GRCh38
rs149418497 Chr.19: 54890246-54890246 on Build GRCh38
rs60304316 Chr.19: 54890108-54890108 on Build GRCh38
rs58560391 Chr.19: 54890081-54890081 on Build GRCh38
rs781322512 Chr.19: 54889752-54889752 on Build GRCh38
rs976788225 Chr.19: 54890187-54890187 on Build GRCh38
rs59309328 Chr.19: 54890115-54890115 on Build GRCh38
rs145316840 Chr.19: 54890030-54890030 on Build GRCh38
rs986062757 Chr.19: 54889899-54889899 on Build GRCh38
rs370887553 Chr.19: 54889786-54889786 on Build GRCh38
rs955325418 Chr.19: 54890098-54890098 on Build GRCh38
rs756301132 Chr.19: 54889833-54889833 on Build GRCh38
rs59587280 Chr.19: 54890260-54890260 on Build GRCh38
rs587738433 Chr.19: 54889889-54889889 on Build GRCh38
rs1014016859 Chr.19: 54889805-54889805 on Build GRCh38
rs1013985904 Chr.19: 54890103-54890103 on Build GRCh38
rs61672126 Chr.19: 54890252-54890252 on Build GRCh38
rs775018175 Chr.19: 54889907-54889907 on Build GRCh38
rs115424230 Chr.19: 54889910-54889910 on Build GRCh38
rs988123114 Chr.19: 54889961-54889961 on Build GRCh38
rs370725161 Chr.19: 54889882-54889882 on Build GRCh38
rs184574417 Chr.19: 54890277-54890277 on Build GRCh38
rs374427185 Chr.19: 54889965-54889965 on Build GRCh38
rs16986050 Chr.19: 54889804-54889804 on Build GRCh38
rs139147488 Chr.19: 54889785-54889785 on Build GRCh38
rs1022872847 Chr.19: 54889789-54889789 on Build GRCh38
rs779411170 Chr.19: 54889862-54889862 on Build GRCh38
rs748435350 Chr.19: 54889874-54889875 on Build GRCh38
rs780089473 Chr.19: 54889757-54889757 on Build GRCh38
rs924228928 Chr.19: 54890190-54890190 on Build GRCh38
rs77103719 Chr.19: 54889758-54889758 on Build GRCh38
rs375614240 Chr.19: 54889803-54889803 on Build GRCh38
rs994274742 Chr.19: 54890089-54890089 on Build GRCh38
rs59103589 Chr.19: 54890046-54890046 on Build GRCh38
rs1020924121 Chr.19: 54890168-54890168 on Build GRCh38
rs972474242 Chr.19: 54889836-54889836 on Build GRCh38
rs888093707 Chr.19: 54889780-54889780 on Build GRCh38
rs375545045 Chr.19: 54889859-54889859 on Build GRCh38
rs587711440 Chr.19: 54890022-54890022 on Build GRCh38
rs1048626756 Chr.19: 54890062-54890062 on Build GRCh38
rs772571031 Chr.19: 54889778-54889778 on Build GRCh38
rs753943019 Chr.19: 54889840-54889840 on Build GRCh38
rs762426689 Chr.19: 54889795-54889795 on Build GRCh38
rs757055373 Chr.19: 54889751-54889751 on Build GRCh38
rs748634993 Chr.19: 54889868-54889868 on Build GRCh38
rs587640721 Chr.19: 54890245-54890245 on Build GRCh38
rs935331665 Chr.19: 54889930-54889930 on Build GRCh38
rs968216707 Chr.19: 54890124-54890124 on Build GRCh38
rs766410785 Chr.19: 54889769-54889769 on Build GRCh38
rs61735070 Chr.19: 54889796-54889796 on Build GRCh38
rs759577351 Chr.19: 54890259-54890260 on Build GRCh38
rs752881089 Chr.19: 54890077-54890077 on Build GRCh38
rs587735181 Chr.19: 54889925-54889925 on Build GRCh38
rs369460200 Chr.19: 54889880-54889880 on Build GRCh38
rs587763431 Chr.19: 54890126-54890126 on Build GRCh38
rs587654005 Chr.19: 54889954-54889954 on Build GRCh38
rs112638842 Chr.19: 54890090-54890090 on Build GRCh38
rs769616486 Chr.19: 54889841-54889841 on Build GRCh38
rs112351594 Chr.19: 54889962-54889962 on Build GRCh38
rs905691356 Chr.19: 54890109-54890109 on Build GRCh38
rs769204676 Chr.19: 54889766-54889766 on Build GRCh38
rs764003301 Chr.19: 54889746-54889746 on Build GRCh38
rs767318512 Chr.19: 54889814-54889814 on Build GRCh38
rs587619994 Chr.19: 54889904-54889904 on Build GRCh38
rs761552903 Chr.19: 54889811-54889811 on Build GRCh38
rs201910524 Chr.19: 54889755-54889755 on Build GRCh38
rs1005394458 Chr.19: 54890049-54890049 on Build GRCh38
rs746116337 Chr.19: 54889754-54889754 on Build GRCh38
rs377578830 Chr.19: 54889871-54889871 on Build GRCh38
rs965377681 Chr.19: 54890179-54890179 on Build GRCh38

View Full Product Details