Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186786421-186786819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4A2
Entrez Gene ID
1974
Gene Name
eukaryotic translation initiation factor 4A2
Gene Aliases
BM-010, DDX2B, EIF4A, EIF4F, eIF-4A-II, eIF4A-II
UniGene
Hs.518475
Gene Chromosome Location
Chr.3: 186783572-186789897 on Build GRCh38
Transcripts
NM_001967.3 NR_031650.1 NR_002989.1

Gene Symbol
MIR1248
Entrez Gene ID
100302143
Gene Name
microRNA 1248
Gene Aliases
MIRN1248, hsa-mir-1248, mir-1248
UniGene
Gene Chromosome Location
Chr.3: 186786672-186786777 on Build GRCh38
Transcripts
NM_001967.3 NR_031650.1 NR_002989.1

Gene Symbol
SNORA81
Entrez Gene ID
677847
Gene Name
small nucleolar RNA, H/ACA box 81
Gene Aliases
HBI-61
UniGene
Hs.688848
Gene Chromosome Location
Chr.3: 186786675-186786852 on Build GRCh38
Transcripts
NM_001967.3 NR_031650.1 NR_002989.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6977782 c.628-29_771+172del345 p.? Chr.3: 186786473-186786817 on Build GRCh38
381531 c.653C>T p.P218L Chr.3: 186786527-186786527 on Build GRCh38
6978656 c.733G>C p.E245Q Chr.3: 186786607-186786607 on Build GRCh38
4920164 c.641C>T p.S214F Chr.3: 186786515-186786515 on Build GRCh38
3846967 c.769G>C p.E257Q Chr.3: 186786643-186786643 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746518055 Chr.3: 186786742-186786742 on Build GRCh38
rs756129308 Chr.3: 186786708-186786708 on Build GRCh38
rs757125227 Chr.3: 186786753-186786754 on Build GRCh38
rs73063489 Chr.3: 186786743-186786743 on Build GRCh38
rs754020514 Chr.3: 186786711-186786711 on Build GRCh38
rs772632941 Chr.3: 186786719-186786719 on Build GRCh38
rs1020529112 Chr.3: 186786792-186786792 on Build GRCh38
rs376257420 Chr.3: 186786714-186786714 on Build GRCh38
rs529062650 Chr.3: 186786748-186786748 on Build GRCh38
rs572865136 Chr.3: 186786688-186786688 on Build GRCh38
rs1035957912 Chr.3: 186786490-186786490 on Build GRCh38
rs749859047 Chr.3: 186786478-186786478 on Build GRCh38
rs778952832 Chr.3: 186786767-186786770 on Build GRCh38
rs765590984 Chr.3: 186786794-186786794 on Build GRCh38
rs751074159 Chr.3: 186786489-186786489 on Build GRCh38
rs781307843 Chr.3: 186786579-186786579 on Build GRCh38
rs776351814 Chr.3: 186786685-186786685 on Build GRCh38
rs757643215 Chr.3: 186786712-186786712 on Build GRCh38
rs890606179 Chr.3: 186786674-186786674 on Build GRCh38
rs139644081 Chr.3: 186786627-186786627 on Build GRCh38
rs760776293 Chr.3: 186786779-186786779 on Build GRCh38
rs896079583 Chr.3: 186786784-186786784 on Build GRCh38
rs367929995 Chr.3: 186786769-186786769 on Build GRCh38
rs750389476 Chr.3: 186786793-186786794 on Build GRCh38
rs964422017 Chr.3: 186786653-186786653 on Build GRCh38
rs749091759 Chr.3: 186786748-186786752 on Build GRCh38
rs757442610 Chr.3: 186786787-186786787 on Build GRCh38
rs746340390 Chr.3: 186786667-186786667 on Build GRCh38
rs548165979 Chr.3: 186786704-186786704 on Build GRCh38
rs771533650 Chr.3: 186786670-186786670 on Build GRCh38
rs768764819 Chr.3: 186786677-186786679 on Build GRCh38
rs905889028 Chr.3: 186786470-186786470 on Build GRCh38
rs371718873 Chr.3: 186786504-186786504 on Build GRCh38
rs923191799 Chr.3: 186786669-186786669 on Build GRCh38
rs776443366 Chr.3: 186786744-186786744 on Build GRCh38
rs746405150 Chr.3: 186786718-186786718 on Build GRCh38
rs769515729 Chr.3: 186786752-186786752 on Build GRCh38
rs774782539 Chr.3: 186786484-186786484 on Build GRCh38
rs755032307 Chr.3: 186786571-186786571 on Build GRCh38
rs569284255 Chr.3: 186786650-186786653 on Build GRCh38
rs568460640 Chr.3: 186786761-186786761 on Build GRCh38
rs1000742171 Chr.3: 186786795-186786795 on Build GRCh38
rs373986173 Chr.3: 186786496-186786496 on Build GRCh38
rs941841627 Chr.3: 186786733-186786733 on Build GRCh38
rs368617464 Chr.3: 186786658-186786658 on Build GRCh38
rs754110393 Chr.3: 186786781-186786781 on Build GRCh38
rs752808710 Chr.3: 186786698-186786698 on Build GRCh38
rs761728419 Chr.3: 186786687-186786687 on Build GRCh38
rs760474964 Chr.3: 186786681-186786681 on Build GRCh38
rs374023366 Chr.3: 186786754-186786754 on Build GRCh38
rs764112971 Chr.3: 186786780-186786780 on Build GRCh38
rs765266132 Chr.3: 186786529-186786529 on Build GRCh38
rs868809630 Chr.3: 186786617-186786617 on Build GRCh38
rs900512595 Chr.3: 186786652-186786652 on Build GRCh38
rs367898551 Chr.3: 186786497-186786497 on Build GRCh38
rs764445015 Chr.3: 186786664-186786666 on Build GRCh38
rs974353983 Chr.3: 186786655-186786655 on Build GRCh38
rs751386721 Chr.3: 186786778-186786778 on Build GRCh38
rs771414497 Chr.3: 186786483-186786483 on Build GRCh38
rs202149708 Chr.3: 186786680-186786680 on Build GRCh38
rs779320192 Chr.3: 186786664-186786664 on Build GRCh38
rs757057055 Chr.3: 186786508-186786508 on Build GRCh38
rs767590752 Chr.3: 186786651-186786652 on Build GRCh38
rs750374080 Chr.3: 186786690-186786690 on Build GRCh38
rs932030011 Chr.3: 186786656-186786656 on Build GRCh38
rs188469767 Chr.3: 186786485-186786485 on Build GRCh38
rs766352944 Chr.3: 186786771-186786771 on Build GRCh38
rs929840704 Chr.3: 186786491-186786491 on Build GRCh38
rs753546148 Chr.3: 186786729-186786729 on Build GRCh38
rs773534417 Chr.3: 186786474-186786474 on Build GRCh38
rs11538612 Chr.3: 186786618-186786618 on Build GRCh38
rs779492327 Chr.3: 186786737-186786737 on Build GRCh38
rs781303236 Chr.3: 186786598-186786598 on Build GRCh38
rs749096721 Chr.3: 186786793-186786793 on Build GRCh38
rs545904669 Chr.3: 186786633-186786633 on Build GRCh38
rs760795365 Chr.3: 186786662-186786663 on Build GRCh38
rs376723460 Chr.3: 186786495-186786495 on Build GRCh38
rs770191956 Chr.3: 186786466-186786466 on Build GRCh38
rs750332310 Chr.3: 186786530-186786530 on Build GRCh38
rs376591367 Chr.3: 186786694-186786694 on Build GRCh38
rs181852973 Chr.3: 186786650-186786650 on Build GRCh38
rs768431834 Chr.3: 186786683-186786683 on Build GRCh38
rs749667058 Chr.3: 186786645-186786645 on Build GRCh38
rs538395242 Chr.3: 186786710-186786710 on Build GRCh38
rs372744748 Chr.3: 186786676-186786676 on Build GRCh38
rs752862272 Chr.3: 186786658-186786659 on Build GRCh38

View Full Product Details