Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70335786-70336382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABS1
Entrez Gene ID
85438
Gene Name
calcium binding protein, spermatid associated 1
Gene Aliases
C4orf35, CLPH, NYD-SP26
UniGene
Hs.120316
Gene Chromosome Location
Chr.4: 70334954-70337116 on Build GRCh38
Transcripts
NM_033122.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7249804 c.1124C>A p.T375N Chr.4: 70336163-70336163 on Build GRCh38
3605426 c.1010C>T p.S337L Chr.4: 70336049-70336049 on Build GRCh38
4567854 c.897_898GG>AA p.M299_E300>IK Chr.4: 70335936-70335937 on Build GRCh38
5805274 c.768A>T p.T256T Chr.4: 70335807-70335807 on Build GRCh38
4520902 c.1082G>A p.G361E Chr.4: 70336121-70336121 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757900279 Chr.4: 70335840-70335840 on Build GRCh38
rs752623263 Chr.4: 70336197-70336197 on Build GRCh38
rs925260963 Chr.4: 70336210-70336210 on Build GRCh38
rs879333550 Chr.4: 70335922-70335922 on Build GRCh38
rs775476199 Chr.4: 70335869-70335869 on Build GRCh38
rs200105962 Chr.4: 70335902-70335902 on Build GRCh38
rs965747161 Chr.4: 70335954-70335954 on Build GRCh38
rs1036788601 Chr.4: 70336311-70336311 on Build GRCh38
rs945420066 Chr.4: 70336167-70336167 on Build GRCh38
rs892646364 Chr.4: 70335934-70335934 on Build GRCh38
rs774967853 Chr.4: 70336151-70336151 on Build GRCh38
rs772877782 Chr.4: 70336263-70336263 on Build GRCh38
rs773936121 Chr.4: 70336145-70336146 on Build GRCh38
rs191296434 Chr.4: 70336161-70336161 on Build GRCh38
rs889989871 Chr.4: 70335929-70335929 on Build GRCh38
rs139939232 Chr.4: 70336009-70336009 on Build GRCh38
rs371605454 Chr.4: 70335825-70335825 on Build GRCh38
rs759844419 Chr.4: 70335913-70335913 on Build GRCh38
rs13039 Chr.4: 70336226-70336226 on Build GRCh38
rs146512314 Chr.4: 70335945-70335945 on Build GRCh38
rs151169789 Chr.4: 70336041-70336041 on Build GRCh38
rs756810359 Chr.4: 70336028-70336028 on Build GRCh38
rs771164751 Chr.4: 70336008-70336008 on Build GRCh38
rs188181084 Chr.4: 70335971-70335971 on Build GRCh38
rs201451430 Chr.4: 70336181-70336181 on Build GRCh38
rs750096993 Chr.4: 70336048-70336048 on Build GRCh38
rs773344110 Chr.4: 70335995-70335995 on Build GRCh38
rs926559363 Chr.4: 70336094-70336094 on Build GRCh38
rs779915158 Chr.4: 70336052-70336052 on Build GRCh38
rs1025906109 Chr.4: 70335968-70335968 on Build GRCh38
rs759930150 Chr.4: 70336010-70336010 on Build GRCh38
rs1041286516 Chr.4: 70335999-70335999 on Build GRCh38
rs376560270 Chr.4: 70336245-70336245 on Build GRCh38
rs752166559 Chr.4: 70335838-70335838 on Build GRCh38
rs142164310 Chr.4: 70336024-70336024 on Build GRCh38
rs771867150 Chr.4: 70336124-70336124 on Build GRCh38
rs1019139090 Chr.4: 70335932-70335932 on Build GRCh38
rs769848096 Chr.4: 70336113-70336113 on Build GRCh38
rs770080535 Chr.4: 70336209-70336209 on Build GRCh38
rs200977898 Chr.4: 70336255-70336255 on Build GRCh38
rs777328061 Chr.4: 70335842-70335842 on Build GRCh38
rs866735630 Chr.4: 70336304-70336304 on Build GRCh38
rs756032770 Chr.4: 70336049-70336049 on Build GRCh38
rs887485213 Chr.4: 70336285-70336285 on Build GRCh38
rs761840071 Chr.4: 70335874-70335874 on Build GRCh38
rs992625940 Chr.4: 70336143-70336143 on Build GRCh38
rs764471835 Chr.4: 70336025-70336025 on Build GRCh38
rs1048088734 Chr.4: 70336280-70336280 on Build GRCh38
rs770962721 Chr.4: 70336097-70336097 on Build GRCh38
rs750629728 Chr.4: 70335811-70335811 on Build GRCh38
rs764377711 Chr.4: 70336201-70336220 on Build GRCh38
rs573600956 Chr.4: 70336279-70336279 on Build GRCh38
rs770456606 Chr.4: 70336056-70336058 on Build GRCh38
rs745621727 Chr.4: 70335865-70335865 on Build GRCh38
rs745910463 Chr.4: 70336207-70336207 on Build GRCh38
rs748967987 Chr.4: 70336060-70336060 on Build GRCh38
rs775916305 Chr.4: 70336019-70336019 on Build GRCh38
rs139430564 Chr.4: 70335866-70335866 on Build GRCh38
rs746891726 Chr.4: 70336183-70336183 on Build GRCh38
rs1008833357 Chr.4: 70336224-70336224 on Build GRCh38
rs754178541 Chr.4: 70336235-70336236 on Build GRCh38
rs1042426905 Chr.4: 70336178-70336178 on Build GRCh38
rs369578007 Chr.4: 70336248-70336248 on Build GRCh38
rs764715464 Chr.4: 70336142-70336142 on Build GRCh38
rs754142979 Chr.4: 70335822-70335822 on Build GRCh38
rs34433477 Chr.4: 70335843-70335843 on Build GRCh38
rs561541971 Chr.4: 70335854-70335854 on Build GRCh38
rs768418266 Chr.4: 70335992-70335992 on Build GRCh38
rs754231118 Chr.4: 70335906-70335906 on Build GRCh38
rs760963716 Chr.4: 70335900-70335900 on Build GRCh38
rs765906296 Chr.4: 70336185-70336185 on Build GRCh38
rs765627574 Chr.4: 70336016-70336016 on Build GRCh38
rs186211014 Chr.4: 70335965-70335965 on Build GRCh38
rs149036625 Chr.4: 70335819-70335819 on Build GRCh38
rs757383116 Chr.4: 70336201-70336201 on Build GRCh38
rs959503798 Chr.4: 70336106-70336106 on Build GRCh38
rs753687991 Chr.4: 70336187-70336187 on Build GRCh38
rs779924073 Chr.4: 70335982-70335982 on Build GRCh38
rs374072903 Chr.4: 70335814-70335814 on Build GRCh38
rs149882290 Chr.4: 70336092-70336092 on Build GRCh38
rs745555076 Chr.4: 70335967-70335967 on Build GRCh38
rs1001089388 Chr.4: 70335870-70335870 on Build GRCh38
rs759224161 Chr.4: 70336199-70336201 on Build GRCh38
rs748801890 Chr.4: 70335883-70335883 on Build GRCh38
rs762493865 Chr.4: 70335871-70335872 on Build GRCh38
rs890031318 Chr.4: 70336218-70336218 on Build GRCh38
rs567242455 Chr.4: 70336090-70336090 on Build GRCh38
rs754706316 Chr.4: 70336190-70336190 on Build GRCh38
rs1003859529 Chr.4: 70336286-70336286 on Build GRCh38
rs754855562 Chr.4: 70336007-70336007 on Build GRCh38
rs749237446 Chr.4: 70335989-70335989 on Build GRCh38
rs2291182 Chr.4: 70335931-70335931 on Build GRCh38
rs778613021 Chr.4: 70335939-70335939 on Build GRCh38
rs755942851 Chr.4: 70335974-70335974 on Build GRCh38
rs760378146 Chr.4: 70336184-70336184 on Build GRCh38
rs753213479 Chr.4: 70335935-70335935 on Build GRCh38
rs182023781 Chr.4: 70335880-70335880 on Build GRCh38
rs767319197 Chr.4: 70336033-70336033 on Build GRCh38
rs368438643 Chr.4: 70336112-70336112 on Build GRCh38
rs895559363 Chr.4: 70336330-70336330 on Build GRCh38
rs776902349 Chr.4: 70336101-70336101 on Build GRCh38
rs144583373 Chr.4: 70335828-70335828 on Build GRCh38
rs765285487 Chr.4: 70336339-70336339 on Build GRCh38
rs182928869 Chr.4: 70336338-70336338 on Build GRCh38
rs985075662 Chr.4: 70336026-70336026 on Build GRCh38
rs780121111 Chr.4: 70336211-70336211 on Build GRCh38
rs369916862 Chr.4: 70335956-70335956 on Build GRCh38
rs1005879607 Chr.4: 70336334-70336334 on Build GRCh38
rs138774071 Chr.4: 70336244-70336244 on Build GRCh38
rs779352271 Chr.4: 70335837-70335837 on Build GRCh38
rs186468035 Chr.4: 70336132-70336132 on Build GRCh38
rs781260280 Chr.4: 70336203-70336203 on Build GRCh38
rs773366011 Chr.4: 70335881-70335881 on Build GRCh38
rs372435553 Chr.4: 70336082-70336082 on Build GRCh38
rs140154037 Chr.4: 70336068-70336068 on Build GRCh38
rs958033545 Chr.4: 70336214-70336214 on Build GRCh38
rs201758701 Chr.4: 70335892-70335892 on Build GRCh38
rs200159017 Chr.4: 70336027-70336027 on Build GRCh38
rs372887159 Chr.4: 70336179-70336179 on Build GRCh38

View Full Product Details