Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186588385-186588949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAT1
Entrez Gene ID
2195
Gene Name
FAT atypical cadherin 1
Gene Aliases
CDHF7, CDHR8, FAT, ME5, hFat1
UniGene
Hs.481371
Gene Chromosome Location
Chr.4: 186587783-186726915 on Build GRCh38
Transcripts
NM_005245.3 XM_005262834.2 XM_005262835.2 XM_006714139.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3940838 c.13587G>A p.A4529A Chr.4: 186588772-186588772 on Build GRCh38
6929458 c.13422G>T p.L4474F Chr.4: 186588937-186588937 on Build GRCh38
1294137 c.13717G>A p.E4573K Chr.4: 186588642-186588642 on Build GRCh38
6211301 c.13599G>C p.E4533D Chr.4: 186588760-186588760 on Build GRCh38
4683468 c.13725G>C p.V4575V Chr.4: 186588634-186588634 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564119304 Chr.4: 186588532-186588532 on Build GRCh38
rs139623858 Chr.4: 186588895-186588895 on Build GRCh38
rs117860756 Chr.4: 186588589-186588589 on Build GRCh38
rs572708850 Chr.4: 186588479-186588479 on Build GRCh38
rs777463346 Chr.4: 186588642-186588642 on Build GRCh38
rs373825887 Chr.4: 186588778-186588778 on Build GRCh38
rs761059913 Chr.4: 186588809-186588809 on Build GRCh38
rs774458628 Chr.4: 186588651-186588651 on Build GRCh38
rs952620390 Chr.4: 186588656-186588656 on Build GRCh38
rs908533298 Chr.4: 186588539-186588539 on Build GRCh38
rs772588307 Chr.4: 186588807-186588807 on Build GRCh38
rs140009081 Chr.4: 186588606-186588606 on Build GRCh38
rs778706452 Chr.4: 186588704-186588704 on Build GRCh38
rs1001738246 Chr.4: 186588617-186588617 on Build GRCh38
rs768453815 Chr.4: 186588782-186588782 on Build GRCh38
rs375820465 Chr.4: 186588877-186588877 on Build GRCh38
rs1043353926 Chr.4: 186588605-186588605 on Build GRCh38
rs865888636 Chr.4: 186588531-186588531 on Build GRCh38
rs757850899 Chr.4: 186588842-186588842 on Build GRCh38
rs147542447 Chr.4: 186588576-186588577 on Build GRCh38
rs116113983 Chr.4: 186588565-186588565 on Build GRCh38
rs543578964 Chr.4: 186588578-186588578 on Build GRCh38
rs961005755 Chr.4: 186588510-186588510 on Build GRCh38
rs775695644 Chr.4: 186588708-186588708 on Build GRCh38
rs146328457 Chr.4: 186588684-186588684 on Build GRCh38
rs761495730 Chr.4: 186588772-186588772 on Build GRCh38
rs558037855 Chr.4: 186588762-186588762 on Build GRCh38
rs755283793 Chr.4: 186588679-186588679 on Build GRCh38
rs201477548 Chr.4: 186588854-186588854 on Build GRCh38
rs367978383 Chr.4: 186588742-186588742 on Build GRCh38
rs35485176 Chr.4: 186588918-186588918 on Build GRCh38
rs764820968 Chr.4: 186588827-186588827 on Build GRCh38
rs764624079 Chr.4: 186588810-186588810 on Build GRCh38
rs770217697 Chr.4: 186588668-186588668 on Build GRCh38
rs993828603 Chr.4: 186588800-186588800 on Build GRCh38
rs549174375 Chr.4: 186588625-186588625 on Build GRCh38
rs745595921 Chr.4: 186588652-186588652 on Build GRCh38
rs553759185 Chr.4: 186588414-186588414 on Build GRCh38
rs758760787 Chr.4: 186588551-186588551 on Build GRCh38
rs911058608 Chr.4: 186588683-186588683 on Build GRCh38
rs773428259 Chr.4: 186588739-186588739 on Build GRCh38
rs752119953 Chr.4: 186588675-186588675 on Build GRCh38
rs749214202 Chr.4: 186588643-186588643 on Build GRCh38
rs749579091 Chr.4: 186588793-186588793 on Build GRCh38
rs540624845 Chr.4: 186588488-186588488 on Build GRCh38
rs189343266 Chr.4: 186588884-186588884 on Build GRCh38
rs374690008 Chr.4: 186588690-186588690 on Build GRCh38
rs779984754 Chr.4: 186588843-186588843 on Build GRCh38
rs920014158 Chr.4: 186588757-186588757 on Build GRCh38
rs759393542 Chr.4: 186588591-186588591 on Build GRCh38
rs769323827 Chr.4: 186588577-186588578 on Build GRCh38
rs1056427147 Chr.4: 186588740-186588740 on Build GRCh38
rs1007899937 Chr.4: 186588436-186588436 on Build GRCh38
rs904722749 Chr.4: 186588459-186588459 on Build GRCh38
rs772054594 Chr.4: 186588875-186588875 on Build GRCh38
rs758669356 Chr.4: 186588511-186588511 on Build GRCh38
rs150973681 Chr.4: 186588871-186588871 on Build GRCh38
rs376718741 Chr.4: 186588721-186588721 on Build GRCh38
rs376173860 Chr.4: 186588905-186588905 on Build GRCh38
rs940022862 Chr.4: 186588712-186588712 on Build GRCh38
rs750900612 Chr.4: 186588609-186588609 on Build GRCh38
rs199927785 Chr.4: 186588883-186588883 on Build GRCh38
rs761609848 Chr.4: 186588824-186588824 on Build GRCh38
rs149629492 Chr.4: 186588449-186588449 on Build GRCh38
rs928578735 Chr.4: 186588476-186588476 on Build GRCh38
rs749489170 Chr.4: 186588863-186588863 on Build GRCh38
rs371451914 Chr.4: 186588714-186588714 on Build GRCh38
rs202146434 Chr.4: 186588911-186588911 on Build GRCh38
rs770883326 Chr.4: 186588794-186588794 on Build GRCh38
rs766893768 Chr.4: 186588614-186588614 on Build GRCh38
rs771965001 Chr.4: 186588654-186588654 on Build GRCh38
rs747545404 Chr.4: 186588577-186588577 on Build GRCh38
rs200386216 Chr.4: 186588796-186588796 on Build GRCh38
rs75546828 Chr.4: 186588460-186588460 on Build GRCh38
rs373515547 Chr.4: 186588601-186588601 on Build GRCh38
rs746739819 Chr.4: 186588781-186588781 on Build GRCh38
rs1552691 Chr.4: 186588655-186588655 on Build GRCh38
rs372186380 Chr.4: 186588768-186588768 on Build GRCh38
rs757448356 Chr.4: 186588864-186588864 on Build GRCh38
rs753512486 Chr.4: 186588814-186588814 on Build GRCh38
rs755374400 Chr.4: 186588626-186588626 on Build GRCh38
rs376002194 Chr.4: 186588637-186588637 on Build GRCh38
rs770939534 Chr.4: 186588650-186588650 on Build GRCh38
rs751555230 Chr.4: 186588846-186588846 on Build GRCh38
rs1008890673 Chr.4: 186588583-186588583 on Build GRCh38
rs779949968 Chr.4: 186588715-186588715 on Build GRCh38
rs1049829513 Chr.4: 186588413-186588413 on Build GRCh38
rs375794457 Chr.4: 186588472-186588472 on Build GRCh38
rs944074220 Chr.4: 186588425-186588425 on Build GRCh38
rs187784855 Chr.4: 186588464-186588464 on Build GRCh38
rs770816787 Chr.4: 186588582-186588582 on Build GRCh38
rs766805655 Chr.4: 186588671-186588671 on Build GRCh38
rs931118497 Chr.4: 186588409-186588409 on Build GRCh38
rs369015571 Chr.4: 186588823-186588823 on Build GRCh38
rs555751691 Chr.4: 186588471-186588471 on Build GRCh38
rs370613233 Chr.4: 186588657-186588657 on Build GRCh38
rs780930037 Chr.4: 186588575-186588575 on Build GRCh38
rs142057401 Chr.4: 186588632-186588632 on Build GRCh38
rs539530918 Chr.4: 186588688-186588688 on Build GRCh38
rs376922855 Chr.4: 186588602-186588602 on Build GRCh38
rs769964561 Chr.4: 186588729-186588729 on Build GRCh38
rs754762942 Chr.4: 186588848-186588848 on Build GRCh38
rs777588238 Chr.4: 186588694-186588694 on Build GRCh38
rs183090437 Chr.4: 186588430-186588430 on Build GRCh38
rs972636112 Chr.4: 186588530-186588530 on Build GRCh38
rs367614609 Chr.4: 186588692-186588692 on Build GRCh38
rs774714048 Chr.4: 186588756-186588756 on Build GRCh38
rs944142258 Chr.4: 186588573-186588573 on Build GRCh38
rs200939422 Chr.4: 186588752-186588752 on Build GRCh38
rs765280609 Chr.4: 186588660-186588660 on Build GRCh38
rs756828769 Chr.4: 186588702-186588702 on Build GRCh38
rs559343059 Chr.4: 186588631-186588631 on Build GRCh38
rs148468016 Chr.4: 186588707-186588707 on Build GRCh38
rs183728932 Chr.4: 186588870-186588870 on Build GRCh38
rs772770486 Chr.4: 186588840-186588840 on Build GRCh38
rs1024646641 Chr.4: 186588505-186588505 on Build GRCh38
rs192609167 Chr.4: 186588777-186588777 on Build GRCh38
rs375893948 Chr.4: 186588799-186588799 on Build GRCh38
rs891756467 Chr.4: 186588445-186588445 on Build GRCh38
rs199584222 Chr.4: 186588562-186588562 on Build GRCh38
rs764878819 Chr.4: 186588910-186588910 on Build GRCh38
rs73873643 Chr.4: 186588450-186588450 on Build GRCh38
rs142612329 Chr.4: 186588769-186588769 on Build GRCh38
rs374285602 Chr.4: 186588661-186588661 on Build GRCh38
rs762511876 Chr.4: 186588917-186588917 on Build GRCh38
rs748762429 Chr.4: 186588699-186588699 on Build GRCh38
rs765370853 Chr.4: 186588607-186588607 on Build GRCh38
rs555931755 Chr.4: 186588773-186588773 on Build GRCh38
rs371459868 Chr.4: 186588687-186588687 on Build GRCh38
rs753054161 Chr.4: 186588763-186588763 on Build GRCh38
rs780438297 Chr.4: 186588559-186588559 on Build GRCh38
rs778036756 Chr.4: 186588533-186588533 on Build GRCh38
rs758955132 Chr.4: 186588612-186588612 on Build GRCh38

View Full Product Details