Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 145762926-145763506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf51
Entrez Gene ID
646603
Gene Name
chromosome 4 open reading frame 51
Gene Aliases
-
UniGene
Hs.452865
Gene Chromosome Location
Chr.4: 145680103-145792248 on Build GRCh38
Transcripts
XM_017008547.1 XM_017008548.1 XM_017008549.1 XM_017008550.1 XM_017008551.1 XM_017008552.1 XM_017008553.1 XM_017008554.1 XM_017008555.1 XM_017008556.1 NM_001306215.1 NM_178835.4 XM_011531634.2 XM_011531635.2 XM_011531645.2 XM_017007768.1 XM_017007769.1 XM_017007770.1 XM_017007771.1 XM_017007772.1 XM_017007773.1 XM_017007774.1 XM_017007775.1 XM_017007777.1

Gene Symbol
ZNF827
Entrez Gene ID
152485
Gene Name
zinc finger protein 827
Gene Aliases
-
UniGene
Hs.133916
Gene Chromosome Location
Chr.4: 145757627-145939008 on Build GRCh38
Transcripts
XM_017008547.1 XM_017008548.1 XM_017008549.1 XM_017008550.1 XM_017008551.1 XM_017008552.1 XM_017008553.1 XM_017008554.1 XM_017008555.1 XM_017008556.1 NM_001306215.1 NM_178835.4 XM_011531634.2 XM_011531635.2 XM_011531645.2 XM_017007768.1 XM_017007769.1 XM_017007770.1 XM_017007771.1 XM_017007772.1 XM_017007773.1 XM_017007774.1 XM_017007775.1 XM_017007777.1

SNP Detail

SNP ID Chromosome Location
rs577181414 Chr.4: 145762961-145762961 on Build GRCh38
rs770679587 Chr.4: 145762985-145762985 on Build GRCh38
rs1020063573 Chr.4: 145762982-145762982 on Build GRCh38
rs183231217 Chr.4: 145762973-145762973 on Build GRCh38
rs776755965 Chr.4: 145763396-145763396 on Build GRCh38
rs963674722 Chr.4: 145763220-145763220 on Build GRCh38
rs764232871 Chr.4: 145763100-145763100 on Build GRCh38
rs75874484 Chr.4: 145763226-145763226 on Build GRCh38
rs759544176 Chr.4: 145763484-145763484 on Build GRCh38
rs74433185 Chr.4: 145762960-145762960 on Build GRCh38
rs1005744372 Chr.4: 145763074-145763074 on Build GRCh38
rs776563149 Chr.4: 145763309-145763309 on Build GRCh38
rs939333745 Chr.4: 145763466-145763466 on Build GRCh38
rs941437981 Chr.4: 145763356-145763356 on Build GRCh38
rs922477103 Chr.4: 145763406-145763406 on Build GRCh38
rs897318510 Chr.4: 145763222-145763222 on Build GRCh38
rs900887886 Chr.4: 145762969-145762969 on Build GRCh38
rs926632296 Chr.4: 145763392-145763392 on Build GRCh38
rs925825162 Chr.4: 145763361-145763361 on Build GRCh38
rs115468844 Chr.4: 145763310-145763310 on Build GRCh38
rs775969368 Chr.4: 145763018-145763018 on Build GRCh38
rs968855986 Chr.4: 145763027-145763027 on Build GRCh38
rs1017945168 Chr.4: 145763146-145763146 on Build GRCh38
rs1049359838 Chr.4: 145762984-145762984 on Build GRCh38
rs933771845 Chr.4: 145762972-145762972 on Build GRCh38
rs961045912 Chr.4: 145763291-145763292 on Build GRCh38
rs990927330 Chr.4: 145763296-145763296 on Build GRCh38
rs934606215 Chr.4: 145763373-145763377 on Build GRCh38
rs770599323 Chr.4: 145763317-145763317 on Build GRCh38
rs746851497 Chr.4: 145763312-145763312 on Build GRCh38
rs978673320 Chr.4: 145763319-145763319 on Build GRCh38
rs1045103897 Chr.4: 145763377-145763377 on Build GRCh38
rs187918472 Chr.4: 145763247-145763247 on Build GRCh38
rs999904577 Chr.4: 145763126-145763126 on Build GRCh38

View Full Product Details