Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 13714323-13714846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH5
Entrez Gene ID
1767
Gene Name
dynein axonemal heavy chain 5
Gene Aliases
CILD3, DNAHC5, HL1, KTGNR, PCD
UniGene
Hs.212360
Gene Chromosome Location
Chr.5: 13690328-14011829 on Build GRCh38
Transcripts
NM_001369.2 XM_005248262.3 XM_017009177.1 XM_017009178.1 XM_017009179.1 XM_017009185.1 XM_017009186.1 XM_017009188.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
589536 c.13061C>T p.A4354V Chr.5: 13714469-13714469 on Build GRCh38
1671510 c.12944G>A p.G4315E Chr.5: 13714586-13714586 on Build GRCh38
4525034 c.13044G>A p.G4348G Chr.5: 13714486-13714486 on Build GRCh38
3608790 c.13084G>A p.D4362N Chr.5: 13714446-13714446 on Build GRCh38
4525047 c.13056G>A p.R4352R Chr.5: 13714474-13714474 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377316131 Chr.5: 13714572-13714572 on Build GRCh38
rs201172384 Chr.5: 13714469-13714469 on Build GRCh38
rs1018426683 Chr.5: 13714757-13714757 on Build GRCh38
rs761705556 Chr.5: 13714499-13714499 on Build GRCh38
rs772248285 Chr.5: 13714394-13714394 on Build GRCh38
rs371027404 Chr.5: 13714493-13714493 on Build GRCh38
rs769824654 Chr.5: 13714501-13714501 on Build GRCh38
rs572638119 Chr.5: 13714806-13714806 on Build GRCh38
rs376585054 Chr.5: 13714475-13714475 on Build GRCh38
rs768902025 Chr.5: 13714567-13714567 on Build GRCh38
rs748008373 Chr.5: 13714574-13714574 on Build GRCh38
rs372853742 Chr.5: 13714735-13714735 on Build GRCh38
rs897444650 Chr.5: 13714699-13714699 on Build GRCh38
rs1010304250 Chr.5: 13714789-13714789 on Build GRCh38
rs530188302 Chr.5: 13714476-13714476 on Build GRCh38
rs755271975 Chr.5: 13714525-13714525 on Build GRCh38
rs1003262420 Chr.5: 13714819-13714819 on Build GRCh38
rs993175666 Chr.5: 13714712-13714712 on Build GRCh38
rs373401142 Chr.5: 13714669-13714669 on Build GRCh38
rs749628828 Chr.5: 13714383-13714383 on Build GRCh38
rs762978621 Chr.5: 13714606-13714606 on Build GRCh38
rs560535018 Chr.5: 13714515-13714515 on Build GRCh38
rs754976780 Chr.5: 13714668-13714668 on Build GRCh38
rs748238895 Chr.5: 13714729-13714729 on Build GRCh38
rs145321489 Chr.5: 13714740-13714740 on Build GRCh38
rs750996382 Chr.5: 13714655-13714655 on Build GRCh38
rs543570342 Chr.5: 13714368-13714368 on Build GRCh38
rs538961013 Chr.5: 13714636-13714636 on Build GRCh38
rs759148086 Chr.5: 13714626-13714626 on Build GRCh38
rs944340640 Chr.5: 13714660-13714660 on Build GRCh38
rs574622154 Chr.5: 13714355-13714355 on Build GRCh38
rs747191200 Chr.5: 13714489-13714489 on Build GRCh38
rs930377551 Chr.5: 13714629-13714629 on Build GRCh38
rs372604448 Chr.5: 13714435-13714435 on Build GRCh38
rs920318129 Chr.5: 13714344-13714344 on Build GRCh38
rs776441005 Chr.5: 13714432-13714432 on Build GRCh38
rs30175 Chr.5: 13714631-13714631 on Build GRCh38
rs886142477 Chr.5: 13714418-13714418 on Build GRCh38
rs554261666 Chr.5: 13714792-13714792 on Build GRCh38
rs536034284 Chr.5: 13714763-13714763 on Build GRCh38
rs762564245 Chr.5: 13714507-13714507 on Build GRCh38
rs752241896 Chr.5: 13714634-13714634 on Build GRCh38
rs2277047 Chr.5: 13714395-13714395 on Build GRCh38
rs200924641 Chr.5: 13714468-13714468 on Build GRCh38
rs907520818 Chr.5: 13714657-13714657 on Build GRCh38
rs548721070 Chr.5: 13714512-13714512 on Build GRCh38
rs768406077 Chr.5: 13714492-13714492 on Build GRCh38
rs780103184 Chr.5: 13714708-13714708 on Build GRCh38
rs372488592 Chr.5: 13714465-13714465 on Build GRCh38
rs773285536 Chr.5: 13714594-13714594 on Build GRCh38
rs893346205 Chr.5: 13714767-13714767 on Build GRCh38
rs10077457 Chr.5: 13714459-13714459 on Build GRCh38
rs141961399 Chr.5: 13714462-13714462 on Build GRCh38
rs750130912 Chr.5: 13714537-13714537 on Build GRCh38
rs780181944 Chr.5: 13714555-13714555 on Build GRCh38
rs765919049 Chr.5: 13714508-13714508 on Build GRCh38
rs764898250 Chr.5: 13714440-13714440 on Build GRCh38
rs758893837 Chr.5: 13714656-13714656 on Build GRCh38
rs763894089 Chr.5: 13714431-13714431 on Build GRCh38
rs144949095 Chr.5: 13714457-13714457 on Build GRCh38
rs753960777 Chr.5: 13714458-13714458 on Build GRCh38
rs539810401 Chr.5: 13714417-13714417 on Build GRCh38
rs959268132 Chr.5: 13714821-13714821 on Build GRCh38
rs757416978 Chr.5: 13714509-13714509 on Build GRCh38
rs971521301 Chr.5: 13714794-13714794 on Build GRCh38
rs768751866 Chr.5: 13714768-13714768 on Build GRCh38
rs148805204 Chr.5: 13714453-13714453 on Build GRCh38
rs900721473 Chr.5: 13714709-13714709 on Build GRCh38
rs746134622 Chr.5: 13714384-13714384 on Build GRCh38
rs369372511 Chr.5: 13714504-13714504 on Build GRCh38
rs780046723 Chr.5: 13714658-13714658 on Build GRCh38
rs183952273 Chr.5: 13714356-13714356 on Build GRCh38
rs754196517 Chr.5: 13714516-13714516 on Build GRCh38
rs200697821 Chr.5: 13714535-13714535 on Build GRCh38
rs747257258 Chr.5: 13714559-13714559 on Build GRCh38
rs779253735 Chr.5: 13714536-13714536 on Build GRCh38
rs368776967 Chr.5: 13714460-13714460 on Build GRCh38
rs751631866 Chr.5: 13714659-13714659 on Build GRCh38
rs1008831613 Chr.5: 13714755-13714755 on Build GRCh38
rs372917600 Chr.5: 13714553-13714553 on Build GRCh38
rs979473530 Chr.5: 13714787-13714787 on Build GRCh38
rs757757096 Chr.5: 13714466-13714466 on Build GRCh38
rs773889655 Chr.5: 13714613-13714613 on Build GRCh38
rs765520528 Chr.5: 13714638-13714638 on Build GRCh38
rs769666008 Chr.5: 13714593-13714593 on Build GRCh38
rs955768429 Chr.5: 13714407-13714407 on Build GRCh38
rs115075057 Chr.5: 13714607-13714607 on Build GRCh38

View Full Product Details