Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 105366301-105366900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALCAM
Entrez Gene ID
214
Gene Name
activated leukocyte cell adhesion molecule
Gene Aliases
CD166, MEMD
UniGene
Hs.591293
Gene Chromosome Location
Chr.3: 105366713-105576913 on Build GRCh38
Transcripts
NM_001243280.1 NM_001243281.1 NM_001243283.1 NM_001627.3

SNP Detail

SNP ID Chromosome Location
rs747521430 Chr.3: 105366607-105366607 on Build GRCh38
rs1001060973 Chr.3: 105366829-105366829 on Build GRCh38
rs766169810 Chr.3: 105366866-105366866 on Build GRCh38
rs944862225 Chr.3: 105366744-105366744 on Build GRCh38
rs1006564890 Chr.3: 105366613-105366613 on Build GRCh38
rs1005007669 Chr.3: 105366811-105366811 on Build GRCh38
rs999022039 Chr.3: 105366337-105366337 on Build GRCh38
rs980786441 Chr.3: 105366558-105366558 on Build GRCh38
rs925581738 Chr.3: 105366707-105366707 on Build GRCh38
rs146071978 Chr.3: 105366675-105366675 on Build GRCh38
rs948408797 Chr.3: 105366514-105366514 on Build GRCh38
rs978948861 Chr.3: 105366538-105366538 on Build GRCh38
rs544976095 Chr.3: 105366343-105366343 on Build GRCh38
rs562432238 Chr.3: 105366436-105366436 on Build GRCh38
rs900530126 Chr.3: 105366590-105366590 on Build GRCh38
rs918451235 Chr.3: 105366731-105366731 on Build GRCh38
rs1039418412 Chr.3: 105366823-105366823 on Build GRCh38
rs531509943 Chr.3: 105366850-105366850 on Build GRCh38
rs890726826 Chr.3: 105366377-105366377 on Build GRCh38
rs992702795 Chr.3: 105366495-105366495 on Build GRCh38
rs1042811883 Chr.3: 105366332-105366332 on Build GRCh38
rs890590093 Chr.3: 105366569-105366569 on Build GRCh38
rs1029766871 Chr.3: 105366424-105366424 on Build GRCh38
rs1006679501 Chr.3: 105366447-105366447 on Build GRCh38
rs573448877 Chr.3: 105366398-105366398 on Build GRCh38
rs969321092 Chr.3: 105366690-105366690 on Build GRCh38
rs929834264 Chr.3: 105366747-105366747 on Build GRCh38
rs142971151 Chr.3: 105366750-105366750 on Build GRCh38
rs925708710 Chr.3: 105366544-105366544 on Build GRCh38
rs982652493 Chr.3: 105366480-105366480 on Build GRCh38
rs551331956 Chr.3: 105366692-105366692 on Build GRCh38
rs978471179 Chr.3: 105366697-105366697 on Build GRCh38
rs994828763 Chr.3: 105366591-105366591 on Build GRCh38
rs540555497 Chr.3: 105366414-105366414 on Build GRCh38
rs527374458 Chr.3: 105366710-105366710 on Build GRCh38
rs1018044812 Chr.3: 105366474-105366474 on Build GRCh38
rs747657940 Chr.3: 105366427-105366427 on Build GRCh38
rs941206254 Chr.3: 105366373-105366373 on Build GRCh38
rs1004935527 Chr.3: 105366617-105366617 on Build GRCh38
rs370496046 Chr.3: 105366585-105366585 on Build GRCh38
rs957207981 Chr.3: 105366539-105366539 on Build GRCh38
rs1049065097 Chr.3: 105366783-105366783 on Build GRCh38
rs1015406213 Chr.3: 105366488-105366488 on Build GRCh38
rs929923649 Chr.3: 105366596-105366596 on Build GRCh38
rs952675352 Chr.3: 105366435-105366435 on Build GRCh38
rs544791282 Chr.3: 105366605-105366605 on Build GRCh38
rs903619832 Chr.3: 105366628-105366628 on Build GRCh38
rs1023199277 Chr.3: 105366858-105366858 on Build GRCh38
rs182830881 Chr.3: 105366718-105366718 on Build GRCh38
rs556397658 Chr.3: 105366781-105366781 on Build GRCh38
rs915621130 Chr.3: 105366506-105366506 on Build GRCh38
rs1031609942 Chr.3: 105366658-105366658 on Build GRCh38
rs900771290 Chr.3: 105366763-105366763 on Build GRCh38
rs900462077 Chr.3: 105366399-105366399 on Build GRCh38
rs956972943 Chr.3: 105366659-105366659 on Build GRCh38
rs776477480 Chr.3: 105366792-105366792 on Build GRCh38
rs904725688 Chr.3: 105366333-105366333 on Build GRCh38
rs1025231243 Chr.3: 105366689-105366689 on Build GRCh38
rs918496301 Chr.3: 105366593-105366593 on Build GRCh38
rs1022878232 Chr.3: 105366688-105366688 on Build GRCh38
rs1001030296 Chr.3: 105366653-105366653 on Build GRCh38
rs773353438 Chr.3: 105366755-105366755 on Build GRCh38
rs1039252788 Chr.3: 105366589-105366589 on Build GRCh38
rs140881080 Chr.3: 105366379-105366379 on Build GRCh38
rs79438935 Chr.3: 105366626-105366626 on Build GRCh38
rs897080730 Chr.3: 105366385-105366385 on Build GRCh38
rs1000487584 Chr.3: 105366876-105366876 on Build GRCh38
rs1051414580 Chr.3: 105366748-105366748 on Build GRCh38
rs549204790 Chr.3: 105366729-105366729 on Build GRCh38
rs1053325807 Chr.3: 105366340-105366340 on Build GRCh38
rs890878772 Chr.3: 105366444-105366444 on Build GRCh38
rs139037862 Chr.3: 105366708-105366708 on Build GRCh38
rs934382756 Chr.3: 105366551-105366551 on Build GRCh38
rs995363914 Chr.3: 105366401-105366401 on Build GRCh38
rs9866561 Chr.3: 105366554-105366554 on Build GRCh38
rs1014080762 Chr.3: 105366840-105366840 on Build GRCh38
rs959950921 Chr.3: 105366492-105366492 on Build GRCh38
rs777181950 Chr.3: 105366651-105366651 on Build GRCh38
rs537605466 Chr.3: 105366751-105366751 on Build GRCh38
rs1056556604 Chr.3: 105366824-105366824 on Build GRCh38
rs147066598 Chr.3: 105366825-105366825 on Build GRCh38

View Full Product Details