Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 79125618-79126199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BHMT
Entrez Gene ID
635
Gene Name
betaine--homocysteine S-methyltransferase
Gene Aliases
BHMT1, HEL-S-61p
UniGene
Hs.80756
Gene Chromosome Location
Chr.5: 79111781-79132290 on Build GRCh38
Transcripts
NM_001713.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3920378 c.632C>T p.S211F Chr.5: 79126052-79126052 on Build GRCh38
6456970 c.736A>T p.S246C Chr.5: 79126156-79126156 on Build GRCh38
5939336 c.700G>A p.G234S Chr.5: 79126120-79126120 on Build GRCh38
139582 c.662C>T p.P221L Chr.5: 79126082-79126082 on Build GRCh38
6611464 c.731T>C p.L244P Chr.5: 79126151-79126151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs57182056 Chr.5: 79125914-79125914 on Build GRCh38
rs771167817 Chr.5: 79126020-79126020 on Build GRCh38
rs545279400 Chr.5: 79125730-79125730 on Build GRCh38
rs750220015 Chr.5: 79126074-79126074 on Build GRCh38
rs921696500 Chr.5: 79125840-79125840 on Build GRCh38
rs201234034 Chr.5: 79125956-79125957 on Build GRCh38
rs1028575294 Chr.5: 79125810-79125810 on Build GRCh38
rs527617816 Chr.5: 79125753-79125753 on Build GRCh38
rs1047358934 Chr.5: 79125752-79125752 on Build GRCh38
rs954383270 Chr.5: 79125813-79125813 on Build GRCh38
rs945581573 Chr.5: 79125930-79125930 on Build GRCh38
rs933019075 Chr.5: 79125875-79125875 on Build GRCh38
rs769947503 Chr.5: 79126049-79126049 on Build GRCh38
rs559793930 Chr.5: 79125934-79125934 on Build GRCh38
rs373915293 Chr.5: 79125957-79125957 on Build GRCh38
rs143822893 Chr.5: 79126071-79126071 on Build GRCh38
rs368015625 Chr.5: 79126010-79126010 on Build GRCh38
rs565111 Chr.5: 79125786-79125786 on Build GRCh38
rs781021530 Chr.5: 79125989-79125989 on Build GRCh38
rs917225572 Chr.5: 79125977-79125977 on Build GRCh38
rs1055650736 Chr.5: 79125979-79125979 on Build GRCh38
rs1020142465 Chr.5: 79125827-79125827 on Build GRCh38
rs758916006 Chr.5: 79126135-79126135 on Build GRCh38
rs79447662 Chr.5: 79125772-79125772 on Build GRCh38
rs983263165 Chr.5: 79125773-79125773 on Build GRCh38
rs1025315346 Chr.5: 79125769-79125772 on Build GRCh38
rs766168363 Chr.5: 79126094-79126094 on Build GRCh38
rs552467826 Chr.5: 79126011-79126011 on Build GRCh38
rs963163919 Chr.5: 79125804-79125804 on Build GRCh38
rs192178437 Chr.5: 79125803-79125803 on Build GRCh38
rs113616862 Chr.5: 79125683-79125683 on Build GRCh38
rs937476832 Chr.5: 79125717-79125717 on Build GRCh38
rs987114701 Chr.5: 79125906-79125906 on Build GRCh38
rs759839401 Chr.5: 79126048-79126048 on Build GRCh38
rs746050748 Chr.5: 79126178-79126178 on Build GRCh38
rs569174724 Chr.5: 79126107-79126107 on Build GRCh38
rs775746997 Chr.5: 79126055-79126055 on Build GRCh38
rs760454939 Chr.5: 79126087-79126087 on Build GRCh38
rs114632405 Chr.5: 79125814-79125814 on Build GRCh38
rs890094553 Chr.5: 79126163-79126163 on Build GRCh38
rs1013587057 Chr.5: 79125768-79125769 on Build GRCh38
rs752243322 Chr.5: 79126120-79126120 on Build GRCh38
rs749339615 Chr.5: 79126028-79126028 on Build GRCh38
rs694290 Chr.5: 79125957-79125957 on Build GRCh38
rs1042545297 Chr.5: 79125680-79125680 on Build GRCh38
rs950247669 Chr.5: 79125749-79125749 on Build GRCh38
rs754247726 Chr.5: 79125693-79125693 on Build GRCh38
rs140065455 Chr.5: 79126050-79126050 on Build GRCh38
rs1056198389 Chr.5: 79125722-79125722 on Build GRCh38
rs912813376 Chr.5: 79125918-79125918 on Build GRCh38
rs772486705 Chr.5: 79126002-79126002 on Build GRCh38
rs140612005 Chr.5: 79126168-79126168 on Build GRCh38
rs936807793 Chr.5: 79125959-79125959 on Build GRCh38
rs559252561 Chr.5: 79125735-79125735 on Build GRCh38
rs3733890 Chr.5: 79126136-79126136 on Build GRCh38
rs546724829 Chr.5: 79125905-79125905 on Build GRCh38
rs950050847 Chr.5: 79125981-79125981 on Build GRCh38
rs879430531 Chr.5: 79126013-79126013 on Build GRCh38
rs898902181 Chr.5: 79125798-79125798 on Build GRCh38
rs573217621 Chr.5: 79125729-79125729 on Build GRCh38
rs529744796 Chr.5: 79125899-79125899 on Build GRCh38
rs549597291 Chr.5: 79125842-79125842 on Build GRCh38
rs754646293 Chr.5: 79126112-79126112 on Build GRCh38
rs778354008 Chr.5: 79126118-79126118 on Build GRCh38
rs781699044 Chr.5: 79126176-79126176 on Build GRCh38
rs941654966 Chr.5: 79126044-79126044 on Build GRCh38
rs76697879 Chr.5: 79126067-79126067 on Build GRCh38
rs761782562 Chr.5: 79126058-79126058 on Build GRCh38
rs557019595 Chr.5: 79125950-79125951 on Build GRCh38
rs777056739 Chr.5: 79126024-79126024 on Build GRCh38
rs967351489 Chr.5: 79125865-79125865 on Build GRCh38
rs559484577 Chr.5: 79125726-79125726 on Build GRCh38

View Full Product Details