Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 134854753-134855322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf24
Entrez Gene ID
134553
Gene Name
chromosome 5 open reading frame 24
Gene Aliases
-
UniGene
Hs.708536
Gene Chromosome Location
Chr.5: 134833603-134859735 on Build GRCh38
Transcripts
NM_001135586.1 NM_001300894.1 NM_152409.3 XM_005271889.3 XM_017009049.1 XM_017009050.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1495909 c.4A>C p.M2L Chr.5: 134854904-134854904 on Build GRCh38
159625 c.162G>C p.Q54H Chr.5: 134855062-134855062 on Build GRCh38
159626 c.243G>C p.K81N Chr.5: 134855143-134855143 on Build GRCh38
1633941 c.129C>T p.S43S Chr.5: 134855029-134855029 on Build GRCh38
3138269 c.43C>T p.P15S Chr.5: 134854943-134854943 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760541667 Chr.5: 134854984-134854984 on Build GRCh38
rs781718375 Chr.5: 134854935-134854935 on Build GRCh38
rs937149388 Chr.5: 134855112-134855112 on Build GRCh38
rs182827108 Chr.5: 134854908-134854908 on Build GRCh38
rs775090596 Chr.5: 134855127-134855127 on Build GRCh38
rs752168056 Chr.5: 134854922-134854922 on Build GRCh38
rs776141095 Chr.5: 134855119-134855119 on Build GRCh38
rs757690818 Chr.5: 134854929-134854929 on Build GRCh38
rs547536550 Chr.5: 134854780-134854780 on Build GRCh38
rs754145538 Chr.5: 134855048-134855048 on Build GRCh38
rs144977700 Chr.5: 134854976-134854976 on Build GRCh38
rs752080201 Chr.5: 134855018-134855018 on Build GRCh38
rs962693392 Chr.5: 134855071-134855071 on Build GRCh38
rs772933950 Chr.5: 134854981-134854981 on Build GRCh38
rs768733990 Chr.5: 134854965-134854965 on Build GRCh38
rs774261454 Chr.5: 134854966-134854966 on Build GRCh38
rs565878997 Chr.5: 134854804-134854804 on Build GRCh38
rs760932124 Chr.5: 134855157-134855157 on Build GRCh38
rs777517568 Chr.5: 134855206-134855206 on Build GRCh38
rs750950317 Chr.5: 134855140-134855140 on Build GRCh38
rs373302777 Chr.5: 134855037-134855037 on Build GRCh38
rs192202876 Chr.5: 134854882-134854882 on Build GRCh38
rs1054830704 Chr.5: 134854799-134854799 on Build GRCh38
rs138898204 Chr.5: 134855107-134855107 on Build GRCh38
rs535222299 Chr.5: 134854856-134854856 on Build GRCh38
rs756502340 Chr.5: 134855040-134855040 on Build GRCh38
rs747027954 Chr.5: 134855208-134855208 on Build GRCh38
rs746080450 Chr.5: 134854940-134854940 on Build GRCh38
rs759060961 Chr.5: 134855004-134855004 on Build GRCh38
rs902397009 Chr.5: 134854866-134854866 on Build GRCh38
rs976675866 Chr.5: 134855148-134855148 on Build GRCh38
rs141164760 Chr.5: 134855302-134855302 on Build GRCh38
rs779130296 Chr.5: 134855181-134855181 on Build GRCh38
rs148188988 Chr.5: 134855276-134855276 on Build GRCh38
rs762578353 Chr.5: 134855128-134855128 on Build GRCh38
rs189210384 Chr.5: 134855042-134855042 on Build GRCh38
rs771952113 Chr.5: 134855098-134855098 on Build GRCh38
rs764630521 Chr.5: 134854913-134854913 on Build GRCh38
rs755244010 Chr.5: 134855174-134855174 on Build GRCh38
rs939085787 Chr.5: 134854779-134854779 on Build GRCh38
rs1006959344 Chr.5: 134855034-134855034 on Build GRCh38
rs376967674 Chr.5: 134855089-134855089 on Build GRCh38
rs767359794 Chr.5: 134854880-134854880 on Build GRCh38
rs957401340 Chr.5: 134854977-134854977 on Build GRCh38
rs775175831 Chr.5: 134855160-134855160 on Build GRCh38
rs146323904 Chr.5: 134855182-134855182 on Build GRCh38
rs770775481 Chr.5: 134855114-134855114 on Build GRCh38
rs754549646 Chr.5: 134854899-134854899 on Build GRCh38
rs866804048 Chr.5: 134854974-134854974 on Build GRCh38
rs201064253 Chr.5: 134854892-134854892 on Build GRCh38
rs764906069 Chr.5: 134855006-134855006 on Build GRCh38
rs780351562 Chr.5: 134854961-134854961 on Build GRCh38
rs1026067656 Chr.5: 134855156-134855156 on Build GRCh38
rs372418986 Chr.5: 134855027-134855027 on Build GRCh38
rs536221047 Chr.5: 134854806-134854806 on Build GRCh38
rs758481757 Chr.5: 134855194-134855194 on Build GRCh38
rs1046834798 Chr.5: 134855227-134855227 on Build GRCh38
rs767841849 Chr.5: 134855139-134855139 on Build GRCh38
rs765895478 Chr.5: 134854887-134854887 on Build GRCh38
rs761536554 Chr.5: 134854858-134854858 on Build GRCh38
rs776352593 Chr.5: 134854991-134854991 on Build GRCh38
rs559205176 Chr.5: 134854946-134854946 on Build GRCh38
rs765130702 Chr.5: 134855029-134855029 on Build GRCh38
rs368441862 Chr.5: 134855023-134855023 on Build GRCh38
rs142019018 Chr.5: 134855159-134855159 on Build GRCh38
rs769405914 Chr.5: 134855125-134855125 on Build GRCh38
rs765908439 Chr.5: 134854987-134854987 on Build GRCh38
rs746715843 Chr.5: 134855108-134855108 on Build GRCh38
rs532341776 Chr.5: 134854777-134854777 on Build GRCh38
rs761229698 Chr.5: 134855138-134855140 on Build GRCh38
rs76797142 Chr.5: 134855025-134855025 on Build GRCh38
rs1043617508 Chr.5: 134854848-134854848 on Build GRCh38
rs760289237 Chr.5: 134854883-134854883 on Build GRCh38
rs769526801 Chr.5: 134855284-134855284 on Build GRCh38
rs368128995 Chr.5: 134855228-134855228 on Build GRCh38
rs890969326 Chr.5: 134855000-134855000 on Build GRCh38
rs773814774 Chr.5: 134854867-134854868 on Build GRCh38
rs755049819 Chr.5: 134855051-134855051 on Build GRCh38
rs779008212 Chr.5: 134855074-134855074 on Build GRCh38
rs894796352 Chr.5: 134854823-134854823 on Build GRCh38
rs775285368 Chr.5: 134855301-134855301 on Build GRCh38
rs748153927 Chr.5: 134854969-134854969 on Build GRCh38
rs7700841 Chr.5: 134854895-134854895 on Build GRCh38
rs745632732 Chr.5: 134855101-134855101 on Build GRCh38
rs946425985 Chr.5: 134854834-134854834 on Build GRCh38
rs745676847 Chr.5: 134855278-134855278 on Build GRCh38
rs759301003 Chr.5: 134855123-134855123 on Build GRCh38

View Full Product Details