Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 142000930-142001491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNPDA1
Entrez Gene ID
10007
Gene Name
glucosamine-6-phosphate deaminase 1
Gene Aliases
GNP1, GNPDA, GNPI, GPI, HLN
UniGene
Hs.633853
Gene Chromosome Location
Chr.5: 142000669-142013055 on Build GRCh38
Transcripts
NM_005471.4 XM_005268348.1 XM_006714747.2 XM_011537537.1

SNP Detail

SNP ID Chromosome Location
rs1014361530 Chr.5: 142001379-142001379 on Build GRCh38
rs930613384 Chr.5: 142001427-142001427 on Build GRCh38
rs113836536 Chr.5: 142001275-142001275 on Build GRCh38
rs55892272 Chr.5: 142001273-142001274 on Build GRCh38
rs1048284370 Chr.5: 142001033-142001033 on Build GRCh38
rs252120 Chr.5: 142001163-142001163 on Build GRCh38
rs1804084 Chr.5: 142001024-142001024 on Build GRCh38
rs576103727 Chr.5: 142001398-142001398 on Build GRCh38
rs1014019409 Chr.5: 142001154-142001154 on Build GRCh38
rs777987754 Chr.5: 142001310-142001310 on Build GRCh38
rs566925111 Chr.5: 142001002-142001002 on Build GRCh38
rs543199593 Chr.5: 142001043-142001043 on Build GRCh38
rs944817421 Chr.5: 142000987-142000987 on Build GRCh38
rs1802147 Chr.5: 142001087-142001087 on Build GRCh38
rs142040050 Chr.5: 142001417-142001417 on Build GRCh38
rs559601788 Chr.5: 142001234-142001234 on Build GRCh38
rs182577638 Chr.5: 142001035-142001035 on Build GRCh38
rs1044381098 Chr.5: 142001092-142001092 on Build GRCh38
rs548642330 Chr.5: 142001292-142001292 on Build GRCh38
rs754889569 Chr.5: 142001423-142001423 on Build GRCh38
rs1009465999 Chr.5: 142001260-142001260 on Build GRCh38
rs752809327 Chr.5: 142001170-142001170 on Build GRCh38
rs137954723 Chr.5: 142001367-142001367 on Build GRCh38
rs867694860 Chr.5: 142001282-142001282 on Build GRCh38
rs150286691 Chr.5: 142001343-142001343 on Build GRCh38
rs1000363633 Chr.5: 142001413-142001413 on Build GRCh38
rs1041249058 Chr.5: 142000999-142000999 on Build GRCh38
rs947972548 Chr.5: 142001041-142001041 on Build GRCh38
rs750533924 Chr.5: 142000957-142000957 on Build GRCh38
rs926215088 Chr.5: 142000959-142000959 on Build GRCh38
rs374236549 Chr.5: 142001273-142001273 on Build GRCh38
rs187189071 Chr.5: 142001218-142001218 on Build GRCh38
rs986095828 Chr.5: 142001228-142001228 on Build GRCh38
rs528482907 Chr.5: 142001267-142001267 on Build GRCh38
rs1001152123 Chr.5: 142001090-142001090 on Build GRCh38
rs562351995 Chr.5: 142001293-142001293 on Build GRCh38
rs886548744 Chr.5: 142001244-142001245 on Build GRCh38
rs916456787 Chr.5: 142001040-142001040 on Build GRCh38
rs563172524 Chr.5: 142001119-142001119 on Build GRCh38
rs140395498 Chr.5: 142000958-142000958 on Build GRCh38
rs868327736 Chr.5: 142001141-142001141 on Build GRCh38
rs947918522 Chr.5: 142001408-142001408 on Build GRCh38
rs781024550 Chr.5: 142001467-142001467 on Build GRCh38
rs531258136 Chr.5: 142001306-142001306 on Build GRCh38
rs902969448 Chr.5: 142001401-142001401 on Build GRCh38
rs937269766 Chr.5: 142001354-142001354 on Build GRCh38
rs931956391 Chr.5: 142000968-142000968 on Build GRCh38
rs900727644 Chr.5: 142001290-142001290 on Build GRCh38
rs972154597 Chr.5: 142001304-142001304 on Build GRCh38
rs897003461 Chr.5: 142001015-142001015 on Build GRCh38
rs953649045 Chr.5: 142001221-142001221 on Build GRCh38
rs978954973 Chr.5: 142001180-142001180 on Build GRCh38
rs73793930 Chr.5: 142001363-142001363 on Build GRCh38
rs758410099 Chr.5: 142001147-142001147 on Build GRCh38
rs1050450451 Chr.5: 142000982-142000982 on Build GRCh38
rs1021879752 Chr.5: 142001301-142001302 on Build GRCh38
rs397702433 Chr.5: 142001274-142001275 on Build GRCh38
rs911925981 Chr.5: 142000985-142000985 on Build GRCh38
rs547181412 Chr.5: 142001082-142001082 on Build GRCh38
rs551316294 Chr.5: 142001340-142001340 on Build GRCh38
rs536990727 Chr.5: 142001308-142001308 on Build GRCh38
rs11557799 Chr.5: 142001094-142001094 on Build GRCh38

View Full Product Details