Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138253961-138254542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFRA3
Entrez Gene ID
2676
Gene Name
GDNF family receptor alpha 3
Gene Aliases
GDNFR3
UniGene
Hs.58042
Gene Chromosome Location
Chr.5: 138252380-138274564 on Build GRCh38
Transcripts
NM_001496.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4509921 c.821C>T p.P274L Chr.5: 138254125-138254125 on Build GRCh38
5672290 c.788C>T p.S263L Chr.5: 138254158-138254158 on Build GRCh38
5709465 c.845C>A p.A282E Chr.5: 138254101-138254101 on Build GRCh38
6469956 c.867A>T p.L289L Chr.5: 138254079-138254079 on Build GRCh38
4561442 c.885G>A p.L295L Chr.5: 138254061-138254061 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551119368 Chr.5: 138254482-138254482 on Build GRCh38
rs769209317 Chr.5: 138254124-138254124 on Build GRCh38
rs896759350 Chr.5: 138254014-138254014 on Build GRCh38
rs76093287 Chr.5: 138254044-138254044 on Build GRCh38
rs569429006 Chr.5: 138254188-138254189 on Build GRCh38
rs115929904 Chr.5: 138254001-138254001 on Build GRCh38
rs777829495 Chr.5: 138254183-138254183 on Build GRCh38
rs375045686 Chr.5: 138254067-138254067 on Build GRCh38
rs769417311 Chr.5: 138254192-138254192 on Build GRCh38
rs113436085 Chr.5: 138254204-138254204 on Build GRCh38
rs1028551733 Chr.5: 138254511-138254511 on Build GRCh38
rs559850984 Chr.5: 138254393-138254393 on Build GRCh38
rs752757685 Chr.5: 138254179-138254179 on Build GRCh38
rs537077148 Chr.5: 138254293-138254293 on Build GRCh38
rs986231246 Chr.5: 138254252-138254252 on Build GRCh38
rs902690324 Chr.5: 138254512-138254512 on Build GRCh38
rs774926607 Chr.5: 138254193-138254193 on Build GRCh38
rs77183860 Chr.5: 138254030-138254030 on Build GRCh38
rs986514001 Chr.5: 138254520-138254520 on Build GRCh38
rs201017144 Chr.5: 138254024-138254024 on Build GRCh38
rs760998410 Chr.5: 138254178-138254181 on Build GRCh38
rs534629288 Chr.5: 138254026-138254026 on Build GRCh38
rs188636781 Chr.5: 138254359-138254359 on Build GRCh38
rs758478294 Chr.5: 138254181-138254181 on Build GRCh38
rs73792434 Chr.5: 138254160-138254160 on Build GRCh38
rs1010694437 Chr.5: 138254442-138254442 on Build GRCh38
rs371365382 Chr.5: 138254100-138254100 on Build GRCh38
rs758922665 Chr.5: 138254283-138254283 on Build GRCh38
rs751052784 Chr.5: 138254187-138254187 on Build GRCh38
rs909268312 Chr.5: 138254334-138254334 on Build GRCh38
rs199567177 Chr.5: 138254128-138254128 on Build GRCh38
rs779013605 Chr.5: 138254045-138254045 on Build GRCh38
rs1021296687 Chr.5: 138254059-138254059 on Build GRCh38
rs762329638 Chr.5: 138254021-138254021 on Build GRCh38
rs750630458 Chr.5: 138254044-138254044 on Build GRCh38
rs761335465 Chr.5: 138254187-138254188 on Build GRCh38
rs896483756 Chr.5: 138254439-138254439 on Build GRCh38
rs754042082 Chr.5: 138254170-138254170 on Build GRCh38
rs568496270 Chr.5: 138254255-138254255 on Build GRCh38
rs754154265 Chr.5: 138254183-138254184 on Build GRCh38
rs532857869 Chr.5: 138254375-138254375 on Build GRCh38
rs766667458 Chr.5: 138254161-138254161 on Build GRCh38
rs746797873 Chr.5: 138254103-138254103 on Build GRCh38
rs949701483 Chr.5: 138254452-138254452 on Build GRCh38
rs199928771 Chr.5: 138254189-138254190 on Build GRCh38
rs569666126 Chr.5: 138253995-138253995 on Build GRCh38
rs773622175 Chr.5: 138254154-138254154 on Build GRCh38
rs6884097 Chr.5: 138254321-138254321 on Build GRCh38
rs374217131 Chr.5: 138254016-138254016 on Build GRCh38
rs755180171 Chr.5: 138254171-138254171 on Build GRCh38
rs797019885 Chr.5: 138254188-138254189 on Build GRCh38
rs766578577 Chr.5: 138254040-138254040 on Build GRCh38
rs919100444 Chr.5: 138254298-138254298 on Build GRCh38
rs577431796 Chr.5: 138254313-138254313 on Build GRCh38
rs1000687242 Chr.5: 138254509-138254509 on Build GRCh38
rs765246437 Chr.5: 138254178-138254178 on Build GRCh38
rs966398827 Chr.5: 138254274-138254274 on Build GRCh38
rs1023044296 Chr.5: 138254012-138254012 on Build GRCh38
rs779391011 Chr.5: 138254148-138254148 on Build GRCh38
rs530560276 Chr.5: 138253998-138253998 on Build GRCh38
rs116268521 Chr.5: 138254190-138254190 on Build GRCh38
rs756686070 Chr.5: 138254188-138254190 on Build GRCh38
rs78661573 Chr.5: 138254202-138254202 on Build GRCh38
rs1035632985 Chr.5: 138254028-138254028 on Build GRCh38
rs766788569 Chr.5: 138254183-138254183 on Build GRCh38
rs768727049 Chr.5: 138254399-138254399 on Build GRCh38
rs979709856 Chr.5: 138254070-138254070 on Build GRCh38
rs754002363 Chr.5: 138254042-138254042 on Build GRCh38
rs367714394 Chr.5: 138254077-138254077 on Build GRCh38
rs776114581 Chr.5: 138254123-138254123 on Build GRCh38
rs762307930 Chr.5: 138254147-138254147 on Build GRCh38
rs748058617 Chr.5: 138254073-138254073 on Build GRCh38
rs367624929 Chr.5: 138254188-138254188 on Build GRCh38
rs113913981 Chr.5: 138254312-138254312 on Build GRCh38
rs1042583378 Chr.5: 138254465-138254465 on Build GRCh38
rs770755573 Chr.5: 138254119-138254119 on Build GRCh38
rs940779283 Chr.5: 138254149-138254149 on Build GRCh38
rs746838783 Chr.5: 138254187-138254187 on Build GRCh38
rs548095377 Chr.5: 138254382-138254382 on Build GRCh38
rs745516331 Chr.5: 138254191-138254191 on Build GRCh38
rs779342373 Chr.5: 138254184-138254185 on Build GRCh38

View Full Product Details