Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 173910319-173910806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPEB4
Entrez Gene ID
80315
Gene Name
cytoplasmic polyadenylation element binding protein 4
Gene Aliases
CPE-BP4, hCPEB-4
UniGene
Hs.127126
Gene Chromosome Location
Chr.5: 173888328-173960991 on Build GRCh38
Transcripts
NM_001308189.1 NM_001308191.1 NM_001308192.1 NM_001308193.1 NM_030627.3 XM_005265994.1 XM_011534660.2 XM_011534661.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5908478 c.1129C>T p.R377C Chr.5: 173910526-173910526 on Build GRCh38
6546947 c.1170C>T p.L390L Chr.5: 173910567-173910567 on Build GRCh38
3373798 c.1161G>A p.E387E Chr.5: 173910558-173910558 on Build GRCh38
1436077 c.1144G>A p.D382N Chr.5: 173910541-173910541 on Build GRCh38
6207000 c.1130G>A p.R377H Chr.5: 173910527-173910527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs972364463 Chr.5: 173910430-173910430 on Build GRCh38
rs750214597 Chr.5: 173910620-173910620 on Build GRCh38
rs760270553 Chr.5: 173910567-173910567 on Build GRCh38
rs1039781923 Chr.5: 173910549-173910549 on Build GRCh38
rs776731763 Chr.5: 173910513-173910513 on Build GRCh38
rs757186137 Chr.5: 173910485-173910485 on Build GRCh38
rs912466415 Chr.5: 173910727-173910727 on Build GRCh38
rs747307733 Chr.5: 173910492-173910492 on Build GRCh38
rs569550736 Chr.5: 173910540-173910540 on Build GRCh38
rs892717050 Chr.5: 173910707-173910707 on Build GRCh38
rs201275485 Chr.5: 173910480-173910480 on Build GRCh38
rs150875114 Chr.5: 173910720-173910720 on Build GRCh38
rs1023194994 Chr.5: 173910494-173910494 on Build GRCh38
rs183397681 Chr.5: 173910373-173910373 on Build GRCh38
rs76997507 Chr.5: 173910621-173910621 on Build GRCh38
rs749840131 Chr.5: 173910641-173910641 on Build GRCh38
rs756982545 Chr.5: 173910598-173910598 on Build GRCh38
rs778235629 Chr.5: 173910640-173910640 on Build GRCh38
rs748158271 Chr.5: 173910627-173910627 on Build GRCh38
rs771887833 Chr.5: 173910429-173910429 on Build GRCh38
rs767101563 Chr.5: 173910553-173910553 on Build GRCh38
rs781406258 Chr.5: 173910622-173910622 on Build GRCh38
rs377449359 Chr.5: 173910489-173910489 on Build GRCh38
rs764123436 Chr.5: 173910591-173910591 on Build GRCh38
rs768589797 Chr.5: 173910510-173910510 on Build GRCh38
rs369037257 Chr.5: 173910527-173910527 on Build GRCh38
rs1051036272 Chr.5: 173910503-173910503 on Build GRCh38
rs773850447 Chr.5: 173910535-173910535 on Build GRCh38
rs1024634570 Chr.5: 173910739-173910739 on Build GRCh38
rs772992740 Chr.5: 173910729-173910729 on Build GRCh38
rs767100811 Chr.5: 173910479-173910479 on Build GRCh38
rs919182421 Chr.5: 173910437-173910437 on Build GRCh38
rs1013879806 Chr.5: 173910709-173910709 on Build GRCh38
rs749028069 Chr.5: 173910484-173910484 on Build GRCh38
rs969068970 Chr.5: 173910742-173910742 on Build GRCh38
rs777870361 Chr.5: 173910483-173910483 on Build GRCh38
rs763296139 Chr.5: 173910520-173910520 on Build GRCh38
rs753884175 Chr.5: 173910595-173910595 on Build GRCh38
rs188863474 Chr.5: 173910763-173910763 on Build GRCh38
rs980547410 Chr.5: 173910399-173910399 on Build GRCh38
rs754430235 Chr.5: 173910477-173910477 on Build GRCh38
rs878997588 Chr.5: 173910648-173910648 on Build GRCh38
rs551244961 Chr.5: 173910529-173910529 on Build GRCh38
rs148567441 Chr.5: 173910541-173910541 on Build GRCh38
rs1001048402 Chr.5: 173910603-173910603 on Build GRCh38
rs749645146 Chr.5: 173910424-173910424 on Build GRCh38
rs200768087 Chr.5: 173910555-173910555 on Build GRCh38
rs770228680 Chr.5: 173910515-173910515 on Build GRCh38
rs777092879 Chr.5: 173910407-173910407 on Build GRCh38
rs778809250 Chr.5: 173910614-173910614 on Build GRCh38
rs769924421 Chr.5: 173910637-173910637 on Build GRCh38
rs756069447 Chr.5: 173910482-173910482 on Build GRCh38
rs759323664 Chr.5: 173910657-173910670 on Build GRCh38
rs773712347 Chr.5: 173910519-173910519 on Build GRCh38
rs148319555 Chr.5: 173910418-173910418 on Build GRCh38
rs528875394 Chr.5: 173910679-173910679 on Build GRCh38
rs932476035 Chr.5: 173910690-173910692 on Build GRCh38

View Full Product Details