Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 158712989-158713485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EBF1
Entrez Gene ID
1879
Gene Name
early B-cell factor 1
Gene Aliases
COE1, EBF, O/E-1, OLF1
UniGene
Hs.573143
Gene Chromosome Location
Chr.5: 158695915-159099762 on Build GRCh38
Transcripts
NM_001290360.2 NM_001324101.1 NM_001324103.1 NM_001324106.1 NM_001324107.1 NM_001324108.1 NM_001324109.1 NM_001324111.1 NM_024007.4 NM_182708.2 XM_017009192.1 XM_017009193.1 XM_017009194.1 XM_017009195.1 XM_017009196.1 XM_017009197.1 XM_017009198.1 XM_017009199.1 XM_017009200.1 XM_017009201.1 XM_017009204.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5356716 c.1309A>C p.N437H Chr.5: 158713030-158713030 on Build GRCh38
7121767 c.1211C>A p.A404E Chr.5: 158713128-158713128 on Build GRCh38
3919508 c.1308G>A p.V436V Chr.5: 158713031-158713031 on Build GRCh38
1292064 c.1305C>T p.G435G Chr.5: 158713034-158713034 on Build GRCh38
244180 c.1266G>A p.P422P Chr.5: 158713073-158713073 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938204362 Chr.5: 158713202-158713202 on Build GRCh38
rs898996839 Chr.5: 158713311-158713311 on Build GRCh38
rs757216565 Chr.5: 158713102-158713102 on Build GRCh38
rs776689677 Chr.5: 158713056-158713056 on Build GRCh38
rs747932991 Chr.5: 158713028-158713028 on Build GRCh38
rs557997304 Chr.5: 158713215-158713215 on Build GRCh38
rs377223081 Chr.5: 158713048-158713048 on Build GRCh38
rs745844207 Chr.5: 158713112-158713112 on Build GRCh38
rs748315881 Chr.5: 158713152-158713152 on Build GRCh38
rs760863289 Chr.5: 158713051-158713051 on Build GRCh38
rs749844004 Chr.5: 158713226-158713226 on Build GRCh38
rs755850355 Chr.5: 158713092-158713092 on Build GRCh38
rs141300956 Chr.5: 158713124-158713124 on Build GRCh38
rs995988467 Chr.5: 158713320-158713320 on Build GRCh38
rs535605217 Chr.5: 158713053-158713053 on Build GRCh38
rs1042404596 Chr.5: 158713227-158713227 on Build GRCh38
rs559493437 Chr.5: 158713420-158713420 on Build GRCh38
rs187382921 Chr.5: 158713361-158713361 on Build GRCh38
rs570120838 Chr.5: 158713049-158713049 on Build GRCh38
rs368950395 Chr.5: 158713070-158713070 on Build GRCh38
rs757596884 Chr.5: 158713221-158713221 on Build GRCh38
rs116052031 Chr.5: 158713282-158713282 on Build GRCh38
rs779028258 Chr.5: 158713107-158713107 on Build GRCh38
rs1011119110 Chr.5: 158713225-158713225 on Build GRCh38
rs765507286 Chr.5: 158713067-158713067 on Build GRCh38
rs756829673 Chr.5: 158713180-158713180 on Build GRCh38
rs540694325 Chr.5: 158713412-158713412 on Build GRCh38
rs763383652 Chr.5: 158713072-158713072 on Build GRCh38
rs964523647 Chr.5: 158713319-158713319 on Build GRCh38
rs940097583 Chr.5: 158713106-158713106 on Build GRCh38
rs370485550 Chr.5: 158713178-158713178 on Build GRCh38
rs777669529 Chr.5: 158713034-158713034 on Build GRCh38
rs951909431 Chr.5: 158713354-158713354 on Build GRCh38
rs774052785 Chr.5: 158713074-158713074 on Build GRCh38
rs780739827 Chr.5: 158713009-158713009 on Build GRCh38
rs1013895582 Chr.5: 158713367-158713367 on Build GRCh38
rs750855534 Chr.5: 158713071-158713071 on Build GRCh38
rs868057434 Chr.5: 158713142-158713142 on Build GRCh38
rs982585964 Chr.5: 158713452-158713452 on Build GRCh38
rs1046508895 Chr.5: 158713115-158713115 on Build GRCh38
rs780793034 Chr.5: 158713079-158713079 on Build GRCh38
rs554321278 Chr.5: 158713169-158713203 on Build GRCh38
rs572150945 Chr.5: 158713206-158713206 on Build GRCh38
rs143017293 Chr.5: 158713073-158713073 on Build GRCh38
rs1022740891 Chr.5: 158713441-158713441 on Build GRCh38
rs773507828 Chr.5: 158713189-158713189 on Build GRCh38
rs1018726967 Chr.5: 158713302-158713302 on Build GRCh38
rs762145923 Chr.5: 158713065-158713065 on Build GRCh38
rs550639939 Chr.5: 158713171-158713171 on Build GRCh38
rs182297797 Chr.5: 158713332-158713332 on Build GRCh38
rs267600519 Chr.5: 158713131-158713131 on Build GRCh38
rs752266713 Chr.5: 158713091-158713091 on Build GRCh38
rs1009057542 Chr.5: 158713288-158713288 on Build GRCh38
rs3734105 Chr.5: 158713214-158713214 on Build GRCh38
rs148200095 Chr.5: 158713076-158713076 on Build GRCh38
rs190925775 Chr.5: 158713386-158713386 on Build GRCh38
rs190918385 Chr.5: 158713259-158713259 on Build GRCh38
rs988400793 Chr.5: 158713180-158713180 on Build GRCh38
rs768598275 Chr.5: 158713123-158713123 on Build GRCh38
rs149065953 Chr.5: 158713033-158713033 on Build GRCh38
rs138007031 Chr.5: 158713025-158713025 on Build GRCh38
rs749094595 Chr.5: 158713093-158713093 on Build GRCh38

View Full Product Details