Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30653131-30653729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf136
Entrez Gene ID
221545
Gene Name
chromosome 6 open reading frame 136
Gene Aliases
-
UniGene
Hs.591787
Gene Chromosome Location
Chr.6: 30647039-30653210 on Build GRCh38
Transcripts
NM_001109938.2 NM_001161376.1 NM_145029.3 NM_001164239.1 NM_003587.4 XM_011514938.2 XM_011514939.2 XM_011514940.2 XM_011514941.2

Gene Symbol
DHX16
Entrez Gene ID
8449
Gene Name
DEAH-box helicase 16
Gene Aliases
DBP2, DDX16, PRO2014, PRP8, PRPF2, Prp2
UniGene
Hs.485060
Gene Chromosome Location
Chr.6: 30653119-30673053 on Build GRCh38
Transcripts
NM_001109938.2 NM_001161376.1 NM_145029.3 NM_001164239.1 NM_003587.4 XM_011514938.2 XM_011514939.2 XM_011514940.2 XM_011514941.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6471048 c.1669G>A p.E557K Chr.6: 30653256-30653256 on Build GRCh38
7163539 c.1609G>A p.A537T Chr.6: 30653316-30653316 on Build GRCh38
4863974 c.1591G>A p.A531T Chr.6: 30653334-30653334 on Build GRCh38
6471051 c.1638G>A p.K546K Chr.6: 30653287-30653287 on Build GRCh38
4426139 c.3011_3012insA p.S1005fs*13 Chr.6: 30653356-30653357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774434206 Chr.6: 30653216-30653216 on Build GRCh38
rs1127955 Chr.6: 30653199-30653199 on Build GRCh38
rs907729487 Chr.6: 30653587-30653587 on Build GRCh38
rs937060046 Chr.6: 30653676-30653676 on Build GRCh38
rs143023855 Chr.6: 30653422-30653424 on Build GRCh38
rs765707126 Chr.6: 30653410-30653410 on Build GRCh38
rs781331484 Chr.6: 30653343-30653343 on Build GRCh38
rs1021032511 Chr.6: 30653168-30653168 on Build GRCh38
rs764628772 Chr.6: 30653320-30653320 on Build GRCh38
rs33932721 Chr.6: 30653617-30653618 on Build GRCh38
rs181681183 Chr.6: 30653501-30653501 on Build GRCh38
rs761762573 Chr.6: 30653243-30653244 on Build GRCh38
rs554492163 Chr.6: 30653707-30653713 on Build GRCh38
rs200009023 Chr.6: 30653270-30653270 on Build GRCh38
rs887757017 Chr.6: 30653344-30653344 on Build GRCh38
rs368094880 Chr.6: 30653197-30653197 on Build GRCh38
rs896641870 Chr.6: 30653433-30653434 on Build GRCh38
rs902628399 Chr.6: 30653456-30653457 on Build GRCh38
rs200945980 Chr.6: 30653641-30653641 on Build GRCh38
rs760210893 Chr.6: 30653257-30653257 on Build GRCh38
rs369800407 Chr.6: 30653635-30653636 on Build GRCh38
rs932156624 Chr.6: 30653663-30653663 on Build GRCh38
rs768566179 Chr.6: 30653204-30653204 on Build GRCh38
rs1037349647 Chr.6: 30653372-30653372 on Build GRCh38
rs776342899 Chr.6: 30653269-30653269 on Build GRCh38
rs897856540 Chr.6: 30653697-30653697 on Build GRCh38
rs1020981834 Chr.6: 30653434-30653434 on Build GRCh38
rs964745773 Chr.6: 30653494-30653494 on Build GRCh38
rs367754684 Chr.6: 30653402-30653402 on Build GRCh38
rs956929737 Chr.6: 30653584-30653584 on Build GRCh38
rs753228386 Chr.6: 30653314-30653314 on Build GRCh38
rs925182333 Chr.6: 30653221-30653221 on Build GRCh38
rs750606024 Chr.6: 30653356-30653356 on Build GRCh38
rs1030707517 Chr.6: 30653570-30653570 on Build GRCh38
rs941661155 Chr.6: 30653361-30653361 on Build GRCh38
rs549822282 Chr.6: 30653465-30653465 on Build GRCh38
rs942165795 Chr.6: 30653691-30653691 on Build GRCh38
rs1040557544 Chr.6: 30653222-30653222 on Build GRCh38
rs986199858 Chr.6: 30653688-30653688 on Build GRCh38
rs1023817814 Chr.6: 30653527-30653527 on Build GRCh38
rs929874338 Chr.6: 30653707-30653707 on Build GRCh38
rs941733314 Chr.6: 30653591-30653591 on Build GRCh38
rs757343846 Chr.6: 30653334-30653334 on Build GRCh38
rs374191584 Chr.6: 30653396-30653396 on Build GRCh38
rs983330473 Chr.6: 30653583-30653583 on Build GRCh38
rs141923733 Chr.6: 30653277-30653277 on Build GRCh38
rs909291874 Chr.6: 30653689-30653689 on Build GRCh38
rs979430700 Chr.6: 30653205-30653205 on Build GRCh38
rs763413391 Chr.6: 30653315-30653315 on Build GRCh38
rs773654815 Chr.6: 30653402-30653402 on Build GRCh38
rs371328692 Chr.6: 30653375-30653375 on Build GRCh38
rs1034800455 Chr.6: 30653457-30653457 on Build GRCh38
rs778801197 Chr.6: 30653383-30653383 on Build GRCh38
rs765530905 Chr.6: 30653305-30653305 on Build GRCh38
rs201164769 Chr.6: 30653642-30653642 on Build GRCh38
rs1030282790 Chr.6: 30653510-30653510 on Build GRCh38
rs967704914 Chr.6: 30653542-30653542 on Build GRCh38
rs140329547 Chr.6: 30653616-30653617 on Build GRCh38
rs28459331 Chr.6: 30653636-30653636 on Build GRCh38
rs750657160 Chr.6: 30653437-30653437 on Build GRCh38
rs771728483 Chr.6: 30653233-30653233 on Build GRCh38
rs145428484 Chr.6: 30653607-30653608 on Build GRCh38
rs533463729 Chr.6: 30653673-30653673 on Build GRCh38
rs1036311183 Chr.6: 30653695-30653695 on Build GRCh38
rs966671810 Chr.6: 30653198-30653198 on Build GRCh38
rs371237906 Chr.6: 30653262-30653262 on Build GRCh38
rs186854310 Chr.6: 30653548-30653548 on Build GRCh38
rs754833325 Chr.6: 30653370-30653370 on Build GRCh38
rs532886137 Chr.6: 30653397-30653397 on Build GRCh38
rs976355765 Chr.6: 30653610-30653610 on Build GRCh38
rs975063561 Chr.6: 30653500-30653500 on Build GRCh38
rs1011974839 Chr.6: 30653469-30653469 on Build GRCh38
rs994077455 Chr.6: 30653442-30653442 on Build GRCh38
rs926905960 Chr.6: 30653613-30653613 on Build GRCh38
rs372632356 Chr.6: 30653414-30653414 on Build GRCh38
rs868571875 Chr.6: 30653628-30653628 on Build GRCh38
rs751636618 Chr.6: 30653321-30653321 on Build GRCh38
rs748126393 Chr.6: 30653387-30653387 on Build GRCh38
rs748038802 Chr.6: 30653232-30653232 on Build GRCh38
rs189393359 Chr.6: 30653393-30653393 on Build GRCh38
rs772640089 Chr.6: 30653245-30653245 on Build GRCh38
rs547315021 Chr.6: 30653299-30653299 on Build GRCh38
rs1013784840 Chr.6: 30653434-30653434 on Build GRCh38
rs770548224 Chr.6: 30653258-30653258 on Build GRCh38

View Full Product Details