Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129260197-129260764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPG1
Entrez Gene ID
22820
Gene Name
coatomer protein complex subunit gamma 1
Gene Aliases
COPG
UniGene
Hs.518250
Gene Chromosome Location
Chr.3: 129249575-129277773 on Build GRCh38
Transcripts
NM_016128.3 XM_011512549.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6634053 c.926G>A p.R309H Chr.3: 129260387-129260387 on Build GRCh38
5004923 c.937A>G p.K313E Chr.3: 129260398-129260398 on Build GRCh38
6305085 c.956C>T p.P319L Chr.3: 129260635-129260635 on Build GRCh38
3060010 c.1072G>A p.D358N Chr.3: 129260751-129260751 on Build GRCh38
3774501 c.891C>T p.S297S Chr.3: 129260352-129260352 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768922461 Chr.3: 129260723-129260723 on Build GRCh38
rs573184481 Chr.3: 129260688-129260688 on Build GRCh38
rs759243223 Chr.3: 129260649-129260649 on Build GRCh38
rs551440948 Chr.3: 129260494-129260494 on Build GRCh38
rs763074625 Chr.3: 129260386-129260386 on Build GRCh38
rs761615073 Chr.3: 129260307-129260307 on Build GRCh38
rs1018343224 Chr.3: 129260233-129260233 on Build GRCh38
rs761725791 Chr.3: 129260405-129260405 on Build GRCh38
rs753512743 Chr.3: 129260447-129260447 on Build GRCh38
rs765976698 Chr.3: 129260431-129260431 on Build GRCh38
rs199810158 Chr.3: 129260284-129260284 on Build GRCh38
rs6802965 Chr.3: 129260413-129260413 on Build GRCh38
rs754515422 Chr.3: 129260341-129260341 on Build GRCh38
rs770525944 Chr.3: 129260363-129260363 on Build GRCh38
rs774282553 Chr.3: 129260397-129260397 on Build GRCh38
rs753616364 Chr.3: 129260606-129260606 on Build GRCh38
rs934185904 Chr.3: 129260564-129260564 on Build GRCh38
rs749511220 Chr.3: 129260708-129260708 on Build GRCh38
rs767165798 Chr.3: 129260396-129260396 on Build GRCh38
rs201750714 Chr.3: 129260319-129260319 on Build GRCh38
rs771946058 Chr.3: 129260635-129260635 on Build GRCh38
rs760542756 Chr.3: 129260490-129260490 on Build GRCh38
rs374818510 Chr.3: 129260292-129260292 on Build GRCh38
rs764088237 Chr.3: 129260596-129260596 on Build GRCh38
rs368438594 Chr.3: 129260375-129260375 on Build GRCh38
rs377553599 Chr.3: 129260697-129260697 on Build GRCh38
rs756483626 Chr.3: 129260704-129260704 on Build GRCh38
rs990785091 Chr.3: 129260219-129260219 on Build GRCh38
rs918809057 Chr.3: 129260518-129260518 on Build GRCh38
rs1034357565 Chr.3: 129260594-129260594 on Build GRCh38
rs111804975 Chr.3: 129260519-129260519 on Build GRCh38
rs556813387 Chr.3: 129260323-129260323 on Build GRCh38
rs371307852 Chr.3: 129260283-129260283 on Build GRCh38
rs780369112 Chr.3: 129260597-129260597 on Build GRCh38
rs892788412 Chr.3: 129260310-129260311 on Build GRCh38
rs757054397 Chr.3: 129260653-129260653 on Build GRCh38
rs4927907 Chr.3: 129260221-129260221 on Build GRCh38
rs886963820 Chr.3: 129260499-129260499 on Build GRCh38
rs374232037 Chr.3: 129260602-129260602 on Build GRCh38
rs767223949 Chr.3: 129260309-129260309 on Build GRCh38
rs1037669770 Chr.3: 129260254-129260254 on Build GRCh38
rs757955471 Chr.3: 129260691-129260691 on Build GRCh38
rs201228229 Chr.3: 129260372-129260372 on Build GRCh38
rs201897170 Chr.3: 129260579-129260579 on Build GRCh38
rs776261640 Chr.3: 129260647-129260647 on Build GRCh38
rs752186723 Chr.3: 129260678-129260678 on Build GRCh38
rs780636747 Chr.3: 129260705-129260705 on Build GRCh38
rs778283780 Chr.3: 129260356-129260356 on Build GRCh38
rs757750673 Chr.3: 129260582-129260582 on Build GRCh38
rs569369828 Chr.3: 129260264-129260264 on Build GRCh38
rs751478484 Chr.3: 129260303-129260303 on Build GRCh38
rs77079402 Chr.3: 129260272-129260272 on Build GRCh38
rs767858436 Chr.3: 129260692-129260692 on Build GRCh38
rs773232882 Chr.3: 129260636-129260636 on Build GRCh38
rs760414395 Chr.3: 129260638-129260638 on Build GRCh38
rs759213346 Chr.3: 129260388-129260388 on Build GRCh38
rs750089239 Chr.3: 129260310-129260310 on Build GRCh38
rs546335559 Chr.3: 129260528-129260528 on Build GRCh38
rs941631769 Chr.3: 129260228-129260228 on Build GRCh38
rs755207836 Chr.3: 129260717-129260717 on Build GRCh38
rs767333882 Chr.3: 129260407-129260407 on Build GRCh38
rs535308051 Chr.3: 129260317-129260317 on Build GRCh38
rs199838147 Chr.3: 129260362-129260362 on Build GRCh38
rs1024751528 Chr.3: 129260521-129260521 on Build GRCh38
rs746062966 Chr.3: 129260373-129260373 on Build GRCh38
rs775881986 Chr.3: 129260383-129260383 on Build GRCh38
rs748097243 Chr.3: 129260609-129260609 on Build GRCh38
rs575780982 Chr.3: 129260457-129260457 on Build GRCh38
rs760291842 Chr.3: 129260414-129260414 on Build GRCh38
rs746190956 Chr.3: 129260587-129260587 on Build GRCh38
rs373912605 Chr.3: 129260294-129260294 on Build GRCh38
rs766166706 Chr.3: 129260642-129260642 on Build GRCh38
rs781586035 Chr.3: 129260583-129260583 on Build GRCh38
rs558168165 Chr.3: 129260574-129260574 on Build GRCh38
rs1004027295 Chr.3: 129260507-129260507 on Build GRCh38
rs200063755 Chr.3: 129260732-129260732 on Build GRCh38
rs368625942 Chr.3: 129260452-129260452 on Build GRCh38
rs879157064 Chr.3: 129260616-129260616 on Build GRCh38
rs4927906 Chr.3: 129260265-129260265 on Build GRCh38
rs753386418 Chr.3: 129260334-129260334 on Build GRCh38
rs1039409256 Chr.3: 129260433-129260433 on Build GRCh38
rs1024827214 Chr.3: 129260273-129260273 on Build GRCh38
rs563072762 Chr.3: 129260268-129260268 on Build GRCh38
rs368123292 Chr.3: 129260271-129260271 on Build GRCh38
rs748140089 Chr.3: 129260738-129260738 on Build GRCh38
rs772216561 Chr.3: 129260742-129260742 on Build GRCh38
rs1054120767 Chr.3: 129260625-129260625 on Build GRCh38
rs764781011 Chr.3: 129260672-129260672 on Build GRCh38

View Full Product Details