Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 154405615-154406199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNKSR3
Entrez Gene ID
154043
Gene Name
CNKSR family member 3
Gene Aliases
MAGI1
UniGene
Hs.16064
Gene Chromosome Location
Chr.6: 154405299-154510619 on Build GRCh38
Transcripts
NM_173515.2 XM_011535485.2

SNP Detail

SNP ID Chromosome Location
rs1027518844 Chr.6: 154406118-154406118 on Build GRCh38
rs541916429 Chr.6: 154405762-154405762 on Build GRCh38
rs2275336 Chr.6: 154405854-154405854 on Build GRCh38
rs894689631 Chr.6: 154405772-154405772 on Build GRCh38
rs10603494 Chr.6: 154405886-154405889 on Build GRCh38
rs777358899 Chr.6: 154405979-154405979 on Build GRCh38
rs548270161 Chr.6: 154406022-154406022 on Build GRCh38
rs138147314 Chr.6: 154406070-154406070 on Build GRCh38
rs763116327 Chr.6: 154406081-154406082 on Build GRCh38
rs558493619 Chr.6: 154405780-154405780 on Build GRCh38
rs921484598 Chr.6: 154405756-154405756 on Build GRCh38
rs41292908 Chr.6: 154405848-154405848 on Build GRCh38
rs1014014257 Chr.6: 154405660-154405660 on Build GRCh38
rs994500831 Chr.6: 154406106-154406109 on Build GRCh38
rs973186581 Chr.6: 154405695-154405695 on Build GRCh38
rs938686799 Chr.6: 154405766-154405766 on Build GRCh38
rs1055548586 Chr.6: 154405943-154405943 on Build GRCh38
rs975080377 Chr.6: 154406050-154406050 on Build GRCh38
rs556327850 Chr.6: 154406150-154406150 on Build GRCh38
rs1016712310 Chr.6: 154406023-154406023 on Build GRCh38
rs987087018 Chr.6: 154406078-154406078 on Build GRCh38
rs1025378605 Chr.6: 154406151-154406151 on Build GRCh38
rs754165979 Chr.6: 154405885-154405886 on Build GRCh38
rs1043313638 Chr.6: 154406044-154406044 on Build GRCh38
rs962787789 Chr.6: 154406034-154406034 on Build GRCh38
rs367546542 Chr.6: 154405889-154405892 on Build GRCh38
rs1043856863 Chr.6: 154405855-154405855 on Build GRCh38
rs550336961 Chr.6: 154405990-154405990 on Build GRCh38
rs532148510 Chr.6: 154405938-154405938 on Build GRCh38
rs34224990 Chr.6: 154405934-154405934 on Build GRCh38
rs544246864 Chr.6: 154405704-154405704 on Build GRCh38
rs41292906 Chr.6: 154405711-154405711 on Build GRCh38
rs567076869 Chr.6: 154406066-154406066 on Build GRCh38
rs925363266 Chr.6: 154406100-154406100 on Build GRCh38
rs559219295 Chr.6: 154406102-154406102 on Build GRCh38
rs745825093 Chr.6: 154406096-154406096 on Build GRCh38
rs145415328 Chr.6: 154405992-154405992 on Build GRCh38
rs113404262 Chr.6: 154405916-154405916 on Build GRCh38
rs976054757 Chr.6: 154405744-154405744 on Build GRCh38
rs973280581 Chr.6: 154405909-154405909 on Build GRCh38
rs1008529877 Chr.6: 154405867-154405867 on Build GRCh38
rs948731545 Chr.6: 154406043-154406043 on Build GRCh38
rs559247753 Chr.6: 154405669-154405669 on Build GRCh38
rs934251701 Chr.6: 154405820-154405820 on Build GRCh38
rs929160358 Chr.6: 154405706-154405706 on Build GRCh38
rs529857436 Chr.6: 154405996-154405996 on Build GRCh38
rs1057160377 Chr.6: 154405769-154405769 on Build GRCh38
rs1034295190 Chr.6: 154405645-154405645 on Build GRCh38
rs886980531 Chr.6: 154406007-154406007 on Build GRCh38
rs1027997372 Chr.6: 154406068-154406068 on Build GRCh38
rs945589979 Chr.6: 154406084-154406084 on Build GRCh38
rs560789204 Chr.6: 154405838-154405838 on Build GRCh38
rs982585379 Chr.6: 154405672-154405672 on Build GRCh38
rs958519001 Chr.6: 154405651-154405651 on Build GRCh38
rs111970860 Chr.6: 154405774-154405774 on Build GRCh38
rs571093746 Chr.6: 154406103-154406103 on Build GRCh38
rs904679212 Chr.6: 154405839-154405839 on Build GRCh38
rs909535501 Chr.6: 154405702-154405702 on Build GRCh38
rs914071376 Chr.6: 154406080-154406080 on Build GRCh38
rs565002809 Chr.6: 154405879-154405879 on Build GRCh38
rs904812407 Chr.6: 154406045-154406045 on Build GRCh38
rs950575281 Chr.6: 154405982-154405982 on Build GRCh38
rs200975287 Chr.6: 154405889-154405889 on Build GRCh38
rs755611715 Chr.6: 154405872-154405872 on Build GRCh38
rs951100603 Chr.6: 154405671-154405671 on Build GRCh38
rs574863649 Chr.6: 154405721-154405721 on Build GRCh38
rs577190579 Chr.6: 154405674-154405674 on Build GRCh38
rs74920838 Chr.6: 154406130-154406130 on Build GRCh38

View Full Product Details