Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28452362-28452904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO16
Entrez Gene ID
157574
Gene Name
F-box protein 16
Gene Aliases
FBX16
UniGene
Hs.741522
Gene Chromosome Location
Chr.8: 28428408-28490318 on Build GRCh38
Transcripts
NM_001258211.1 NM_172366.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4751762 c.599G>A p.R200Q Chr.8: 28452385-28452385 on Build GRCh38
6180730 c.549A>G p.L183L Chr.8: 28452435-28452435 on Build GRCh38
3648006 c.519G>A p.K173K Chr.8: 28452465-28452465 on Build GRCh38
2788602 c.598C>T p.R200W Chr.8: 28452386-28452386 on Build GRCh38
4589857 c.522_523insA p.G175fs*6 Chr.8: 28452461-28452462 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs942262337 Chr.8: 28452625-28452625 on Build GRCh38
rs141686487 Chr.8: 28452789-28452789 on Build GRCh38
rs530000588 Chr.8: 28452528-28452528 on Build GRCh38
rs761550556 Chr.8: 28452803-28452804 on Build GRCh38
rs397891454 Chr.8: 28452516-28452517 on Build GRCh38
rs983519604 Chr.8: 28452589-28452589 on Build GRCh38
rs754859131 Chr.8: 28452450-28452450 on Build GRCh38
rs758524216 Chr.8: 28452512-28452512 on Build GRCh38
rs973307715 Chr.8: 28452644-28452644 on Build GRCh38
rs907673077 Chr.8: 28452621-28452621 on Build GRCh38
rs191927649 Chr.8: 28452539-28452539 on Build GRCh38
rs780209458 Chr.8: 28452513-28452513 on Build GRCh38
rs960045340 Chr.8: 28452844-28452844 on Build GRCh38
rs752459879 Chr.8: 28452498-28452498 on Build GRCh38
rs762809282 Chr.8: 28452444-28452444 on Build GRCh38
rs35042301 Chr.8: 28452514-28452515 on Build GRCh38
rs773801245 Chr.8: 28452446-28452446 on Build GRCh38
rs1012155759 Chr.8: 28452804-28452808 on Build GRCh38
rs185767660 Chr.8: 28452518-28452518 on Build GRCh38
rs375104603 Chr.8: 28452431-28452431 on Build GRCh38
rs375993733 Chr.8: 28452449-28452449 on Build GRCh38
rs777601007 Chr.8: 28452519-28452519 on Build GRCh38
rs577122351 Chr.8: 28452740-28452740 on Build GRCh38
rs777844947 Chr.8: 28452568-28452568 on Build GRCh38
rs369379563 Chr.8: 28452400-28452400 on Build GRCh38
rs566672358 Chr.8: 28452537-28452537 on Build GRCh38
rs139855119 Chr.8: 28452858-28452858 on Build GRCh38
rs769607803 Chr.8: 28452419-28452419 on Build GRCh38
rs754315496 Chr.8: 28452486-28452486 on Build GRCh38
rs1052410210 Chr.8: 28452765-28452765 on Build GRCh38
rs891970706 Chr.8: 28452511-28452512 on Build GRCh38
rs1043261455 Chr.8: 28452820-28452823 on Build GRCh38
rs906094352 Chr.8: 28452830-28452830 on Build GRCh38
rs137863210 Chr.8: 28452733-28452733 on Build GRCh38
rs567996211 Chr.8: 28452670-28452670 on Build GRCh38
rs764560735 Chr.8: 28452472-28452472 on Build GRCh38
rs1001763641 Chr.8: 28452832-28452832 on Build GRCh38
rs372068495 Chr.8: 28452626-28452626 on Build GRCh38
rs749245600 Chr.8: 28452520-28452520 on Build GRCh38
rs952410726 Chr.8: 28452411-28452411 on Build GRCh38
rs111399097 Chr.8: 28452689-28452689 on Build GRCh38
rs541993578 Chr.8: 28452864-28452864 on Build GRCh38
rs750514213 Chr.8: 28452499-28452499 on Build GRCh38
rs1041684924 Chr.8: 28452458-28452458 on Build GRCh38
rs964563701 Chr.8: 28452682-28452682 on Build GRCh38
rs1052360154 Chr.8: 28452480-28452480 on Build GRCh38
rs747889711 Chr.8: 28452414-28452414 on Build GRCh38
rs993847120 Chr.8: 28452576-28452576 on Build GRCh38
rs1025288849 Chr.8: 28452581-28452581 on Build GRCh38
rs753376225 Chr.8: 28452406-28452406 on Build GRCh38
rs747235939 Chr.8: 28452516-28452516 on Build GRCh38
rs747195706 Chr.8: 28452658-28452658 on Build GRCh38
rs145958705 Chr.8: 28452834-28452834 on Build GRCh38
rs752423663 Chr.8: 28452470-28452470 on Build GRCh38
rs984321769 Chr.8: 28452424-28452424 on Build GRCh38
rs1052190713 Chr.8: 28452800-28452800 on Build GRCh38
rs745824099 Chr.8: 28452864-28452865 on Build GRCh38
rs770462983 Chr.8: 28452445-28452445 on Build GRCh38
rs546960357 Chr.8: 28452533-28452533 on Build GRCh38
rs935564216 Chr.8: 28452761-28452761 on Build GRCh38
rs112300865 Chr.8: 28452575-28452575 on Build GRCh38
rs778438604 Chr.8: 28452474-28452477 on Build GRCh38
rs138522609 Chr.8: 28452402-28452402 on Build GRCh38
rs149428353 Chr.8: 28452405-28452405 on Build GRCh38
rs963535680 Chr.8: 28452859-28452861 on Build GRCh38
rs747801384 Chr.8: 28452479-28452481 on Build GRCh38
rs984262695 Chr.8: 28452657-28452657 on Build GRCh38
rs1045556042 Chr.8: 28452839-28452839 on Build GRCh38
rs757768910 Chr.8: 28452495-28452495 on Build GRCh38
rs976914537 Chr.8: 28452732-28452732 on Build GRCh38
rs770819079 Chr.8: 28452848-28452850 on Build GRCh38
rs921842187 Chr.8: 28452702-28452702 on Build GRCh38
rs554525855 Chr.8: 28452630-28452630 on Build GRCh38
rs765205390 Chr.8: 28452473-28452473 on Build GRCh38
rs754748031 Chr.8: 28452414-28452415 on Build GRCh38
rs377149575 Chr.8: 28452598-28452598 on Build GRCh38
rs1001782035 Chr.8: 28452881-28452881 on Build GRCh38
rs144465484 Chr.8: 28452552-28452552 on Build GRCh38
rs922819385 Chr.8: 28452459-28452459 on Build GRCh38
rs534212467 Chr.8: 28452694-28452694 on Build GRCh38
rs778914888 Chr.8: 28452401-28452401 on Build GRCh38
rs773065842 Chr.8: 28452437-28452437 on Build GRCh38
rs1042924742 Chr.8: 28452529-28452529 on Build GRCh38
rs532300677 Chr.8: 28452538-28452538 on Build GRCh38

View Full Product Details