Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 154187347-154187819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF26
Entrez Gene ID
26084
Gene Name
Rho guanine nucleotide exchange factor 26
Gene Aliases
CSGEF, HMFN1864, SGEF
UniGene
Hs.240845
Gene Chromosome Location
Chr.3: 154121003-154257827 on Build GRCh38
Transcripts
NM_001251962.1 NM_001251963.1 NM_015595.3 XM_011512672.1 XM_017006160.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1040148 c.1622G>A p.R541Q Chr.3: 154187819-154187819 on Build GRCh38
1040145 c.1538A>T p.D513V Chr.3: 154187735-154187735 on Build GRCh38
4977787 c.1621C>T p.R541* Chr.3: 154187818-154187818 on Build GRCh38
5659232 c.1552G>T p.V518L Chr.3: 154187749-154187749 on Build GRCh38
5501505 c.1611C>T p.V537V Chr.3: 154187808-154187808 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772566646 Chr.3: 154187596-154187596 on Build GRCh38
rs764194661 Chr.3: 154187636-154187636 on Build GRCh38
rs765375391 Chr.3: 154187767-154187767 on Build GRCh38
rs769397878 Chr.3: 154187693-154187693 on Build GRCh38
rs530204792 Chr.3: 154187631-154187631 on Build GRCh38
rs1048521843 Chr.3: 154187581-154187581 on Build GRCh38
rs775741517 Chr.3: 154187660-154187660 on Build GRCh38
rs35918903 Chr.3: 154187399-154187400 on Build GRCh38
rs996787161 Chr.3: 154187488-154187488 on Build GRCh38
rs920345367 Chr.3: 154187611-154187611 on Build GRCh38
rs1010331917 Chr.3: 154187511-154187511 on Build GRCh38
rs781264698 Chr.3: 154187676-154187676 on Build GRCh38
rs924667141 Chr.3: 154187515-154187515 on Build GRCh38
rs746787364 Chr.3: 154187670-154187670 on Build GRCh38
rs959825636 Chr.3: 154187599-154187599 on Build GRCh38
rs772871529 Chr.3: 154187701-154187701 on Build GRCh38
rs756334014 Chr.3: 154187413-154187413 on Build GRCh38
rs947387907 Chr.3: 154187486-154187486 on Build GRCh38
rs759804204 Chr.3: 154187723-154187723 on Build GRCh38
rs377034117 Chr.3: 154187380-154187381 on Build GRCh38
rs143464714 Chr.3: 154187494-154187494 on Build GRCh38
rs748912005 Chr.3: 154187710-154187710 on Build GRCh38
rs6781620 Chr.3: 154187426-154187426 on Build GRCh38
rs956068222 Chr.3: 154187692-154187692 on Build GRCh38
rs995289304 Chr.3: 154187578-154187578 on Build GRCh38
rs757436159 Chr.3: 154187764-154187764 on Build GRCh38
rs1054875837 Chr.3: 154187399-154187399 on Build GRCh38
rs757447601 Chr.3: 154187664-154187664 on Build GRCh38
rs886812158 Chr.3: 154187593-154187593 on Build GRCh38
rs943549093 Chr.3: 154187555-154187555 on Build GRCh38
rs779843965 Chr.3: 154187665-154187665 on Build GRCh38
rs183851298 Chr.3: 154187730-154187730 on Build GRCh38
rs937855536 Chr.3: 154187388-154187388 on Build GRCh38
rs369279368 Chr.3: 154187451-154187451 on Build GRCh38
rs191762970 Chr.3: 154187531-154187531 on Build GRCh38
rs1009131729 Chr.3: 154187473-154187473 on Build GRCh38
rs959898150 Chr.3: 154187395-154187395 on Build GRCh38
rs554546806 Chr.3: 154187643-154187643 on Build GRCh38
rs543945754 Chr.3: 154187747-154187747 on Build GRCh38
rs766135978 Chr.3: 154187659-154187660 on Build GRCh38
rs779493610 Chr.3: 154187709-154187709 on Build GRCh38
rs769253478 Chr.3: 154187492-154187492 on Build GRCh38
rs115792432 Chr.3: 154187475-154187475 on Build GRCh38
rs376544460 Chr.3: 154187400-154187400 on Build GRCh38
rs764185574 Chr.3: 154187788-154187788 on Build GRCh38
rs1035608067 Chr.3: 154187598-154187598 on Build GRCh38
rs770493001 Chr.3: 154187711-154187711 on Build GRCh38
rs775941680 Chr.3: 154187729-154187729 on Build GRCh38
rs1047880763 Chr.3: 154187448-154187448 on Build GRCh38
rs890585148 Chr.3: 154187551-154187551 on Build GRCh38
rs1052007199 Chr.3: 154187537-154187537 on Build GRCh38
rs750717319 Chr.3: 154187781-154187781 on Build GRCh38
rs766215111 Chr.3: 154187650-154187650 on Build GRCh38
rs991530493 Chr.3: 154187457-154187457 on Build GRCh38
rs748146786 Chr.3: 154187684-154187684 on Build GRCh38
rs6794673 Chr.3: 154187402-154187402 on Build GRCh38
rs1012767272 Chr.3: 154187610-154187610 on Build GRCh38
rs1039400960 Chr.3: 154187569-154187569 on Build GRCh38
rs202052244 Chr.3: 154187652-154187652 on Build GRCh38
rs147136183 Chr.3: 154187572-154187572 on Build GRCh38
rs920381620 Chr.3: 154187411-154187411 on Build GRCh38
rs774117651 Chr.3: 154187717-154187717 on Build GRCh38
rs753931512 Chr.3: 154187739-154187739 on Build GRCh38
rs760983064 Chr.3: 154187753-154187753 on Build GRCh38
rs1035511763 Chr.3: 154187386-154187386 on Build GRCh38
rs1055001542 Chr.3: 154187744-154187744 on Build GRCh38
rs930308531 Chr.3: 154187422-154187422 on Build GRCh38
rs561487483 Chr.3: 154187728-154187728 on Build GRCh38
rs1033079286 Chr.3: 154187489-154187489 on Build GRCh38
rs796544085 Chr.3: 154187375-154187375 on Build GRCh38
rs569124579 Chr.3: 154187625-154187625 on Build GRCh38
rs540229946 Chr.3: 154187671-154187671 on Build GRCh38

View Full Product Details