Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 130051985-130052571 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASAP1
Entrez Gene ID
50807
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 1
Gene Aliases
AMAP1, CENTB4, DDEF1, PAG2, PAP, ZG14P
UniGene
Hs.655552
Gene Chromosome Location
Chr.8: 130052104-130443848 on Build GRCh38
Transcripts
NM_001247996.1 NM_018482.3 XM_005250925.1 XM_006716563.3 XM_006716564.1 XM_006716565.3 XM_006716566.1 XM_011517052.2 XM_011517053.1 XM_017013467.1 XM_017013468.1 XM_017013469.1

SNP Detail

SNP ID Chromosome Location
rs936061535 Chr.8: 130052335-130052335 on Build GRCh38
rs1018430145 Chr.8: 130052121-130052121 on Build GRCh38
rs754581461 Chr.8: 130052172-130052172 on Build GRCh38
rs977060754 Chr.8: 130052176-130052176 on Build GRCh38
rs967110462 Chr.8: 130052110-130052110 on Build GRCh38
rs964541883 Chr.8: 130052126-130052129 on Build GRCh38
rs926052816 Chr.8: 130052333-130052333 on Build GRCh38
rs900360599 Chr.8: 130052048-130052048 on Build GRCh38
rs549915130 Chr.8: 130052531-130052531 on Build GRCh38
rs977357642 Chr.8: 130052306-130052306 on Build GRCh38
rs560474729 Chr.8: 130052220-130052220 on Build GRCh38
rs1048732775 Chr.8: 130052541-130052541 on Build GRCh38
rs944355404 Chr.8: 130052526-130052526 on Build GRCh38
rs527272520 Chr.8: 130052022-130052022 on Build GRCh38
rs944645266 Chr.8: 130052027-130052027 on Build GRCh38
rs1035668313 Chr.8: 130052123-130052123 on Build GRCh38
rs36017481 Chr.8: 130052536-130052537 on Build GRCh38
rs531928466 Chr.8: 130052433-130052433 on Build GRCh38
rs991513604 Chr.8: 130052384-130052384 on Build GRCh38
rs1014192960 Chr.8: 130052032-130052032 on Build GRCh38
rs951785444 Chr.8: 130052299-130052299 on Build GRCh38
rs991005739 Chr.8: 130052247-130052247 on Build GRCh38
rs959552504 Chr.8: 130052156-130052156 on Build GRCh38
rs1003833735 Chr.8: 130052114-130052114 on Build GRCh38
rs754555752 Chr.8: 130052455-130052455 on Build GRCh38
rs1020956958 Chr.8: 130052105-130052105 on Build GRCh38
rs897699302 Chr.8: 130052498-130052498 on Build GRCh38
rs949914776 Chr.8: 130052414-130052415 on Build GRCh38
rs1030320291 Chr.8: 130052257-130052257 on Build GRCh38
rs893905351 Chr.8: 130052063-130052063 on Build GRCh38
rs907565413 Chr.8: 130052438-130052438 on Build GRCh38
rs915773826 Chr.8: 130052391-130052391 on Build GRCh38
rs894413384 Chr.8: 130052019-130052019 on Build GRCh38
rs545429576 Chr.8: 130052030-130052030 on Build GRCh38
rs1010909969 Chr.8: 130052081-130052081 on Build GRCh38
rs781649457 Chr.8: 130052487-130052487 on Build GRCh38
rs1054192358 Chr.8: 130052060-130052060 on Build GRCh38
rs967405840 Chr.8: 130052286-130052286 on Build GRCh38
rs574537626 Chr.8: 130052315-130052316 on Build GRCh38
rs920734002 Chr.8: 130052292-130052292 on Build GRCh38
rs1027411134 Chr.8: 130052098-130052098 on Build GRCh38
rs983569327 Chr.8: 130052351-130052351 on Build GRCh38
rs6470796 Chr.8: 130052053-130052053 on Build GRCh38

View Full Product Details