Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 175008304-175008839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACYBP
Entrez Gene ID
27101
Gene Name
calcyclin binding protein
Gene Aliases
GIG5, PNAS-107, S100A6BP, SIP
UniGene
Hs.508524
Gene Chromosome Location
Chr.1: 174999435-175012027 on Build GRCh38
Transcripts
NM_001007214.1 NM_014412.2 XM_017001046.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4778953 c.456A>G p.I152M Chr.1: 175008632-175008632 on Build GRCh38
6618799 c.463_466delAGAA p.K157fs*9 Chr.1: 175008639-175008642 on Build GRCh38
4550659 c.505G>T p.V169F Chr.1: 175008681-175008681 on Build GRCh38
6563441 c.511A>C p.K171Q Chr.1: 175008687-175008687 on Build GRCh38
1473017 c.474G>A p.V158V Chr.1: 175008650-175008650 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916459779 Chr.1: 175008515-175008515 on Build GRCh38
rs1030458817 Chr.1: 175008805-175008805 on Build GRCh38
rs535491219 Chr.1: 175008506-175008506 on Build GRCh38
rs1025592012 Chr.1: 175008375-175008375 on Build GRCh38
rs78902398 Chr.1: 175008715-175008715 on Build GRCh38
rs1048324594 Chr.1: 175008387-175008387 on Build GRCh38
rs1007078037 Chr.1: 175008325-175008325 on Build GRCh38
rs953382555 Chr.1: 175008526-175008526 on Build GRCh38
rs772852816 Chr.1: 175008706-175008706 on Build GRCh38
rs988760221 Chr.1: 175008606-175008606 on Build GRCh38
rs765577153 Chr.1: 175008728-175008728 on Build GRCh38
rs769492990 Chr.1: 175008358-175008359 on Build GRCh38
rs771704453 Chr.1: 175008469-175008469 on Build GRCh38
rs749334544 Chr.1: 175008716-175008716 on Build GRCh38
rs572313069 Chr.1: 175008457-175008457 on Build GRCh38
rs901390173 Chr.1: 175008757-175008757 on Build GRCh38
rs772433099 Chr.1: 175008629-175008629 on Build GRCh38
rs976683270 Chr.1: 175008374-175008374 on Build GRCh38
rs559835241 Chr.1: 175008613-175008613 on Build GRCh38
rs1004501272 Chr.1: 175008424-175008424 on Build GRCh38
rs747587553 Chr.1: 175008641-175008641 on Build GRCh38
rs143328471 Chr.1: 175008721-175008721 on Build GRCh38
rs778246676 Chr.1: 175008737-175008737 on Build GRCh38
rs1007436667 Chr.1: 175008750-175008750 on Build GRCh38
rs562779249 Chr.1: 175008754-175008754 on Build GRCh38
rs774026693 Chr.1: 175008758-175008758 on Build GRCh38
rs780223023 Chr.1: 175008581-175008581 on Build GRCh38
rs753947654 Chr.1: 175008583-175008583 on Build GRCh38
rs1018915081 Chr.1: 175008359-175008359 on Build GRCh38
rs539892444 Chr.1: 175008551-175008551 on Build GRCh38
rs745985565 Chr.1: 175008582-175008583 on Build GRCh38
rs768110141 Chr.1: 175008756-175008756 on Build GRCh38
rs888354917 Chr.1: 175008396-175008396 on Build GRCh38
rs775036033 Chr.1: 175008662-175008662 on Build GRCh38
rs755049178 Chr.1: 175008725-175008725 on Build GRCh38
rs1015428348 Chr.1: 175008443-175008443 on Build GRCh38
rs762442803 Chr.1: 175008672-175008672 on Build GRCh38
rs758517552 Chr.1: 175008732-175008732 on Build GRCh38
rs941828447 Chr.1: 175008473-175008473 on Build GRCh38
rs1046382856 Chr.1: 175008557-175008557 on Build GRCh38
rs150031571 Chr.1: 175008690-175008690 on Build GRCh38
rs771316635 Chr.1: 175008650-175008650 on Build GRCh38
rs901034588 Chr.1: 175008775-175008775 on Build GRCh38
rs984593080 Chr.1: 175008450-175008450 on Build GRCh38
rs757847038 Chr.1: 175008741-175008741 on Build GRCh38
rs551544326 Chr.1: 175008807-175008807 on Build GRCh38
rs888069084 Chr.1: 175008746-175008746 on Build GRCh38
rs113031677 Chr.1: 175008573-175008573 on Build GRCh38
rs6678823 Chr.1: 175008446-175008446 on Build GRCh38
rs113585434 Chr.1: 175008560-175008560 on Build GRCh38
rs779208262 Chr.1: 175008591-175008591 on Build GRCh38
rs761532555 Chr.1: 175008564-175008564 on Build GRCh38
rs1046107680 Chr.1: 175008330-175008330 on Build GRCh38
rs773937358 Chr.1: 175008683-175008683 on Build GRCh38
rs1025876576 Chr.1: 175008803-175008803 on Build GRCh38
rs949278404 Chr.1: 175008527-175008527 on Build GRCh38
rs754040198 Chr.1: 175008724-175008724 on Build GRCh38
rs1001814361 Chr.1: 175008384-175008384 on Build GRCh38
rs755310650 Chr.1: 175008585-175008585 on Build GRCh38
rs768304595 Chr.1: 175008681-175008681 on Build GRCh38
rs748535167 Chr.1: 175008599-175008599 on Build GRCh38
rs756179966 Chr.1: 175008577-175008577 on Build GRCh38
rs377382736 Chr.1: 175008565-175008565 on Build GRCh38
rs369805680 Chr.1: 175008559-175008559 on Build GRCh38
rs778172955 Chr.1: 175008639-175008639 on Build GRCh38
rs750330318 Chr.1: 175008574-175008574 on Build GRCh38
rs373727208 Chr.1: 175008744-175008744 on Build GRCh38
rs115397485 Chr.1: 175008365-175008365 on Build GRCh38
rs951358886 Chr.1: 175008819-175008819 on Build GRCh38
rs565791192 Chr.1: 175008784-175008784 on Build GRCh38
rs763433103 Chr.1: 175008433-175008433 on Build GRCh38
rs760716194 Chr.1: 175008712-175008712 on Build GRCh38

View Full Product Details