Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 170983095-170983518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MROH9
Entrez Gene ID
80133
Gene Name
maestro heat like repeat family member 9
Gene Aliases
ARMC11, C1orf129
UniGene
Hs.591489
Gene Chromosome Location
Chr.1: 170935471-171066341 on Build GRCh38
Transcripts
NM_001163629.1 NM_025063.2 XM_011510005.2 XM_011510006.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3478211 c.667C>T p.P223S Chr.1: 170983472-170983472 on Build GRCh38
3478212 c.667C>T p.P223S Chr.1: 170983472-170983472 on Build GRCh38
5777026 c.630T>G p.N210K Chr.1: 170983435-170983435 on Build GRCh38
5567709 c.656G>T p.S219I Chr.1: 170983461-170983461 on Build GRCh38
7171794 c.617-1G>C p.? Chr.1: 170983421-170983421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376392385 Chr.1: 170983403-170983403 on Build GRCh38
rs758710250 Chr.1: 170983468-170983468 on Build GRCh38
rs201549824 Chr.1: 170983485-170983485 on Build GRCh38
rs369675316 Chr.1: 170983160-170983160 on Build GRCh38
rs370652735 Chr.1: 170983381-170983381 on Build GRCh38
rs1047826740 Chr.1: 170983278-170983278 on Build GRCh38
rs116251875 Chr.1: 170983456-170983456 on Build GRCh38
rs752904080 Chr.1: 170983458-170983458 on Build GRCh38
rs988271721 Chr.1: 170983209-170983209 on Build GRCh38
rs536309064 Chr.1: 170983482-170983482 on Build GRCh38
rs761520213 Chr.1: 170983421-170983421 on Build GRCh38
rs147253408 Chr.1: 170983292-170983292 on Build GRCh38
rs898993551 Chr.1: 170983455-170983455 on Build GRCh38
rs762495463 Chr.1: 170983390-170983390 on Build GRCh38
rs1008599648 Chr.1: 170983282-170983282 on Build GRCh38
rs975660190 Chr.1: 170983281-170983281 on Build GRCh38
rs375869633 Chr.1: 170983280-170983280 on Build GRCh38
rs1044603401 Chr.1: 170983246-170983246 on Build GRCh38
rs779204268 Chr.1: 170983457-170983457 on Build GRCh38
rs371155414 Chr.1: 170983425-170983425 on Build GRCh38
rs940531009 Chr.1: 170983426-170983426 on Build GRCh38
rs917355188 Chr.1: 170983198-170983198 on Build GRCh38
rs762206507 Chr.1: 170983473-170983475 on Build GRCh38
rs191819562 Chr.1: 170983308-170983308 on Build GRCh38
rs764687608 Chr.1: 170983404-170983404 on Build GRCh38
rs373670994 Chr.1: 170983435-170983435 on Build GRCh38
rs773980157 Chr.1: 170983404-170983404 on Build GRCh38
rs367575706 Chr.1: 170983447-170983447 on Build GRCh38
rs767232814 Chr.1: 170983422-170983422 on Build GRCh38
rs760479378 Chr.1: 170983438-170983438 on Build GRCh38
rs946987046 Chr.1: 170983251-170983251 on Build GRCh38
rs995974129 Chr.1: 170983309-170983309 on Build GRCh38
rs774824147 Chr.1: 170983384-170983384 on Build GRCh38
rs761315275 Chr.1: 170983404-170983405 on Build GRCh38
rs979130597 Chr.1: 170983145-170983145 on Build GRCh38
rs199994744 Chr.1: 170983487-170983487 on Build GRCh38
rs930914935 Chr.1: 170983261-170983261 on Build GRCh38
rs915506644 Chr.1: 170983217-170983217 on Build GRCh38
rs539531291 Chr.1: 170983159-170983159 on Build GRCh38
rs573162421 Chr.1: 170983222-170983222 on Build GRCh38
rs1031364738 Chr.1: 170983480-170983480 on Build GRCh38
rs948886768 Chr.1: 170983237-170983237 on Build GRCh38
rs766369095 Chr.1: 170983443-170983443 on Build GRCh38
rs754322656 Chr.1: 170983455-170983455 on Build GRCh38
rs967999015 Chr.1: 170983142-170983142 on Build GRCh38
rs982240542 Chr.1: 170983131-170983131 on Build GRCh38
rs373241096 Chr.1: 170983391-170983391 on Build GRCh38

View Full Product Details