Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 214618810-214619374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPF
Entrez Gene ID
1063
Gene Name
centromere protein F
Gene Aliases
CENF, CILD31, PRO1779, STROMS, hcp-1
UniGene
Hs.497741
Gene Chromosome Location
Chr.1: 214603181-214664574 on Build GRCh38
Transcripts
NM_016343.3 XM_011509082.2 XM_017000086.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6997821 c.573+3A>G p.? Chr.1: 214619223-214619223 on Build GRCh38
6541707 c.564G>T p.L188F Chr.1: 214619211-214619211 on Build GRCh38
903736 c.522G>A p.E174E Chr.1: 214619169-214619169 on Build GRCh38
903735 c.505G>T p.E169* Chr.1: 214619152-214619152 on Build GRCh38
4028000 c.532C>T p.R178* Chr.1: 214619179-214619179 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748425553 Chr.1: 214619162-214619162 on Build GRCh38
rs578131699 Chr.1: 214618852-214618853 on Build GRCh38
rs1034391294 Chr.1: 214619157-214619157 on Build GRCh38
rs759584040 Chr.1: 214619082-214619082 on Build GRCh38
rs61732049 Chr.1: 214619209-214619209 on Build GRCh38
rs760951737 Chr.1: 214619180-214619180 on Build GRCh38
rs1022324040 Chr.1: 214619338-214619338 on Build GRCh38
rs768673812 Chr.1: 214619156-214619160 on Build GRCh38
rs929687332 Chr.1: 214619121-214619121 on Build GRCh38
rs755220921 Chr.1: 214619133-214619133 on Build GRCh38
rs144898977 Chr.1: 214619163-214619163 on Build GRCh38
rs781065445 Chr.1: 214619249-214619249 on Build GRCh38
rs576346142 Chr.1: 214619206-214619206 on Build GRCh38
rs1046949436 Chr.1: 214619288-214619288 on Build GRCh38
rs750836467 Chr.1: 214619090-214619090 on Build GRCh38
rs765373633 Chr.1: 214619219-214619219 on Build GRCh38
rs373504409 Chr.1: 214619232-214619232 on Build GRCh38
rs1055515790 Chr.1: 214619328-214619328 on Build GRCh38
rs755059466 Chr.1: 214619214-214619214 on Build GRCh38
rs746873907 Chr.1: 214619233-214619233 on Build GRCh38
rs774953042 Chr.1: 214619194-214619194 on Build GRCh38
rs756648156 Chr.1: 214619094-214619094 on Build GRCh38
rs918342723 Chr.1: 214619115-214619115 on Build GRCh38
rs1024412664 Chr.1: 214619348-214619348 on Build GRCh38
rs752798318 Chr.1: 214619224-214619224 on Build GRCh38
rs189214553 Chr.1: 214618839-214618839 on Build GRCh38
rs998196177 Chr.1: 214618959-214618959 on Build GRCh38
rs572212022 Chr.1: 214619260-214619260 on Build GRCh38
rs992473195 Chr.1: 214619095-214619095 on Build GRCh38
rs751918226 Chr.1: 214619079-214619079 on Build GRCh38
rs1003805669 Chr.1: 214619096-214619096 on Build GRCh38
rs149042046 Chr.1: 214619185-214619185 on Build GRCh38
rs767943545 Chr.1: 214619084-214619084 on Build GRCh38
rs746919842 Chr.1: 214619170-214619170 on Build GRCh38
rs1053514392 Chr.1: 214618935-214618935 on Build GRCh38
rs967416964 Chr.1: 214619189-214619189 on Build GRCh38
rs757152620 Chr.1: 214619236-214619236 on Build GRCh38
rs767706745 Chr.1: 214619200-214619200 on Build GRCh38
rs769320835 Chr.1: 214619191-214619191 on Build GRCh38
rs977708671 Chr.1: 214618885-214618885 on Build GRCh38
rs374684412 Chr.1: 214619083-214619083 on Build GRCh38
rs774400128 Chr.1: 214619031-214619031 on Build GRCh38
rs79890895 Chr.1: 214619330-214619330 on Build GRCh38
rs888717078 Chr.1: 214618903-214618903 on Build GRCh38
rs564982357 Chr.1: 214619347-214619347 on Build GRCh38
rs771457456 Chr.1: 214618941-214618941 on Build GRCh38
rs1005336989 Chr.1: 214619326-214619327 on Build GRCh38
rs927215247 Chr.1: 214619058-214619058 on Build GRCh38
rs910726264 Chr.1: 214619234-214619234 on Build GRCh38
rs994346344 Chr.1: 214618858-214618858 on Build GRCh38
rs942378817 Chr.1: 214618968-214618968 on Build GRCh38
rs2292377 Chr.1: 214619006-214619006 on Build GRCh38
rs1031539967 Chr.1: 214618967-214618967 on Build GRCh38
rs538665660 Chr.1: 214619042-214619042 on Build GRCh38
rs767035015 Chr.1: 214619208-214619208 on Build GRCh38
rs892570700 Chr.1: 214619285-214619285 on Build GRCh38
rs374687775 Chr.1: 214619009-214619009 on Build GRCh38
rs994463747 Chr.1: 214619032-214619032 on Build GRCh38
rs888395950 Chr.1: 214619304-214619304 on Build GRCh38
rs140195013 Chr.1: 214619159-214619159 on Build GRCh38
rs375674714 Chr.1: 214619185-214619186 on Build GRCh38
rs554011682 Chr.1: 214619057-214619057 on Build GRCh38
rs956755673 Chr.1: 214618993-214618993 on Build GRCh38
rs368611046 Chr.1: 214619112-214619113 on Build GRCh38
rs775069080 Chr.1: 214619272-214619272 on Build GRCh38
rs868555756 Chr.1: 214619135-214619135 on Build GRCh38
rs199800194 Chr.1: 214619179-214619179 on Build GRCh38
rs1057308761 Chr.1: 214618837-214618837 on Build GRCh38
rs888263417 Chr.1: 214619067-214619067 on Build GRCh38
rs777632803 Chr.1: 214619167-214619167 on Build GRCh38
rs979958622 Chr.1: 214619350-214619350 on Build GRCh38
rs745824249 Chr.1: 214619255-214619255 on Build GRCh38
rs11120367 Chr.1: 214618937-214618937 on Build GRCh38
rs1048553763 Chr.1: 214618870-214618870 on Build GRCh38
rs541187994 Chr.1: 214619279-214619279 on Build GRCh38
rs2292378 Chr.1: 214618906-214618906 on Build GRCh38
rs200203293 Chr.1: 214619122-214619122 on Build GRCh38
rs762679372 Chr.1: 214619196-214619196 on Build GRCh38
rs754027997 Chr.1: 214619127-214619127 on Build GRCh38
rs143816659 Chr.1: 214619230-214619230 on Build GRCh38

View Full Product Details