Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6624495-6625021 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCHS1
Entrez Gene ID
8642
Gene Name
dachsous cadherin-related 1
Gene Aliases
CDH19, CDH25, CDHR6, FIB1, MVP2, PCDH16, VMLDS1
UniGene
Hs.199850
Gene Chromosome Location
Chr.11: 6621324-6655849 on Build GRCh38
Transcripts
NM_003737.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6461986 c.7147G>A p.V2383I Chr.11: 6624868-6624868 on Build GRCh38
4035807 c.7248T>G p.A2416A Chr.11: 6624767-6624767 on Build GRCh38
7224220 c.7147-9A>G p.? Chr.11: 6624877-6624877 on Build GRCh38
4557297 c.7225G>C p.G2409R Chr.11: 6624790-6624790 on Build GRCh38
3398067 c.7191C>T p.S2397S Chr.11: 6624824-6624824 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781008254 Chr.11: 6624889-6624889 on Build GRCh38
rs773321588 Chr.11: 6624727-6624727 on Build GRCh38
rs1013647566 Chr.11: 6624626-6624626 on Build GRCh38
rs370143518 Chr.11: 6624824-6624824 on Build GRCh38
rs902593712 Chr.11: 6624912-6624912 on Build GRCh38
rs1002097659 Chr.11: 6624581-6624581 on Build GRCh38
rs1028741665 Chr.11: 6624946-6624946 on Build GRCh38
rs764657384 Chr.11: 6624865-6624865 on Build GRCh38
rs756642692 Chr.11: 6624761-6624761 on Build GRCh38
rs763128348 Chr.11: 6624740-6624740 on Build GRCh38
rs975023322 Chr.11: 6624716-6624716 on Build GRCh38
rs1021459802 Chr.11: 6624634-6624634 on Build GRCh38
rs746802857 Chr.11: 6624687-6624687 on Build GRCh38
rs759992482 Chr.11: 6624754-6624754 on Build GRCh38
rs772006722 Chr.11: 6624913-6624920 on Build GRCh38
rs768108958 Chr.11: 6624633-6624633 on Build GRCh38
rs768441388 Chr.11: 6624791-6624791 on Build GRCh38
rs773236358 Chr.11: 6624812-6624812 on Build GRCh38
rs10458926 Chr.11: 6624877-6624877 on Build GRCh38
rs141901540 Chr.11: 6624808-6624808 on Build GRCh38
rs749114081 Chr.11: 6624907-6624907 on Build GRCh38
rs778334140 Chr.11: 6624772-6624772 on Build GRCh38
rs766622753 Chr.11: 6624748-6624748 on Build GRCh38
rs779739540 Chr.11: 6624787-6624787 on Build GRCh38
rs117657616 Chr.11: 6624664-6624664 on Build GRCh38
rs531727583 Chr.11: 6624656-6624656 on Build GRCh38
rs1032330149 Chr.11: 6624924-6624924 on Build GRCh38
rs749959353 Chr.11: 6624868-6624868 on Build GRCh38
rs553904945 Chr.11: 6624875-6624875 on Build GRCh38
rs747329572 Chr.11: 6624893-6624893 on Build GRCh38
rs751240647 Chr.11: 6624881-6624881 on Build GRCh38
rs371931841 Chr.11: 6624689-6624689 on Build GRCh38
rs771075826 Chr.11: 6624908-6624908 on Build GRCh38
rs768634030 Chr.11: 6624708-6624708 on Build GRCh38
rs780922563 Chr.11: 6624792-6624792 on Build GRCh38
rs763992730 Chr.11: 6624649-6624650 on Build GRCh38
rs751693445 Chr.11: 6624604-6624604 on Build GRCh38
rs939823287 Chr.11: 6624800-6624800 on Build GRCh38
rs202152693 Chr.11: 6624680-6624680 on Build GRCh38
rs964940386 Chr.11: 6624692-6624695 on Build GRCh38
rs774526091 Chr.11: 6624913-6624913 on Build GRCh38
rs138322204 Chr.11: 6624844-6624844 on Build GRCh38
rs960835910 Chr.11: 6624624-6624624 on Build GRCh38
rs923407513 Chr.11: 6624726-6624726 on Build GRCh38
rs371690696 Chr.11: 6624757-6624757 on Build GRCh38
rs747935543 Chr.11: 6624897-6624897 on Build GRCh38
rs536782875 Chr.11: 6625002-6625002 on Build GRCh38
rs1038033367 Chr.11: 6624529-6624529 on Build GRCh38
rs777784008 Chr.11: 6624905-6624905 on Build GRCh38
rs368218406 Chr.11: 6624932-6624932 on Build GRCh38
rs759792901 Chr.11: 6624839-6624839 on Build GRCh38
rs764235436 Chr.11: 6624543-6624543 on Build GRCh38
rs953123693 Chr.11: 6624949-6624949 on Build GRCh38
rs754574588 Chr.11: 6624888-6624888 on Build GRCh38
rs551678165 Chr.11: 6624639-6624639 on Build GRCh38
rs964722599 Chr.11: 6624984-6624984 on Build GRCh38
rs763080180 Chr.11: 6624821-6624821 on Build GRCh38
rs898414175 Chr.11: 6624573-6624573 on Build GRCh38
rs769682025 Chr.11: 6624900-6624900 on Build GRCh38
rs749915710 Chr.11: 6624773-6624773 on Build GRCh38
rs757260564 Chr.11: 6624623-6624623 on Build GRCh38
rs373710080 Chr.11: 6624758-6624758 on Build GRCh38
rs746602849 Chr.11: 6624790-6624790 on Build GRCh38
rs933504957 Chr.11: 6624746-6624746 on Build GRCh38
rs780575460 Chr.11: 6624850-6624850 on Build GRCh38
rs758053271 Chr.11: 6624775-6624775 on Build GRCh38
rs975818736 Chr.11: 6624582-6624582 on Build GRCh38
rs367851277 Chr.11: 6624876-6624876 on Build GRCh38
rs751931235 Chr.11: 6624752-6624752 on Build GRCh38
rs769792980 Chr.11: 6624723-6624723 on Build GRCh38
rs949582086 Chr.11: 6624807-6624807 on Build GRCh38
rs921705169 Chr.11: 6624615-6624615 on Build GRCh38
rs1037980967 Chr.11: 6624803-6624803 on Build GRCh38
rs145939548 Chr.11: 6624823-6624823 on Build GRCh38
rs368729057 Chr.11: 6624681-6624681 on Build GRCh38
rs776558735 Chr.11: 6624717-6624717 on Build GRCh38
rs761124968 Chr.11: 6624851-6624851 on Build GRCh38
rs750889492 Chr.11: 6624718-6624718 on Build GRCh38
rs34538572 Chr.11: 6624914-6624915 on Build GRCh38

View Full Product Details