Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 82731868-82732445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM181B
Entrez Gene ID
220382
Gene Name
family with sequence similarity 181 member B
Gene Aliases
-
UniGene
Hs.448218
Gene Chromosome Location
Chr.11: 82732004-82733864 on Build GRCh38
Transcripts
NM_175885.3

SNP Detail

SNP ID Chromosome Location
rs908870635 Chr.11: 82732267-82732267 on Build GRCh38
rs3780 Chr.11: 82732375-82732375 on Build GRCh38
rs779294924 Chr.11: 82732405-82732405 on Build GRCh38
rs75304464 Chr.11: 82732098-82732098 on Build GRCh38
rs200966416 Chr.11: 82732416-82732416 on Build GRCh38
rs922738711 Chr.11: 82731999-82731999 on Build GRCh38
rs904300339 Chr.11: 82732281-82732281 on Build GRCh38
rs1009908657 Chr.11: 82731906-82731906 on Build GRCh38
rs926591476 Chr.11: 82732425-82732425 on Build GRCh38
rs937714457 Chr.11: 82732294-82732294 on Build GRCh38
rs999848748 Chr.11: 82732300-82732300 on Build GRCh38
rs571977398 Chr.11: 82731979-82731979 on Build GRCh38
rs909989226 Chr.11: 82732048-82732048 on Build GRCh38
rs145469223 Chr.11: 82731969-82731969 on Build GRCh38
rs191303825 Chr.11: 82732004-82732004 on Build GRCh38
rs558085460 Chr.11: 82732231-82732234 on Build GRCh38
rs554450973 Chr.11: 82732249-82732249 on Build GRCh38
rs895613997 Chr.11: 82732203-82732203 on Build GRCh38
rs941356297 Chr.11: 82732060-82732060 on Build GRCh38
rs968411309 Chr.11: 82731946-82731946 on Build GRCh38
rs187572073 Chr.11: 82731954-82731954 on Build GRCh38
rs762745287 Chr.11: 82731962-82731962 on Build GRCh38
rs139556739 Chr.11: 82731907-82731907 on Build GRCh38
rs1056984006 Chr.11: 82732171-82732171 on Build GRCh38
rs571456849 Chr.11: 82732064-82732064 on Build GRCh38
rs372639261 Chr.11: 82732240-82732243 on Build GRCh38
rs895664783 Chr.11: 82731896-82731896 on Build GRCh38
rs540955833 Chr.11: 82732394-82732394 on Build GRCh38
rs374151454 Chr.11: 82732163-82732163 on Build GRCh38
rs963093778 Chr.11: 82732411-82732411 on Build GRCh38
rs748478892 Chr.11: 82732406-82732406 on Build GRCh38
rs1042266138 Chr.11: 82732398-82732398 on Build GRCh38
rs985552688 Chr.11: 82732052-82732052 on Build GRCh38
rs772168430 Chr.11: 82732337-82732337 on Build GRCh38
rs964314944 Chr.11: 82731963-82731963 on Build GRCh38
rs866629059 Chr.11: 82732069-82732069 on Build GRCh38
rs572653243 Chr.11: 82732332-82732332 on Build GRCh38
rs750637763 Chr.11: 82732368-82732368 on Build GRCh38
rs896318962 Chr.11: 82731890-82731890 on Build GRCh38
rs1020849745 Chr.11: 82731923-82731923 on Build GRCh38
rs998397717 Chr.11: 82732373-82732373 on Build GRCh38
rs1000847896 Chr.11: 82731961-82731961 on Build GRCh38
rs899514956 Chr.11: 82732370-82732370 on Build GRCh38
rs984360380 Chr.11: 82732260-82732260 on Build GRCh38
rs114353631 Chr.11: 82732409-82732409 on Build GRCh38
rs1018336728 Chr.11: 82732323-82732323 on Build GRCh38
rs559239707 Chr.11: 82732056-82732056 on Build GRCh38
rs983381773 Chr.11: 82732395-82732395 on Build GRCh38
rs982882721 Chr.11: 82732019-82732019 on Build GRCh38
rs757068852 Chr.11: 82732103-82732103 on Build GRCh38
rs1041459607 Chr.11: 82732255-82732255 on Build GRCh38

View Full Product Details