Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6581047-6581555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHD4
Entrez Gene ID
1108
Gene Name
chromodomain helicase DNA binding protein 4
Gene Aliases
CHD-4, Mi-2b, Mi2-BETA
UniGene
Hs.162233
Gene Chromosome Location
Chr.12: 6570082-6607439 on Build GRCh38
Transcripts
NM_001273.3 NM_001297553.1 XM_005253668.3 XM_006718958.1 XM_006718959.1 XM_006718960.1 XM_006718961.2 XM_006718962.1 XM_017018725.1 XM_017018726.1 XM_017018727.1 XM_017018728.1 XM_017018729.1 XM_017018730.1 XM_017018731.1 XM_017018732.1 XM_017018733.1 XM_017018734.1 NR_003012.1

Gene Symbol
SCARNA11
Entrez Gene ID
677780
Gene Name
small Cajal body-specific RNA 11
Gene Aliases
ACA57
UniGene
Hs.689692
Gene Chromosome Location
Chr.12: 6581473-6581609 on Build GRCh38
Transcripts
NM_001273.3 NM_001297553.1 XM_005253668.3 XM_006718958.1 XM_006718959.1 XM_006718960.1 XM_006718961.2 XM_006718962.1 XM_017018725.1 XM_017018726.1 XM_017018727.1 XM_017018728.1 XM_017018729.1 XM_017018730.1 XM_017018731.1 XM_017018732.1 XM_017018733.1 XM_017018734.1 NR_003012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1476829 c.4812T>C p.D1604D Chr.12: 6581141-6581141 on Build GRCh38
6628029 c.4882C>T p.P1628S Chr.12: 6581155-6581155 on Build GRCh38
6627898 c.4774A>G p.I1592V Chr.12: 6581296-6581296 on Build GRCh38
6231750 c.4965A>T p.T1655T Chr.12: 6581072-6581072 on Build GRCh38
4044297 c.4747G>T p.V1583F Chr.12: 6581323-6581323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544032396 Chr.12: 6581462-6581462 on Build GRCh38
rs763767059 Chr.12: 6581080-6581080 on Build GRCh38
rs771161802 Chr.12: 6581421-6581421 on Build GRCh38
rs563783252 Chr.12: 6581485-6581485 on Build GRCh38
rs201417262 Chr.12: 6581167-6581167 on Build GRCh38
rs760291708 Chr.12: 6581191-6581192 on Build GRCh38
rs765159128 Chr.12: 6581407-6581410 on Build GRCh38
rs746810026 Chr.12: 6581335-6581335 on Build GRCh38
rs746720098 Chr.12: 6581157-6581157 on Build GRCh38
rs910538116 Chr.12: 6581532-6581532 on Build GRCh38
rs776901924 Chr.12: 6581455-6581455 on Build GRCh38
rs376011163 Chr.12: 6581305-6581305 on Build GRCh38
rs1051742094 Chr.12: 6581516-6581516 on Build GRCh38
rs769254265 Chr.12: 6581261-6581261 on Build GRCh38
rs565146222 Chr.12: 6581506-6581506 on Build GRCh38
rs754606685 Chr.12: 6581385-6581385 on Build GRCh38
rs761725960 Chr.12: 6581354-6581356 on Build GRCh38
rs750489736 Chr.12: 6581463-6581463 on Build GRCh38
rs771735196 Chr.12: 6581183-6581183 on Build GRCh38
rs749193541 Chr.12: 6581132-6581132 on Build GRCh38
rs761036067 Chr.12: 6581436-6581436 on Build GRCh38
rs193226938 Chr.12: 6581095-6581095 on Build GRCh38
rs756171975 Chr.12: 6581122-6581122 on Build GRCh38
rs765295381 Chr.12: 6581308-6581308 on Build GRCh38
rs752892972 Chr.12: 6581186-6581186 on Build GRCh38
rs200698074 Chr.12: 6581431-6581431 on Build GRCh38
rs370021329 Chr.12: 6581391-6581391 on Build GRCh38
rs759940656 Chr.12: 6581175-6581175 on Build GRCh38
rs746616224 Chr.12: 6581241-6581241 on Build GRCh38
rs766420024 Chr.12: 6581474-6581474 on Build GRCh38
rs775187139 Chr.12: 6581278-6581278 on Build GRCh38
rs201992075 Chr.12: 6581131-6581131 on Build GRCh38
rs750548651 Chr.12: 6581435-6581436 on Build GRCh38
rs762922027 Chr.12: 6581470-6581470 on Build GRCh38
rs771561972 Chr.12: 6581148-6581148 on Build GRCh38
rs142642013 Chr.12: 6581111-6581111 on Build GRCh38
rs371075410 Chr.12: 6581192-6581192 on Build GRCh38
rs776219606 Chr.12: 6581253-6581253 on Build GRCh38
rs752982860 Chr.12: 6581118-6581118 on Build GRCh38
rs138939193 Chr.12: 6581369-6581369 on Build GRCh38
rs781474417 Chr.12: 6581531-6581532 on Build GRCh38
rs763316829 Chr.12: 6581164-6581164 on Build GRCh38
rs764490880 Chr.12: 6581439-6581441 on Build GRCh38
rs780033123 Chr.12: 6581220-6581220 on Build GRCh38
rs200530735 Chr.12: 6581170-6581170 on Build GRCh38
rs1047539372 Chr.12: 6581524-6581524 on Build GRCh38
rs756370515 Chr.12: 6581187-6581187 on Build GRCh38
rs775084830 Chr.12: 6581187-6581192 on Build GRCh38
rs751875901 Chr.12: 6581490-6581492 on Build GRCh38
rs750355009 Chr.12: 6581310-6581310 on Build GRCh38
rs753973569 Chr.12: 6581196-6581196 on Build GRCh38
rs559514209 Chr.12: 6581345-6581345 on Build GRCh38
rs764120768 Chr.12: 6581191-6581191 on Build GRCh38
rs781396205 Chr.12: 6581113-6581113 on Build GRCh38
rs755212825 Chr.12: 6581510-6581510 on Build GRCh38
rs761993543 Chr.12: 6581456-6581456 on Build GRCh38
rs746346870 Chr.12: 6581425-6581425 on Build GRCh38
rs201471909 Chr.12: 6581143-6581143 on Build GRCh38
rs747812828 Chr.12: 6581397-6581397 on Build GRCh38
rs766609805 Chr.12: 6581109-6581109 on Build GRCh38
rs201942144 Chr.12: 6581266-6581266 on Build GRCh38
rs533921472 Chr.12: 6581182-6581183 on Build GRCh38
rs755755550 Chr.12: 6581511-6581511 on Build GRCh38
rs749512768 Chr.12: 6581379-6581379 on Build GRCh38
rs767838481 Chr.12: 6581270-6581270 on Build GRCh38
rs751612749 Chr.12: 6581484-6581484 on Build GRCh38
rs548105495 Chr.12: 6581129-6581129 on Build GRCh38
rs376175590 Chr.12: 6581140-6581140 on Build GRCh38
rs904678204 Chr.12: 6581169-6581169 on Build GRCh38
rs960717003 Chr.12: 6581316-6581316 on Build GRCh38
rs889175642 Chr.12: 6581121-6581121 on Build GRCh38
rs768247783 Chr.12: 6581161-6581161 on Build GRCh38
rs77843719 Chr.12: 6581487-6581487 on Build GRCh38
rs374652277 Chr.12: 6581444-6581444 on Build GRCh38
rs142136215 Chr.12: 6581082-6581082 on Build GRCh38
rs752362997 Chr.12: 6581491-6581491 on Build GRCh38
rs769323293 Chr.12: 6581413-6581413 on Build GRCh38
rs933321388 Chr.12: 6581499-6581499 on Build GRCh38
rs772592474 Chr.12: 6581427-6581427 on Build GRCh38
rs561803347 Chr.12: 6581295-6581295 on Build GRCh38
rs780246528 Chr.12: 6581458-6581459 on Build GRCh38
rs1013169627 Chr.12: 6581312-6581312 on Build GRCh38
rs376819567 Chr.12: 6581265-6581265 on Build GRCh38
rs778917476 Chr.12: 6581199-6581199 on Build GRCh38
rs763846015 Chr.12: 6581193-6581194 on Build GRCh38
rs758439369 Chr.12: 6581317-6581317 on Build GRCh38
rs139854365 Chr.12: 6581517-6581517 on Build GRCh38
rs776309576 Chr.12: 6581163-6581163 on Build GRCh38
rs760839505 Chr.12: 6581273-6581273 on Build GRCh38
rs963534634 Chr.12: 6581446-6581446 on Build GRCh38
rs756243539 Chr.12: 6581100-6581100 on Build GRCh38
rs373615711 Chr.12: 6581251-6581251 on Build GRCh38
rs184412521 Chr.12: 6581418-6581418 on Build GRCh38
rs764314849 Chr.12: 6581278-6581278 on Build GRCh38
rs199773470 Chr.12: 6581174-6581174 on Build GRCh38
rs745862017 Chr.12: 6581205-6581205 on Build GRCh38
rs779943343 Chr.12: 6581332-6581332 on Build GRCh38
rs758257024 Chr.12: 6581219-6581219 on Build GRCh38
rs767926188 Chr.12: 6581185-6581185 on Build GRCh38
rs757358084 Chr.12: 6581301-6581301 on Build GRCh38
rs757088917 Chr.12: 6581198-6581198 on Build GRCh38
rs775864340 Chr.12: 6581272-6581272 on Build GRCh38
rs775280935 Chr.12: 6581155-6581155 on Build GRCh38

View Full Product Details