Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8032758-8033355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXJ2
Entrez Gene ID
55810
Gene Name
forkhead box J2
Gene Aliases
FHX
UniGene
Hs.120844
Gene Chromosome Location
Chr.12: 8032680-8055522 on Build GRCh38
Transcripts
NM_018416.2 XM_011520760.2 XM_011520761.2

SNP Detail

SNP ID Chromosome Location
rs929188158 Chr.12: 8032933-8032933 on Build GRCh38
rs750962300 Chr.12: 8033200-8033200 on Build GRCh38
rs75621466 Chr.12: 8033239-8033239 on Build GRCh38
rs374163772 Chr.12: 8033246-8033246 on Build GRCh38
rs946879351 Chr.12: 8032903-8032903 on Build GRCh38
rs185882887 Chr.12: 8033307-8033307 on Build GRCh38
rs779693945 Chr.12: 8032855-8032855 on Build GRCh38
rs897045200 Chr.12: 8033294-8033294 on Build GRCh38
rs966447505 Chr.12: 8033063-8033063 on Build GRCh38
rs920461362 Chr.12: 8032925-8032925 on Build GRCh38
rs981800187 Chr.12: 8033238-8033238 on Build GRCh38
rs1056165875 Chr.12: 8032939-8032939 on Build GRCh38
rs950759469 Chr.12: 8032808-8032808 on Build GRCh38
rs1032895184 Chr.12: 8033048-8033048 on Build GRCh38
rs937848213 Chr.12: 8032861-8032861 on Build GRCh38
rs968321818 Chr.12: 8033323-8033323 on Build GRCh38
rs983501220 Chr.12: 8032820-8032820 on Build GRCh38
rs750285661 Chr.12: 8033296-8033296 on Build GRCh38
rs749338699 Chr.12: 8033127-8033127 on Build GRCh38
rs923708347 Chr.12: 8032921-8032921 on Build GRCh38
rs996957761 Chr.12: 8033018-8033018 on Build GRCh38
rs1029234175 Chr.12: 8033308-8033308 on Build GRCh38
rs753465352 Chr.12: 8032896-8032896 on Build GRCh38
rs959360943 Chr.12: 8033244-8033244 on Build GRCh38
rs917563502 Chr.12: 8033216-8033216 on Build GRCh38
rs896388976 Chr.12: 8032946-8032946 on Build GRCh38
rs1027716234 Chr.12: 8032782-8032782 on Build GRCh38
rs1019607483 Chr.12: 8032852-8032852 on Build GRCh38
rs968189460 Chr.12: 8033129-8033129 on Build GRCh38
rs1004934935 Chr.12: 8033274-8033274 on Build GRCh38
rs970382611 Chr.12: 8032809-8032809 on Build GRCh38
rs994854143 Chr.12: 8032781-8032781 on Build GRCh38
rs1018908901 Chr.12: 8033057-8033057 on Build GRCh38
rs981853934 Chr.12: 8032824-8032824 on Build GRCh38
rs769887379 Chr.12: 8033217-8033217 on Build GRCh38
rs3741857 Chr.12: 8032955-8032955 on Build GRCh38
rs886945564 Chr.12: 8033258-8033258 on Build GRCh38
rs35297839 Chr.12: 8033257-8033257 on Build GRCh38
rs913534586 Chr.12: 8032793-8032793 on Build GRCh38
rs995325650 Chr.12: 8032993-8032993 on Build GRCh38
rs756566133 Chr.12: 8033085-8033085 on Build GRCh38
rs1005291007 Chr.12: 8033025-8033025 on Build GRCh38
rs763807529 Chr.12: 8032902-8032902 on Build GRCh38
rs970372861 Chr.12: 8033212-8033212 on Build GRCh38
rs181550299 Chr.12: 8033013-8033013 on Build GRCh38
rs759664419 Chr.12: 8033148-8033148 on Build GRCh38
rs947733035 Chr.12: 8033236-8033236 on Build GRCh38
rs923676155 Chr.12: 8033154-8033154 on Build GRCh38
rs898344140 Chr.12: 8032777-8032777 on Build GRCh38
rs778214342 Chr.12: 8033027-8033027 on Build GRCh38
rs771498313 Chr.12: 8033109-8033109 on Build GRCh38
rs1044977044 Chr.12: 8032958-8032958 on Build GRCh38
rs1027682257 Chr.12: 8033012-8033012 on Build GRCh38
rs188383230 Chr.12: 8033328-8033328 on Build GRCh38
rs377500663 Chr.12: 8032989-8032989 on Build GRCh38
rs992318697 Chr.12: 8033215-8033215 on Build GRCh38
rs1047636383 Chr.12: 8033332-8033332 on Build GRCh38
rs979151477 Chr.12: 8032916-8032916 on Build GRCh38
rs911844945 Chr.12: 8033335-8033335 on Build GRCh38
rs764308105 Chr.12: 8032884-8032884 on Build GRCh38
rs929158051 Chr.12: 8033302-8033302 on Build GRCh38
rs3741858 Chr.12: 8033322-8033322 on Build GRCh38
rs1041103696 Chr.12: 8033008-8033008 on Build GRCh38
rs941324102 Chr.12: 8033064-8033064 on Build GRCh38
rs1040871852 Chr.12: 8033290-8033290 on Build GRCh38
rs934976869 Chr.12: 8032924-8032924 on Build GRCh38
rs1047646205 Chr.12: 8032949-8032949 on Build GRCh38

View Full Product Details